Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99499946-99500446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF394
Entrez Gene ID
84124
Gene Name
zinc finger protein 394
Gene Aliases
ZKSCAN14, ZSCAN46
UniGene
Hs.386324
Gene Chromosome Location
Chr.7: 99486519-99500302 on Build GRCh38
Transcripts
NM_032164.2 XM_017012710.1 XM_017012711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829817 c.84G>T p.A28A Chr.7: 99500010-99500010 on Build GRCh38
4932343 c.110T>C p.L37S Chr.7: 99499984-99499984 on Build GRCh38
4006805 c.138C>T p.P46P Chr.7: 99499956-99499956 on Build GRCh38
2867243 c.18C>T p.T6T Chr.7: 99500076-99500076 on Build GRCh38
421832 c.112C>T p.P38S Chr.7: 99499982-99499982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962254292 Chr.7: 99500203-99500203 on Build GRCh38
rs770543901 Chr.7: 99500098-99500098 on Build GRCh38
rs993524865 Chr.7: 99500177-99500177 on Build GRCh38
rs960589902 Chr.7: 99499982-99499982 on Build GRCh38
rs377751850 Chr.7: 99500139-99500139 on Build GRCh38
rs1038378238 Chr.7: 99500300-99500300 on Build GRCh38
rs1051646615 Chr.7: 99500278-99500278 on Build GRCh38
rs769530636 Chr.7: 99500004-99500004 on Build GRCh38
rs1002981525 Chr.7: 99500372-99500372 on Build GRCh38
rs560686849 Chr.7: 99499980-99499980 on Build GRCh38
rs373393024 Chr.7: 99500134-99500134 on Build GRCh38
rs760655912 Chr.7: 99500125-99500125 on Build GRCh38
rs910978602 Chr.7: 99500396-99500396 on Build GRCh38
rs756522300 Chr.7: 99500076-99500076 on Build GRCh38
rs908069184 Chr.7: 99499989-99499989 on Build GRCh38
rs80010515 Chr.7: 99500397-99500397 on Build GRCh38
rs971506478 Chr.7: 99500199-99500199 on Build GRCh38
rs770440444 Chr.7: 99500012-99500012 on Build GRCh38
rs1010029389 Chr.7: 99500091-99500091 on Build GRCh38
rs747520068 Chr.7: 99499991-99499991 on Build GRCh38
rs151209671 Chr.7: 99500071-99500071 on Build GRCh38
rs546288043 Chr.7: 99500219-99500219 on Build GRCh38
rs150978993 Chr.7: 99500080-99500080 on Build GRCh38
rs766477445 Chr.7: 99500063-99500063 on Build GRCh38
rs1047278085 Chr.7: 99500333-99500333 on Build GRCh38
rs763225331 Chr.7: 99500138-99500138 on Build GRCh38
rs760498183 Chr.7: 99500025-99500025 on Build GRCh38
rs1051165037 Chr.7: 99500101-99500101 on Build GRCh38
rs898747299 Chr.7: 99500146-99500146 on Build GRCh38
rs1035215515 Chr.7: 99500383-99500383 on Build GRCh38
rs754656854 Chr.7: 99500346-99500346 on Build GRCh38
rs774319525 Chr.7: 99499967-99499967 on Build GRCh38
rs889809313 Chr.7: 99500105-99500105 on Build GRCh38
rs778432932 Chr.7: 99500100-99500100 on Build GRCh38
rs759428661 Chr.7: 99499973-99499973 on Build GRCh38
rs1014905679 Chr.7: 99500359-99500359 on Build GRCh38
rs904351336 Chr.7: 99500309-99500309 on Build GRCh38
rs570752672 Chr.7: 99500289-99500289 on Build GRCh38
rs117398779 Chr.7: 99500216-99500216 on Build GRCh38
rs1034791276 Chr.7: 99500149-99500149 on Build GRCh38
rs761746689 Chr.7: 99500031-99500031 on Build GRCh38
rs1025773713 Chr.7: 99500186-99500186 on Build GRCh38
rs748527102 Chr.7: 99499998-99499998 on Build GRCh38
rs962307736 Chr.7: 99500368-99500368 on Build GRCh38
rs977974698 Chr.7: 99500257-99500257 on Build GRCh38
rs1006881492 Chr.7: 99500107-99500107 on Build GRCh38
rs776357193 Chr.7: 99500135-99500135 on Build GRCh38
rs1021968528 Chr.7: 99500419-99500419 on Build GRCh38
rs775430877 Chr.7: 99500115-99500115 on Build GRCh38
rs895849866 Chr.7: 99500354-99500354 on Build GRCh38
rs142219532 Chr.7: 99500007-99500007 on Build GRCh38
rs994389616 Chr.7: 99500330-99500330 on Build GRCh38
rs1042242706 Chr.7: 99500059-99500059 on Build GRCh38
rs765397392 Chr.7: 99500030-99500030 on Build GRCh38
rs548926619 Chr.7: 99500096-99500096 on Build GRCh38
rs117567365 Chr.7: 99500328-99500328 on Build GRCh38
rs758528701 Chr.7: 99499985-99499985 on Build GRCh38
rs573972119 Chr.7: 99500358-99500359 on Build GRCh38
rs745389817 Chr.7: 99500103-99500103 on Build GRCh38
rs767291882 Chr.7: 99499975-99499975 on Build GRCh38
rs889441941 Chr.7: 99500048-99500048 on Build GRCh38
rs755608809 Chr.7: 99499997-99499997 on Build GRCh38
rs1014263340 Chr.7: 99500355-99500355 on Build GRCh38
rs953566064 Chr.7: 99500391-99500391 on Build GRCh38
rs773797023 Chr.7: 99500013-99500013 on Build GRCh38
rs534886366 Chr.7: 99500292-99500292 on Build GRCh38
rs553108923 Chr.7: 99500303-99500303 on Build GRCh38
rs554577201 Chr.7: 99500405-99500405 on Build GRCh38
rs139549797 Chr.7: 99500247-99500247 on Build GRCh38
rs1049478232 Chr.7: 99500238-99500238 on Build GRCh38
rs763126461 Chr.7: 99500054-99500054 on Build GRCh38
rs990491365 Chr.7: 99500231-99500231 on Build GRCh38
rs752549935 Chr.7: 99499979-99499979 on Build GRCh38
rs770446007 Chr.7: 99500276-99500276 on Build GRCh38
rs967959792 Chr.7: 99500249-99500249 on Build GRCh38
rs772938523 Chr.7: 99500006-99500006 on Build GRCh38
rs548582434 Chr.7: 99500193-99500193 on Build GRCh38
rs372313159 Chr.7: 99500066-99500066 on Build GRCh38
rs891723692 Chr.7: 99500265-99500265 on Build GRCh38
rs1002057189 Chr.7: 99500314-99500314 on Build GRCh38
rs201107533 Chr.7: 99499990-99499990 on Build GRCh38
rs907402715 Chr.7: 99500344-99500344 on Build GRCh38
rs189150179 Chr.7: 99500110-99500110 on Build GRCh38
rs923788623 Chr.7: 99500264-99500264 on Build GRCh38
rs959326260 Chr.7: 99500227-99500227 on Build GRCh38
rs759499978 Chr.7: 99500018-99500018 on Build GRCh38
rs1040961740 Chr.7: 99500067-99500067 on Build GRCh38
rs372558510 Chr.7: 99500019-99500019 on Build GRCh38
rs530100126 Chr.7: 99500185-99500185 on Build GRCh38
rs750638228 Chr.7: 99500053-99500053 on Build GRCh38
rs534639386 Chr.7: 99500418-99500418 on Build GRCh38
rs182242623 Chr.7: 99500169-99500169 on Build GRCh38
rs994342077 Chr.7: 99500170-99500170 on Build GRCh38
rs778681546 Chr.7: 99500034-99500034 on Build GRCh38
rs575929664 Chr.7: 99500410-99500410 on Build GRCh38
rs200460121 Chr.7: 99500022-99500022 on Build GRCh38
rs973634007 Chr.7: 99500371-99500371 on Build GRCh38
rs114235151 Chr.7: 99500218-99500218 on Build GRCh38
rs1042891606 Chr.7: 99500287-99500287 on Build GRCh38
rs911303773 Chr.7: 99500284-99500284 on Build GRCh38
rs931075009 Chr.7: 99500223-99500223 on Build GRCh38
rs750433022 Chr.7: 99499984-99499984 on Build GRCh38

View Full Product Details