Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100602928-100603439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCOLCE
Entrez Gene ID
5118
Gene Name
procollagen C-endopeptidase enhancer
Gene Aliases
PCPE, PCPE-1, PCPE1
UniGene
Hs.202097
Gene Chromosome Location
Chr.7: 100602259-100608175 on Build GRCh38
Transcripts
NM_002593.3 NR_038910.1

Gene Symbol
PCOLCE-AS1
Entrez Gene ID
100129845
Gene Name
PCOLCE antisense RNA 1
Gene Aliases
-
UniGene
Hs.714123
Gene Chromosome Location
Chr.7: 100589401-100604038 on Build GRCh38
Transcripts
NM_002593.3 NR_038910.1

SNP Detail

SNP ID Chromosome Location
rs763571474 Chr.7: 100603381-100603381 on Build GRCh38
rs987262827 Chr.7: 100602981-100602981 on Build GRCh38
rs187975898 Chr.7: 100603390-100603390 on Build GRCh38
rs192208025 Chr.7: 100603414-100603414 on Build GRCh38
rs537134886 Chr.7: 100603163-100603163 on Build GRCh38
rs543540108 Chr.7: 100603413-100603413 on Build GRCh38
rs746584613 Chr.7: 100603106-100603106 on Build GRCh38
rs951976860 Chr.7: 100603352-100603352 on Build GRCh38
rs780578065 Chr.7: 100603209-100603209 on Build GRCh38
rs920744665 Chr.7: 100602953-100602953 on Build GRCh38
rs1031189020 Chr.7: 100603183-100603183 on Build GRCh38
rs1043590384 Chr.7: 100603127-100603127 on Build GRCh38
rs202228979 Chr.7: 100603051-100603052 on Build GRCh38
rs571005264 Chr.7: 100603102-100603102 on Build GRCh38
rs939143147 Chr.7: 100603070-100603070 on Build GRCh38
rs577752127 Chr.7: 100603210-100603210 on Build GRCh38
rs1043376652 Chr.7: 100603114-100603114 on Build GRCh38
rs1056598031 Chr.7: 100603092-100603092 on Build GRCh38
rs995922671 Chr.7: 100603333-100603333 on Build GRCh38
rs769760439 Chr.7: 100603380-100603380 on Build GRCh38
rs34838915 Chr.7: 100603196-100603197 on Build GRCh38
rs892186443 Chr.7: 100603073-100603073 on Build GRCh38
rs771425848 Chr.7: 100603159-100603159 on Build GRCh38
rs1027669221 Chr.7: 100603176-100603176 on Build GRCh38
rs554782186 Chr.7: 100603371-100603371 on Build GRCh38
rs530283894 Chr.7: 100603005-100603005 on Build GRCh38
rs964110964 Chr.7: 100603310-100603310 on Build GRCh38
rs920505014 Chr.7: 100603045-100603045 on Build GRCh38
rs930528700 Chr.7: 100603049-100603049 on Build GRCh38
rs536422704 Chr.7: 100603343-100603343 on Build GRCh38
rs373535996 Chr.7: 100603186-100603186 on Build GRCh38
rs747663245 Chr.7: 100602992-100602992 on Build GRCh38
rs752008401 Chr.7: 100603392-100603392 on Build GRCh38
rs911599301 Chr.7: 100603046-100603046 on Build GRCh38
rs905017256 Chr.7: 100603130-100603130 on Build GRCh38
rs570820111 Chr.7: 100603226-100603226 on Build GRCh38
rs2142104 Chr.7: 100602971-100602971 on Build GRCh38
rs1014236328 Chr.7: 100603304-100603304 on Build GRCh38
rs907683817 Chr.7: 100603054-100603054 on Build GRCh38
rs534707945 Chr.7: 100603231-100603231 on Build GRCh38
rs974726881 Chr.7: 100603032-100603032 on Build GRCh38
rs538523505 Chr.7: 100603302-100603302 on Build GRCh38
rs1026904880 Chr.7: 100603350-100603350 on Build GRCh38
rs142787996 Chr.7: 100603044-100603044 on Build GRCh38
rs2686815 Chr.7: 100603051-100603051 on Build GRCh38
rs964372967 Chr.7: 100603015-100603015 on Build GRCh38
rs865990725 Chr.7: 100603338-100603338 on Build GRCh38
rs553992601 Chr.7: 100603277-100603277 on Build GRCh38

View Full Product Details