Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100720708-100721208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPO
Entrez Gene ID
2056
Gene Name
erythropoietin
Gene Aliases
EP, MVCD2
UniGene
Hs.2303
Gene Chromosome Location
Chr.7: 100720800-100723700 on Build GRCh38
Transcripts
NM_000799.2

SNP Detail

SNP ID Chromosome Location
rs746962384 Chr.7: 100720969-100720969 on Build GRCh38
rs183591549 Chr.7: 100721126-100721126 on Build GRCh38
rs369468201 Chr.7: 100721003-100721003 on Build GRCh38
rs187459673 Chr.7: 100721136-100721136 on Build GRCh38
rs933864666 Chr.7: 100720789-100720789 on Build GRCh38
rs997834614 Chr.7: 100721179-100721179 on Build GRCh38
rs543789146 Chr.7: 100720768-100720768 on Build GRCh38
rs768734974 Chr.7: 100721013-100721014 on Build GRCh38
rs768536667 Chr.7: 100720972-100720972 on Build GRCh38
rs753057539 Chr.7: 100721028-100721028 on Build GRCh38
rs1039890761 Chr.7: 100720999-100720999 on Build GRCh38
rs1031173251 Chr.7: 100721112-100721112 on Build GRCh38
rs529760857 Chr.7: 100721115-100721115 on Build GRCh38
rs911147235 Chr.7: 100720844-100720844 on Build GRCh38
rs990497132 Chr.7: 100720734-100720734 on Build GRCh38
rs769398632 Chr.7: 100721011-100721011 on Build GRCh38
rs540884608 Chr.7: 100721019-100721019 on Build GRCh38
rs977864253 Chr.7: 100721060-100721060 on Build GRCh38
rs547772793 Chr.7: 100721120-100721120 on Build GRCh38
rs376713532 Chr.7: 100720809-100720809 on Build GRCh38
rs542975821 Chr.7: 100720819-100720840 on Build GRCh38
rs978272210 Chr.7: 100720764-100720764 on Build GRCh38
rs958118037 Chr.7: 100720801-100720801 on Build GRCh38
rs868861841 Chr.7: 100721040-100721040 on Build GRCh38
rs1021946906 Chr.7: 100721029-100721029 on Build GRCh38
rs914950945 Chr.7: 100720738-100720738 on Build GRCh38
rs776099080 Chr.7: 100720986-100720986 on Build GRCh38
rs777893099 Chr.7: 100720939-100720939 on Build GRCh38
rs566438241 Chr.7: 100721150-100721150 on Build GRCh38
rs771913982 Chr.7: 100720961-100720961 on Build GRCh38
rs779789360 Chr.7: 100720964-100720964 on Build GRCh38
rs916583344 Chr.7: 100720887-100720887 on Build GRCh38
rs141701177 Chr.7: 100720941-100720941 on Build GRCh38
rs942894192 Chr.7: 100721147-100721147 on Build GRCh38
rs761220295 Chr.7: 100721033-100721033 on Build GRCh38
rs559371377 Chr.7: 100721057-100721057 on Build GRCh38
rs757521071 Chr.7: 100720945-100720945 on Build GRCh38
rs372062547 Chr.7: 100721014-100721014 on Build GRCh38
rs1048312896 Chr.7: 100720977-100720977 on Build GRCh38
rs756641457 Chr.7: 100720935-100720935 on Build GRCh38
rs888155162 Chr.7: 100720954-100720954 on Build GRCh38
rs759840144 Chr.7: 100721025-100721025 on Build GRCh38
rs776523447 Chr.7: 100720973-100720973 on Build GRCh38
rs1050932151 Chr.7: 100720790-100720790 on Build GRCh38
rs936491288 Chr.7: 100721134-100721134 on Build GRCh38
rs562990454 Chr.7: 100721124-100721124 on Build GRCh38
rs576397148 Chr.7: 100720761-100720761 on Build GRCh38
rs900938585 Chr.7: 100721151-100721151 on Build GRCh38
rs745609348 Chr.7: 100720951-100720951 on Build GRCh38
rs761765700 Chr.7: 100720813-100720813 on Build GRCh38
rs774652091 Chr.7: 100721018-100721019 on Build GRCh38
rs1020652113 Chr.7: 100720748-100720748 on Build GRCh38
rs759518255 Chr.7: 100720989-100720989 on Build GRCh38
rs910043789 Chr.7: 100721145-100721145 on Build GRCh38
rs535187285 Chr.7: 100721158-100721158 on Build GRCh38
rs371400417 Chr.7: 100720735-100720735 on Build GRCh38
rs967188034 Chr.7: 100720767-100720767 on Build GRCh38
rs967803415 Chr.7: 100720746-100720746 on Build GRCh38
rs866747830 Chr.7: 100720987-100720987 on Build GRCh38
rs746847015 Chr.7: 100720926-100720926 on Build GRCh38
rs1048551551 Chr.7: 100720952-100720952 on Build GRCh38
rs750459802 Chr.7: 100721181-100721181 on Build GRCh38
rs34144627 Chr.7: 100720926-100720927 on Build GRCh38
rs546879710 Chr.7: 100721167-100721167 on Build GRCh38

View Full Product Details