Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158294434-158294952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002784.1 XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002784.1 XM_017002785.1 NM_001765.2 XM_005245579.4

SNP Detail

SNP ID Chromosome Location
rs950498476 Chr.1: 158294787-158294787 on Build GRCh38
rs558012591 Chr.1: 158294616-158294616 on Build GRCh38
rs977127780 Chr.1: 158294601-158294601 on Build GRCh38
rs745723875 Chr.1: 158294843-158294843 on Build GRCh38
rs771217801 Chr.1: 158294798-158294798 on Build GRCh38
rs150910090 Chr.1: 158294641-158294641 on Build GRCh38
rs758528311 Chr.1: 158294670-158294670 on Build GRCh38
rs907234420 Chr.1: 158294834-158294834 on Build GRCh38
rs1011717696 Chr.1: 158294919-158294919 on Build GRCh38
rs922958887 Chr.1: 158294630-158294630 on Build GRCh38
rs966995429 Chr.1: 158294596-158294596 on Build GRCh38
rs58054630 Chr.1: 158294854-158294854 on Build GRCh38
rs991876095 Chr.1: 158294464-158294464 on Build GRCh38
rs562971518 Chr.1: 158294773-158294773 on Build GRCh38
rs984453825 Chr.1: 158294518-158294518 on Build GRCh38
rs1021833504 Chr.1: 158294571-158294571 on Build GRCh38
rs974695727 Chr.1: 158294929-158294929 on Build GRCh38
rs1021805369 Chr.1: 158294925-158294925 on Build GRCh38
rs184101700 Chr.1: 158294916-158294916 on Build GRCh38
rs551373367 Chr.1: 158294824-158294824 on Build GRCh38
rs763524839 Chr.1: 158294661-158294661 on Build GRCh38
rs933115587 Chr.1: 158294638-158294638 on Build GRCh38
rs994333719 Chr.1: 158294783-158294783 on Build GRCh38
rs530262109 Chr.1: 158294801-158294801 on Build GRCh38
rs1047038422 Chr.1: 158294802-158294802 on Build GRCh38
rs777677034 Chr.1: 158294710-158294710 on Build GRCh38
rs1001814127 Chr.1: 158294709-158294709 on Build GRCh38
rs1021666092 Chr.1: 158294678-158294678 on Build GRCh38
rs941853246 Chr.1: 158294711-158294711 on Build GRCh38
rs867703926 Chr.1: 158294487-158294487 on Build GRCh38
rs542331883 Chr.1: 158294848-158294848 on Build GRCh38
rs781548717 Chr.1: 158294812-158294812 on Build GRCh38
rs894608162 Chr.1: 158294918-158294918 on Build GRCh38
rs905842705 Chr.1: 158294688-158294688 on Build GRCh38
rs1039340467 Chr.1: 158294612-158294612 on Build GRCh38
rs929054060 Chr.1: 158294488-158294488 on Build GRCh38
rs1047560169 Chr.1: 158294462-158294462 on Build GRCh38
rs964702260 Chr.1: 158294888-158294888 on Build GRCh38
rs908896462 Chr.1: 158294877-158294877 on Build GRCh38
rs1037585436 Chr.1: 158294723-158294723 on Build GRCh38
rs929235351 Chr.1: 158294779-158294779 on Build GRCh38
rs943323599 Chr.1: 158294543-158294543 on Build GRCh38
rs1025758808 Chr.1: 158294784-158294784 on Build GRCh38
rs901688963 Chr.1: 158294626-158294626 on Build GRCh38
rs543611447 Chr.1: 158294736-158294736 on Build GRCh38
rs545814946 Chr.1: 158294530-158294530 on Build GRCh38
rs1011661418 Chr.1: 158294671-158294671 on Build GRCh38
rs986326495 Chr.1: 158294659-158294659 on Build GRCh38

View Full Product Details