Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159302565-159303075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER1A
Entrez Gene ID
2205
Gene Name
Fc fragment of IgE receptor Ia
Gene Aliases
FCE1A, FcERI
UniGene
Hs.897
Gene Chromosome Location
Chr.1: 159283888-159308224 on Build GRCh38
Transcripts
NM_002001.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6310819 c.76+1G>T p.? Chr.1: 159302875-159302875 on Build GRCh38
6058761 c.65G>T p.G22V Chr.1: 159302863-159302863 on Build GRCh38
3476464 c.61G>A p.D21N Chr.1: 159302859-159302859 on Build GRCh38
3399871 c.67G>A p.V23M Chr.1: 159302865-159302865 on Build GRCh38
897995 c.73G>A p.A25T Chr.1: 159302871-159302871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762834291 Chr.1: 159302824-159302824 on Build GRCh38
rs763791207 Chr.1: 159302879-159302879 on Build GRCh38
rs1001649423 Chr.1: 159302690-159302690 on Build GRCh38
rs971990972 Chr.1: 159302946-159302946 on Build GRCh38
rs1029062049 Chr.1: 159302954-159302954 on Build GRCh38
rs1041970903 Chr.1: 159302994-159302994 on Build GRCh38
rs777195714 Chr.1: 159302833-159302833 on Build GRCh38
rs749102254 Chr.1: 159302845-159302845 on Build GRCh38
rs977228370 Chr.1: 159303011-159303011 on Build GRCh38
rs973066330 Chr.1: 159302626-159302626 on Build GRCh38
rs188572451 Chr.1: 159302832-159302832 on Build GRCh38
rs563285584 Chr.1: 159302855-159302855 on Build GRCh38
rs769647822 Chr.1: 159302834-159302834 on Build GRCh38
rs984627284 Chr.1: 159302902-159302902 on Build GRCh38
rs561675998 Chr.1: 159302821-159302821 on Build GRCh38
rs142162478 Chr.1: 159302865-159302865 on Build GRCh38
rs750868238 Chr.1: 159302818-159302818 on Build GRCh38
rs752365661 Chr.1: 159302803-159302803 on Build GRCh38
rs76953025 Chr.1: 159302604-159302604 on Build GRCh38
rs1044799640 Chr.1: 159302659-159302659 on Build GRCh38
rs949076384 Chr.1: 159302949-159302949 on Build GRCh38
rs201427609 Chr.1: 159302806-159302806 on Build GRCh38
rs987266623 Chr.1: 159302721-159302721 on Build GRCh38
rs3819246 Chr.1: 159302947-159302947 on Build GRCh38
rs370235095 Chr.1: 159302624-159302624 on Build GRCh38
rs758933299 Chr.1: 159302820-159302820 on Build GRCh38
rs967150891 Chr.1: 159302979-159302979 on Build GRCh38
rs572416083 Chr.1: 159302994-159302994 on Build GRCh38
rs925850066 Chr.1: 159303034-159303034 on Build GRCh38
rs3845626 Chr.1: 159302618-159302618 on Build GRCh38
rs143021708 Chr.1: 159302838-159302838 on Build GRCh38
rs138086620 Chr.1: 159303032-159303032 on Build GRCh38
rs767709769 Chr.1: 159302919-159302919 on Build GRCh38
rs151155086 Chr.1: 159302864-159302864 on Build GRCh38
rs543210024 Chr.1: 159302747-159302747 on Build GRCh38
rs770869369 Chr.1: 159302852-159302852 on Build GRCh38
rs1028716556 Chr.1: 159302642-159302642 on Build GRCh38
rs550191583 Chr.1: 159302996-159302996 on Build GRCh38
rs766892309 Chr.1: 159302910-159302910 on Build GRCh38
rs752033444 Chr.1: 159302913-159302913 on Build GRCh38
rs200624480 Chr.1: 159302628-159302628 on Build GRCh38
rs752912017 Chr.1: 159302920-159302920 on Build GRCh38
rs12119226 Chr.1: 159302974-159302974 on Build GRCh38
rs774203947 Chr.1: 159302854-159302854 on Build GRCh38
rs531574535 Chr.1: 159302746-159302746 on Build GRCh38
rs970950954 Chr.1: 159302587-159302587 on Build GRCh38
rs147014787 Chr.1: 159302973-159302973 on Build GRCh38
rs551494976 Chr.1: 159302900-159302900 on Build GRCh38
rs569644449 Chr.1: 159302938-159302938 on Build GRCh38
rs893391830 Chr.1: 159302746-159302748 on Build GRCh38
rs755047262 Chr.1: 159302916-159302916 on Build GRCh38
rs528286521 Chr.1: 159302740-159302740 on Build GRCh38
rs140386098 Chr.1: 159302951-159302951 on Build GRCh38
rs145144273 Chr.1: 159302858-159302858 on Build GRCh38
rs758446481 Chr.1: 159303029-159303029 on Build GRCh38
rs906077875 Chr.1: 159302660-159302660 on Build GRCh38
rs977300400 Chr.1: 159302765-159302765 on Build GRCh38
rs368188084 Chr.1: 159302956-159302956 on Build GRCh38
rs529137742 Chr.1: 159302825-159302825 on Build GRCh38
rs762121643 Chr.1: 159302805-159302805 on Build GRCh38
rs773625724 Chr.1: 159302897-159302897 on Build GRCh38
rs953261461 Chr.1: 159302693-159302693 on Build GRCh38
rs184864413 Chr.1: 159302625-159302625 on Build GRCh38
rs901875658 Chr.1: 159303024-159303024 on Build GRCh38
rs1054570004 Chr.1: 159302968-159302968 on Build GRCh38

View Full Product Details