Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160216728-160217226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8
Entrez Gene ID
50717
Gene Name
DDB1 and CUL4 associated factor 8
Gene Aliases
GAN2, H326, WDR42A
UniGene
Hs.632447
Gene Chromosome Location
Chr.1: 160215715-160262560 on Build GRCh38
Transcripts
NM_015726.3 NR_028103.1 NR_028104.1

SNP Detail

SNP ID Chromosome Location
rs2696 Chr.1: 160216884-160216884 on Build GRCh38
rs937014346 Chr.1: 160217127-160217127 on Build GRCh38
rs779045417 Chr.1: 160217055-160217055 on Build GRCh38
rs571161821 Chr.1: 160217099-160217100 on Build GRCh38
rs16831617 Chr.1: 160216983-160216983 on Build GRCh38
rs1037899827 Chr.1: 160217047-160217047 on Build GRCh38
rs751338030 Chr.1: 160216809-160216809 on Build GRCh38
rs532351636 Chr.1: 160216761-160216761 on Build GRCh38
rs967518683 Chr.1: 160216959-160216959 on Build GRCh38
rs549502921 Chr.1: 160217102-160217102 on Build GRCh38
rs1034229545 Chr.1: 160216873-160216873 on Build GRCh38
rs954369926 Chr.1: 160216878-160216878 on Build GRCh38
rs180997028 Chr.1: 160216935-160216935 on Build GRCh38
rs1029991808 Chr.1: 160216841-160216841 on Build GRCh38
rs1032548994 Chr.1: 160217170-160217170 on Build GRCh38
rs1053553 Chr.1: 160216900-160216900 on Build GRCh38
rs74125505 Chr.1: 160216907-160216907 on Build GRCh38
rs900341534 Chr.1: 160217061-160217061 on Build GRCh38
rs755349841 Chr.1: 160217020-160217020 on Build GRCh38
rs979624694 Chr.1: 160217116-160217116 on Build GRCh38
rs1006062637 Chr.1: 160217039-160217039 on Build GRCh38
rs11265356 Chr.1: 160217103-160217103 on Build GRCh38
rs1010293279 Chr.1: 160217104-160217104 on Build GRCh38
rs995800921 Chr.1: 160216798-160216798 on Build GRCh38
rs994219006 Chr.1: 160216786-160216786 on Build GRCh38
rs1011440438 Chr.1: 160216908-160216908 on Build GRCh38
rs16831619 Chr.1: 160217011-160217011 on Build GRCh38
rs773943463 Chr.1: 160216909-160216909 on Build GRCh38
rs1047827307 Chr.1: 160217008-160217008 on Build GRCh38
rs925564088 Chr.1: 160216956-160216956 on Build GRCh38
rs1055378771 Chr.1: 160216962-160216962 on Build GRCh38
rs754225028 Chr.1: 160216961-160216961 on Build GRCh38
rs1001381055 Chr.1: 160216864-160216864 on Build GRCh38
rs527400720 Chr.1: 160217013-160217013 on Build GRCh38
rs748398683 Chr.1: 160217100-160217100 on Build GRCh38
rs572525144 Chr.1: 160216767-160216767 on Build GRCh38
rs906836513 Chr.1: 160217114-160217114 on Build GRCh38
rs959516636 Chr.1: 160217173-160217173 on Build GRCh38
rs953362916 Chr.1: 160217100-160217100 on Build GRCh38
rs912953032 Chr.1: 160216932-160216932 on Build GRCh38
rs949348019 Chr.1: 160216986-160216986 on Build GRCh38
rs373432093 Chr.1: 160217101-160217101 on Build GRCh38
rs537828874 Chr.1: 160217106-160217106 on Build GRCh38
rs377530193 Chr.1: 160217107-160217108 on Build GRCh38
rs906890492 Chr.1: 160216845-160216845 on Build GRCh38
rs966919946 Chr.1: 160216934-160216934 on Build GRCh38
rs544135623 Chr.1: 160216844-160216844 on Build GRCh38
rs1045430567 Chr.1: 160216834-160216835 on Build GRCh38
rs893092582 Chr.1: 160217179-160217179 on Build GRCh38
rs74125506 Chr.1: 160217109-160217109 on Build GRCh38
rs1055628998 Chr.1: 160217136-160217136 on Build GRCh38

View Full Product Details