Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 104184029-104184578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC5
Entrez Gene ID
5001
Gene Name
origin recognition complex subunit 5
Gene Aliases
ORC5L, ORC5P, ORC5T, PPP1R117
UniGene
Hs.432948
Gene Chromosome Location
Chr.7: 104126341-104208047 on Build GRCh38
Transcripts
NM_002553.3 NM_181747.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
376116 c.733+1G>T p.? Chr.7: 104184122-104184122 on Build GRCh38
1150525 c.734-1G>T p.? Chr.7: 104184034-104184034 on Build GRCh38
3876871 c.718G>A p.V240M Chr.7: 104184138-104184138 on Build GRCh38
3876870 c.718G>A p.V240M Chr.7: 104184138-104184138 on Build GRCh38
744297 c.734-1G>T p.? Chr.7: 104184034-104184034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775054447 Chr.7: 104184055-104184055 on Build GRCh38
rs773907332 Chr.7: 104184171-104184171 on Build GRCh38
rs562286245 Chr.7: 104184275-104184275 on Build GRCh38
rs968666029 Chr.7: 104184432-104184432 on Build GRCh38
rs962766586 Chr.7: 104184244-104184244 on Build GRCh38
rs386716399 Chr.7: 104184272-104184276 on Build GRCh38
rs774940496 Chr.7: 104184154-104184154 on Build GRCh38
rs1006712468 Chr.7: 104184496-104184496 on Build GRCh38
rs189732175 Chr.7: 104184515-104184515 on Build GRCh38
rs756082614 Chr.7: 104184224-104184224 on Build GRCh38
rs186620370 Chr.7: 104184437-104184437 on Build GRCh38
rs781323352 Chr.7: 104184107-104184107 on Build GRCh38
rs766936316 Chr.7: 104184174-104184174 on Build GRCh38
rs544721054 Chr.7: 104184215-104184215 on Build GRCh38
rs2307410 Chr.7: 104184113-104184113 on Build GRCh38
rs539519347 Chr.7: 104184402-104184402 on Build GRCh38
rs550922470 Chr.7: 104184344-104184344 on Build GRCh38
rs745710374 Chr.7: 104184139-104184139 on Build GRCh38
rs7807929 Chr.7: 104184546-104184546 on Build GRCh38
rs763599138 Chr.7: 104184058-104184058 on Build GRCh38
rs756426224 Chr.7: 104184210-104184210 on Build GRCh38
rs369602537 Chr.7: 104184325-104184325 on Build GRCh38
rs976926146 Chr.7: 104184474-104184474 on Build GRCh38
rs762002911 Chr.7: 104184184-104184184 on Build GRCh38
rs200547080 Chr.7: 104184214-104184214 on Build GRCh38
rs77344002 Chr.7: 104184472-104184472 on Build GRCh38
rs755221188 Chr.7: 104184106-104184106 on Build GRCh38
rs947447198 Chr.7: 104184427-104184427 on Build GRCh38
rs117174769 Chr.7: 104184494-104184494 on Build GRCh38
rs924880100 Chr.7: 104184463-104184464 on Build GRCh38
rs145655201 Chr.7: 104184478-104184478 on Build GRCh38
rs1023073005 Chr.7: 104184411-104184411 on Build GRCh38
rs760046294 Chr.7: 104184057-104184057 on Build GRCh38
rs776079142 Chr.7: 104184059-104184060 on Build GRCh38
rs751911247 Chr.7: 104184102-104184102 on Build GRCh38
rs981705342 Chr.7: 104184318-104184318 on Build GRCh38
rs758489410 Chr.7: 104184458-104184458 on Build GRCh38
rs11384924 Chr.7: 104184276-104184277 on Build GRCh38
rs933302261 Chr.7: 104184451-104184451 on Build GRCh38
rs148778994 Chr.7: 104184464-104184464 on Build GRCh38
rs201838712 Chr.7: 104184156-104184156 on Build GRCh38
rs2307408 Chr.7: 104184183-104184184 on Build GRCh38
rs751936608 Chr.7: 104184179-104184179 on Build GRCh38
rs772638589 Chr.7: 104184453-104184453 on Build GRCh38
rs763421811 Chr.7: 104184172-104184172 on Build GRCh38
rs924625214 Chr.7: 104184439-104184439 on Build GRCh38
rs571460763 Chr.7: 104184191-104184193 on Build GRCh38
rs767925103 Chr.7: 104184203-104184203 on Build GRCh38
rs199595122 Chr.7: 104184103-104184103 on Build GRCh38
rs369009943 Chr.7: 104184138-104184138 on Build GRCh38
rs760406922 Chr.7: 104184155-104184155 on Build GRCh38
rs201554196 Chr.7: 104184071-104184071 on Build GRCh38
rs755236544 Chr.7: 104184190-104184190 on Build GRCh38
rs748411334 Chr.7: 104184109-104184109 on Build GRCh38
rs915943466 Chr.7: 104184168-104184168 on Build GRCh38
rs7807228 Chr.7: 104184219-104184219 on Build GRCh38
rs756191361 Chr.7: 104184110-104184110 on Build GRCh38
rs951054357 Chr.7: 104184314-104184314 on Build GRCh38
rs200692562 Chr.7: 104184066-104184066 on Build GRCh38
rs530245222 Chr.7: 104184199-104184199 on Build GRCh38
rs207468353 Chr.7: 104184364-104184364 on Build GRCh38
rs749187818 Chr.7: 104184116-104184116 on Build GRCh38
rs140233642 Chr.7: 104184309-104184309 on Build GRCh38
rs752767191 Chr.7: 104184356-104184356 on Build GRCh38
rs551907249 Chr.7: 104184403-104184403 on Build GRCh38
rs763161122 Chr.7: 104184071-104184071 on Build GRCh38
rs776170846 Chr.7: 104184160-104184160 on Build GRCh38
rs566830506 Chr.7: 104184420-104184420 on Build GRCh38
rs397690351 Chr.7: 104184278-104184279 on Build GRCh38
rs771916034 Chr.7: 104184150-104184150 on Build GRCh38
rs1042103515 Chr.7: 104184188-104184188 on Build GRCh38
rs960138343 Chr.7: 104184361-104184361 on Build GRCh38
rs544272424 Chr.7: 104184234-104184234 on Build GRCh38
rs17326187 Chr.7: 104184231-104184231 on Build GRCh38
rs771039759 Chr.7: 104184126-104184126 on Build GRCh38
rs1003375932 Chr.7: 104184326-104184326 on Build GRCh38
rs766711388 Chr.7: 104184080-104184080 on Build GRCh38
rs937497437 Chr.7: 104184338-104184338 on Build GRCh38
rs1018017096 Chr.7: 104184240-104184240 on Build GRCh38
rs73715103 Chr.7: 104184208-104184208 on Build GRCh38
rs34556202 Chr.7: 104184274-104184274 on Build GRCh38
rs764555114 Chr.7: 104184070-104184071 on Build GRCh38
rs569449062 Chr.7: 104184347-104184347 on Build GRCh38
rs745823404 Chr.7: 104184222-104184222 on Build GRCh38

View Full Product Details