Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121099789-121100310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPED1
Entrez Gene ID
79974
Gene Name
cadherin like and PC-esterase domain containing 1
Gene Aliases
C7orf58
UniGene
Hs.189652
Gene Chromosome Location
Chr.7: 120988697-121297444 on Build GRCh38
Transcripts
NM_001105533.1 NM_024913.4 XM_017012649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3949825 c.848C>G p.P283R Chr.7: 121100024-121100024 on Build GRCh38
3698122 c.904C>T p.R302C Chr.7: 121100080-121100080 on Build GRCh38
4161736 c.768T>A p.L256L Chr.7: 121099944-121099944 on Build GRCh38
5823264 c.798A>G p.E266E Chr.7: 121099974-121099974 on Build GRCh38
5450322 c.821C>T p.A274V Chr.7: 121099997-121099997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037681274 Chr.7: 121100208-121100208 on Build GRCh38
rs545633007 Chr.7: 121100194-121100194 on Build GRCh38
rs1055436281 Chr.7: 121099885-121099885 on Build GRCh38
rs781025339 Chr.7: 121100010-121100010 on Build GRCh38
rs747926803 Chr.7: 121099909-121099912 on Build GRCh38
rs538924558 Chr.7: 121100231-121100231 on Build GRCh38
rs930454621 Chr.7: 121099854-121099854 on Build GRCh38
rs869188684 Chr.7: 121099921-121099923 on Build GRCh38
rs113695658 Chr.7: 121100055-121100055 on Build GRCh38
rs781291584 Chr.7: 121099908-121099909 on Build GRCh38
rs377306245 Chr.7: 121100000-121100000 on Build GRCh38
rs749298766 Chr.7: 121099965-121099965 on Build GRCh38
rs374401699 Chr.7: 121099948-121099948 on Build GRCh38
rs1917112 Chr.7: 121100171-121100171 on Build GRCh38
rs375548007 Chr.7: 121100133-121100133 on Build GRCh38
rs777376616 Chr.7: 121100000-121100001 on Build GRCh38
rs777108232 Chr.7: 121100069-121100069 on Build GRCh38
rs199844199 Chr.7: 121099909-121099910 on Build GRCh38
rs897444345 Chr.7: 121100214-121100214 on Build GRCh38
rs200929500 Chr.7: 121099903-121099903 on Build GRCh38
rs183590846 Chr.7: 121099923-121099923 on Build GRCh38
rs33990520 Chr.7: 121099909-121099909 on Build GRCh38
rs1023115117 Chr.7: 121099939-121099939 on Build GRCh38
rs139654343 Chr.7: 121099950-121099950 on Build GRCh38
rs548078791 Chr.7: 121099910-121099911 on Build GRCh38
rs779941795 Chr.7: 121100029-121100029 on Build GRCh38
rs199878690 Chr.7: 121099989-121099989 on Build GRCh38
rs1049418490 Chr.7: 121100114-121100114 on Build GRCh38
rs770238716 Chr.7: 121100139-121100139 on Build GRCh38
rs1054437326 Chr.7: 121099903-121099908 on Build GRCh38
rs201996698 Chr.7: 121100030-121100030 on Build GRCh38
rs879196224 Chr.7: 121099910-121099911 on Build GRCh38
rs553437004 Chr.7: 121100170-121100170 on Build GRCh38
rs764504558 Chr.7: 121100145-121100145 on Build GRCh38
rs760995327 Chr.7: 121100051-121100051 on Build GRCh38
rs964105374 Chr.7: 121099955-121099955 on Build GRCh38
rs770335227 Chr.7: 121099900-121099903 on Build GRCh38
rs772467238 Chr.7: 121100045-121100045 on Build GRCh38
rs767731316 Chr.7: 121099972-121099972 on Build GRCh38
rs746509670 Chr.7: 121099942-121099942 on Build GRCh38
rs569709456 Chr.7: 121099971-121099971 on Build GRCh38
rs753671189 Chr.7: 121100086-121100086 on Build GRCh38
rs111441190 Chr.7: 121100243-121100243 on Build GRCh38
rs756765281 Chr.7: 121100002-121100002 on Build GRCh38
rs796247008 Chr.7: 121099909-121099910 on Build GRCh38
rs199742777 Chr.7: 121099899-121099899 on Build GRCh38
rs762055654 Chr.7: 121100143-121100143 on Build GRCh38
rs536984919 Chr.7: 121099911-121099912 on Build GRCh38
rs758533071 Chr.7: 121100195-121100195 on Build GRCh38
rs778844625 Chr.7: 121100104-121100104 on Build GRCh38
rs894156858 Chr.7: 121099890-121099890 on Build GRCh38
rs372819946 Chr.7: 121100031-121100031 on Build GRCh38
rs869061812 Chr.7: 121099923-121099923 on Build GRCh38
rs149864758 Chr.7: 121099823-121099823 on Build GRCh38
rs775978243 Chr.7: 121100141-121100141 on Build GRCh38
rs201397213 Chr.7: 121099884-121099884 on Build GRCh38
rs771962292 Chr.7: 121100107-121100107 on Build GRCh38
rs533090612 Chr.7: 121100225-121100225 on Build GRCh38
rs145131821 Chr.7: 121100080-121100080 on Build GRCh38
rs1041921711 Chr.7: 121099855-121099855 on Build GRCh38
rs200638100 Chr.7: 121099934-121099934 on Build GRCh38
rs187314481 Chr.7: 121099967-121099967 on Build GRCh38
rs553687081 Chr.7: 121099839-121099839 on Build GRCh38
rs767723334 Chr.7: 121099913-121099913 on Build GRCh38
rs760968774 Chr.7: 121099980-121099980 on Build GRCh38
rs773441913 Chr.7: 121099976-121099976 on Build GRCh38
rs567896255 Chr.7: 121100098-121100098 on Build GRCh38
rs1027351589 Chr.7: 121100241-121100241 on Build GRCh38
rs936142278 Chr.7: 121100012-121100012 on Build GRCh38
rs779561603 Chr.7: 121100082-121100082 on Build GRCh38
rs748131754 Chr.7: 121100105-121100105 on Build GRCh38
rs766221115 Chr.7: 121100224-121100224 on Build GRCh38
rs1017365496 Chr.7: 121100271-121100271 on Build GRCh38
rs879072673 Chr.7: 121099910-121099910 on Build GRCh38
rs798937 Chr.7: 121100049-121100049 on Build GRCh38
rs754119316 Chr.7: 121099991-121099991 on Build GRCh38
rs765821693 Chr.7: 121099998-121099998 on Build GRCh38
rs766323294 Chr.7: 121100181-121100181 on Build GRCh38
rs775313324 Chr.7: 121099952-121099952 on Build GRCh38
rs200483154 Chr.7: 121099966-121099966 on Build GRCh38
rs574258870 Chr.7: 121099821-121099821 on Build GRCh38
rs536820540 Chr.7: 121100124-121100124 on Build GRCh38
rs777971197 Chr.7: 121100115-121100115 on Build GRCh38
rs527698812 Chr.7: 121099909-121099911 on Build GRCh38
rs1044474069 Chr.7: 121100127-121100127 on Build GRCh38
rs754726185 Chr.7: 121100100-121100100 on Build GRCh38
rs757025021 Chr.7: 121099945-121099945 on Build GRCh38
rs774206526 Chr.7: 121100046-121100046 on Build GRCh38
rs115702920 Chr.7: 121099879-121099879 on Build GRCh38
rs755604681 Chr.7: 121099996-121099996 on Build GRCh38
rs530575190 Chr.7: 121100013-121100013 on Build GRCh38
rs1007291198 Chr.7: 121100244-121100244 on Build GRCh38
rs910452805 Chr.7: 121099871-121099871 on Build GRCh38
rs926150585 Chr.7: 121100007-121100007 on Build GRCh38
rs147197905 Chr.7: 121100073-121100073 on Build GRCh38
rs750798769 Chr.7: 121099916-121099916 on Build GRCh38
rs765846204 Chr.7: 121100074-121100074 on Build GRCh38
rs375819148 Chr.7: 121100081-121100081 on Build GRCh38
rs751877872 Chr.7: 121099907-121099909 on Build GRCh38
rs150593278 Chr.7: 121099936-121099936 on Build GRCh38
rs567876875 Chr.7: 121099911-121099911 on Build GRCh38
rs142662878 Chr.7: 121099924-121099924 on Build GRCh38
rs766825335 Chr.7: 121099928-121099928 on Build GRCh38
rs200075911 Chr.7: 121100075-121100075 on Build GRCh38
rs746635114 Chr.7: 121100125-121100126 on Build GRCh38
rs755414515 Chr.7: 121099935-121099935 on Build GRCh38
rs897482204 Chr.7: 121100065-121100065 on Build GRCh38
rs369996126 Chr.7: 121100125-121100125 on Build GRCh38
rs149714091 Chr.7: 121100048-121100048 on Build GRCh38
rs201671315 Chr.7: 121100109-121100109 on Build GRCh38

View Full Product Details