Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161156870-161157374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFC1
Entrez Gene ID
51506
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
HSPC155
UniGene
Hs.301412
Gene Chromosome Location
Chr.1: 161132754-161158856 on Build GRCh38
Transcripts
NM_016406.3 XM_005245254.1 XM_017001450.1 XM_017001451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668153 c.138C>G p.N46K Chr.1: 161156964-161156964 on Build GRCh38
5046364 c.220G>A p.D74N Chr.1: 161157282-161157282 on Build GRCh38
1319819 c.208T>G p.W70G Chr.1: 161157270-161157270 on Build GRCh38
4739028 c.255+1G>A p.? Chr.1: 161157318-161157318 on Build GRCh38
3477178 c.223C>T p.L75F Chr.1: 161157285-161157285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760085314 Chr.1: 161157058-161157058 on Build GRCh38
rs980022635 Chr.1: 161157125-161157125 on Build GRCh38
rs529045897 Chr.1: 161157308-161157308 on Build GRCh38
rs115695944 Chr.1: 161156964-161156964 on Build GRCh38
rs764017795 Chr.1: 161157218-161157218 on Build GRCh38
rs754143657 Chr.1: 161157233-161157233 on Build GRCh38
rs756328544 Chr.1: 161156931-161156931 on Build GRCh38
rs753938820 Chr.1: 161157010-161157010 on Build GRCh38
rs1011787814 Chr.1: 161157013-161157013 on Build GRCh38
rs928450360 Chr.1: 161157221-161157221 on Build GRCh38
rs992777702 Chr.1: 161157136-161157136 on Build GRCh38
rs776075286 Chr.1: 161157328-161157328 on Build GRCh38
rs765167891 Chr.1: 161157242-161157242 on Build GRCh38
rs763408590 Chr.1: 161156981-161156981 on Build GRCh38
rs574100964 Chr.1: 161156989-161156989 on Build GRCh38
rs368611076 Chr.1: 161157084-161157084 on Build GRCh38
rs918534548 Chr.1: 161157180-161157180 on Build GRCh38
rs145750773 Chr.1: 161156899-161156899 on Build GRCh38
rs777476658 Chr.1: 161156945-161156945 on Build GRCh38
rs910048155 Chr.1: 161157305-161157305 on Build GRCh38
rs778834205 Chr.1: 161157017-161157017 on Build GRCh38
rs745871078 Chr.1: 161157018-161157018 on Build GRCh38
rs755055965 Chr.1: 161156910-161156910 on Build GRCh38
rs191173120 Chr.1: 161157016-161157016 on Build GRCh38
rs778224896 Chr.1: 161157280-161157280 on Build GRCh38
rs374413995 Chr.1: 161157061-161157061 on Build GRCh38
rs776546585 Chr.1: 161157342-161157342 on Build GRCh38
rs768013508 Chr.1: 161157034-161157034 on Build GRCh38
rs960055260 Chr.1: 161157135-161157135 on Build GRCh38
rs147604779 Chr.1: 161157317-161157317 on Build GRCh38
rs148546741 Chr.1: 161156906-161156906 on Build GRCh38
rs763272547 Chr.1: 161156903-161156903 on Build GRCh38
rs200218667 Chr.1: 161156962-161156962 on Build GRCh38
rs761314808 Chr.1: 161157048-161157048 on Build GRCh38
rs752983006 Chr.1: 161156921-161156921 on Build GRCh38
rs557240094 Chr.1: 161156980-161156980 on Build GRCh38
rs951563082 Chr.1: 161157163-161157163 on Build GRCh38
rs541155454 Chr.1: 161157137-161157137 on Build GRCh38
rs918440885 Chr.1: 161156973-161156973 on Build GRCh38
rs986614413 Chr.1: 161157126-161157126 on Build GRCh38
rs565767505 Chr.1: 161157318-161157318 on Build GRCh38
rs778085299 Chr.1: 161157007-161157007 on Build GRCh38
rs563623271 Chr.1: 161157195-161157195 on Build GRCh38
rs772108711 Chr.1: 161157029-161157029 on Build GRCh38
rs780172906 Chr.1: 161157301-161157301 on Build GRCh38
rs372460141 Chr.1: 161156977-161156977 on Build GRCh38
rs771343038 Chr.1: 161157054-161157054 on Build GRCh38
rs756569237 Chr.1: 161157228-161157228 on Build GRCh38
rs76617482 Chr.1: 161156915-161156915 on Build GRCh38
rs28449692 Chr.1: 161156932-161156932 on Build GRCh38
rs766933349 Chr.1: 161156905-161156905 on Build GRCh38
rs78466983 Chr.1: 161157138-161157138 on Build GRCh38
rs760810685 Chr.1: 161157217-161157217 on Build GRCh38
rs764297659 Chr.1: 161157340-161157340 on Build GRCh38
rs747874867 Chr.1: 161157310-161157310 on Build GRCh38
rs963348736 Chr.1: 161157172-161157172 on Build GRCh38
rs745738273 Chr.1: 161156967-161156967 on Build GRCh38
rs142789001 Chr.1: 161156979-161156979 on Build GRCh38
rs757419243 Chr.1: 161157240-161157240 on Build GRCh38
rs749127647 Chr.1: 161156953-161156953 on Build GRCh38
rs752749910 Chr.1: 161157002-161157002 on Build GRCh38
rs1055429965 Chr.1: 161157272-161157272 on Build GRCh38
rs532997448 Chr.1: 161157176-161157176 on Build GRCh38
rs1034229270 Chr.1: 161157130-161157130 on Build GRCh38
rs775935466 Chr.1: 161157205-161157205 on Build GRCh38
rs767625014 Chr.1: 161156998-161156998 on Build GRCh38
rs756096248 Chr.1: 161157005-161157005 on Build GRCh38
rs747407933 Chr.1: 161157043-161157043 on Build GRCh38
rs1030535054 Chr.1: 161157335-161157335 on Build GRCh38
rs199514509 Chr.1: 161156990-161156990 on Build GRCh38
rs937113965 Chr.1: 161157226-161157226 on Build GRCh38
rs947025377 Chr.1: 161157306-161157306 on Build GRCh38
rs777489287 Chr.1: 161157054-161157055 on Build GRCh38
rs139173256 Chr.1: 161157309-161157309 on Build GRCh38
rs761874084 Chr.1: 161157296-161157296 on Build GRCh38
rs12031437 Chr.1: 161157185-161157185 on Build GRCh38
rs776352697 Chr.1: 161157041-161157041 on Build GRCh38
rs774665995 Chr.1: 161157056-161157056 on Build GRCh38
rs968549100 Chr.1: 161157086-161157086 on Build GRCh38
rs923353174 Chr.1: 161156898-161156898 on Build GRCh38
rs376658014 Chr.1: 161157303-161157303 on Build GRCh38
rs572091473 Chr.1: 161157085-161157085 on Build GRCh38
rs766587193 Chr.1: 161156988-161156988 on Build GRCh38
rs769414580 Chr.1: 161157311-161157311 on Build GRCh38
rs771769527 Chr.1: 161156974-161156974 on Build GRCh38
rs373077112 Chr.1: 161157334-161157334 on Build GRCh38
rs750223673 Chr.1: 161157246-161157246 on Build GRCh38
rs1030106038 Chr.1: 161156991-161156991 on Build GRCh38

View Full Product Details