Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167053842-167054376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017000005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899012 c.934G>T p.E312* Chr.1: 167054360-167054360 on Build GRCh38
5993357 c.932G>A p.R311H Chr.1: 167054362-167054362 on Build GRCh38
6856148 c.941C>T p.P314L Chr.1: 167054353-167054353 on Build GRCh38
1733535 c.931C>T p.R311C Chr.1: 167054363-167054363 on Build GRCh38
5719393 c.930G>A p.G310G Chr.1: 167054364-167054364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553060907 Chr.1: 167054124-167054124 on Build GRCh38
rs1029392225 Chr.1: 167054328-167054328 on Build GRCh38
rs1033582521 Chr.1: 167054020-167054020 on Build GRCh38
rs3820388 Chr.1: 167054189-167054189 on Build GRCh38
rs909038577 Chr.1: 167054097-167054097 on Build GRCh38
rs367913743 Chr.1: 167054284-167054284 on Build GRCh38
rs534624445 Chr.1: 167054108-167054108 on Build GRCh38
rs892962514 Chr.1: 167054072-167054072 on Build GRCh38
rs931853971 Chr.1: 167053872-167053872 on Build GRCh38
rs146949681 Chr.1: 167054319-167054319 on Build GRCh38
rs55881654 Chr.1: 167053912-167053912 on Build GRCh38
rs1001003975 Chr.1: 167054231-167054231 on Build GRCh38
rs953529951 Chr.1: 167054333-167054333 on Build GRCh38
rs1011916396 Chr.1: 167053942-167053942 on Build GRCh38
rs1039399085 Chr.1: 167054117-167054117 on Build GRCh38
rs1030068837 Chr.1: 167053965-167053965 on Build GRCh38
rs904970174 Chr.1: 167054205-167054205 on Build GRCh38
rs778599635 Chr.1: 167054346-167054346 on Build GRCh38
rs138642494 Chr.1: 167054005-167054005 on Build GRCh38
rs374024652 Chr.1: 167054283-167054283 on Build GRCh38
rs750060636 Chr.1: 167054352-167054352 on Build GRCh38
rs575655173 Chr.1: 167053880-167053880 on Build GRCh38
rs201876151 Chr.1: 167054297-167054297 on Build GRCh38
rs546752990 Chr.1: 167053978-167053978 on Build GRCh38
rs766281753 Chr.1: 167054342-167054342 on Build GRCh38
rs181146882 Chr.1: 167054044-167054044 on Build GRCh38
rs189874689 Chr.1: 167054037-167054037 on Build GRCh38
rs148351388 Chr.1: 167054353-167054353 on Build GRCh38
rs570324429 Chr.1: 167054075-167054075 on Build GRCh38
rs950487574 Chr.1: 167053976-167053976 on Build GRCh38
rs957666750 Chr.1: 167054021-167054021 on Build GRCh38
rs145794171 Chr.1: 167053866-167053866 on Build GRCh38
rs528566247 Chr.1: 167053946-167053946 on Build GRCh38
rs41270694 Chr.1: 167054244-167054244 on Build GRCh38
rs546171516 Chr.1: 167053892-167053892 on Build GRCh38
rs569207400 Chr.1: 167054048-167054048 on Build GRCh38
rs997215361 Chr.1: 167053956-167053956 on Build GRCh38
rs372035414 Chr.1: 167054315-167054315 on Build GRCh38
rs997164380 Chr.1: 167054143-167054143 on Build GRCh38
rs940596440 Chr.1: 167053897-167053897 on Build GRCh38
rs767693291 Chr.1: 167054324-167054324 on Build GRCh38
rs893025006 Chr.1: 167053927-167053927 on Build GRCh38
rs1017234155 Chr.1: 167053943-167053943 on Build GRCh38
rs1036245336 Chr.1: 167054115-167054115 on Build GRCh38
rs777178171 Chr.1: 167054302-167054302 on Build GRCh38
rs887732321 Chr.1: 167053884-167053884 on Build GRCh38
rs1045096812 Chr.1: 167054195-167054195 on Build GRCh38
rs759757858 Chr.1: 167054323-167054323 on Build GRCh38
rs565884581 Chr.1: 167054318-167054318 on Build GRCh38
rs552127531 Chr.1: 167054061-167054061 on Build GRCh38
rs984667389 Chr.1: 167054095-167054095 on Build GRCh38
rs998713313 Chr.1: 167053911-167053911 on Build GRCh38
rs893299195 Chr.1: 167054253-167054253 on Build GRCh38
rs953576069 Chr.1: 167054084-167054084 on Build GRCh38
rs990013431 Chr.1: 167054339-167054339 on Build GRCh38
rs916576927 Chr.1: 167054034-167054034 on Build GRCh38
rs76170587 Chr.1: 167054049-167054049 on Build GRCh38
rs376733018 Chr.1: 167053977-167053977 on Build GRCh38
rs991073953 Chr.1: 167054268-167054268 on Build GRCh38
rs776012221 Chr.1: 167054135-167054135 on Build GRCh38
rs748664530 Chr.1: 167054310-167054310 on Build GRCh38
rs545808958 Chr.1: 167054334-167054334 on Build GRCh38
rs1001722410 Chr.1: 167054018-167054018 on Build GRCh38
rs35152579 Chr.1: 167054343-167054343 on Build GRCh38
rs916839525 Chr.1: 167054272-167054272 on Build GRCh38
rs950435375 Chr.1: 167054197-167054197 on Build GRCh38
rs763105903 Chr.1: 167054314-167054314 on Build GRCh38
rs375327487 Chr.1: 167054094-167054094 on Build GRCh38
rs773584975 Chr.1: 167054312-167054312 on Build GRCh38
rs1004589120 Chr.1: 167054201-167054201 on Build GRCh38
rs940647184 Chr.1: 167054102-167054102 on Build GRCh38
rs747496437 Chr.1: 167054019-167054019 on Build GRCh38
rs752628346 Chr.1: 167054331-167054331 on Build GRCh38
rs772675633 Chr.1: 167053994-167053994 on Build GRCh38
rs1052924199 Chr.1: 167054071-167054071 on Build GRCh38
rs770094266 Chr.1: 167054311-167054311 on Build GRCh38

View Full Product Details