Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73789500-73790037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2W
Entrez Gene ID
55284
Gene Name
ubiquitin conjugating enzyme E2 W (putative)
Gene Aliases
UBC-16, UBC16
UniGene
Hs.128841
Gene Chromosome Location
Chr.8: 73780097-73878910 on Build GRCh38
Transcripts
NM_001001481.2 NM_001271015.1 NM_018299.4 NR_073119.1 NR_073120.1 NR_073121.1

SNP Detail

SNP ID Chromosome Location
rs921975712 Chr.8: 73789832-73789832 on Build GRCh38
rs1035979406 Chr.8: 73789876-73789876 on Build GRCh38
rs956775444 Chr.8: 73789741-73789741 on Build GRCh38
rs141930626 Chr.8: 73789990-73789990 on Build GRCh38
rs1044429038 Chr.8: 73790000-73790000 on Build GRCh38
rs878912963 Chr.8: 73789651-73789651 on Build GRCh38
rs1003628900 Chr.8: 73789671-73789671 on Build GRCh38
rs970182114 Chr.8: 73789715-73789715 on Build GRCh38
rs896758021 Chr.8: 73789976-73789976 on Build GRCh38
rs762022623 Chr.8: 73789813-73789813 on Build GRCh38
rs930370357 Chr.8: 73789953-73789953 on Build GRCh38
rs750996696 Chr.8: 73790004-73790004 on Build GRCh38
rs765403076 Chr.8: 73789842-73789842 on Build GRCh38
rs1049365694 Chr.8: 73789524-73789524 on Build GRCh38
rs946765218 Chr.8: 73789838-73789839 on Build GRCh38
rs944341911 Chr.8: 73789855-73789855 on Build GRCh38
rs1014359560 Chr.8: 73789622-73789622 on Build GRCh38
rs950565351 Chr.8: 73789743-73789743 on Build GRCh38
rs970692081 Chr.8: 73789646-73789646 on Build GRCh38
rs544748154 Chr.8: 73789597-73789597 on Build GRCh38
rs551814993 Chr.8: 73789871-73789871 on Build GRCh38
rs1022699150 Chr.8: 73789640-73789640 on Build GRCh38
rs549227376 Chr.8: 73789882-73789882 on Build GRCh38
rs150223229 Chr.8: 73789717-73789717 on Build GRCh38
rs1031055267 Chr.8: 73789675-73789675 on Build GRCh38
rs986764688 Chr.8: 73789831-73789831 on Build GRCh38
rs983365992 Chr.8: 73789791-73789791 on Build GRCh38
rs747053899 Chr.8: 73789988-73789988 on Build GRCh38
rs540471745 Chr.8: 73789561-73789561 on Build GRCh38
rs761306811 Chr.8: 73789820-73789820 on Build GRCh38
rs1043800291 Chr.8: 73789893-73789893 on Build GRCh38
rs181615714 Chr.8: 73789566-73789566 on Build GRCh38
rs895478463 Chr.8: 73789621-73789621 on Build GRCh38
rs924102260 Chr.8: 73789948-73789948 on Build GRCh38
rs919047530 Chr.8: 73789896-73789896 on Build GRCh38
rs911562125 Chr.8: 73789833-73789833 on Build GRCh38
rs532850990 Chr.8: 73789585-73789585 on Build GRCh38
rs1056621702 Chr.8: 73789962-73789962 on Build GRCh38
rs989690217 Chr.8: 73789812-73789812 on Build GRCh38
rs16938727 Chr.8: 73789533-73789533 on Build GRCh38
rs367562118 Chr.8: 73789704-73789704 on Build GRCh38
rs776969134 Chr.8: 73789792-73789792 on Build GRCh38
rs138896690 Chr.8: 73789872-73789872 on Build GRCh38
rs953669431 Chr.8: 73789822-73789822 on Build GRCh38

View Full Product Details