Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86488161-86488698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
UniGene
Hs.145386
Gene Chromosome Location
Chr.8: 86467116-86514348 on Build GRCh38
Transcripts
NM_001286707.1 NM_001286719.1 NM_001317807.1 NM_016033.2 XM_005250944.3 XM_005250947.3 XM_005250948.3 XM_006716573.3 XM_011517090.2 XM_011517091.2 XM_011517092.2 XM_011517093.2 XM_011517094.2 XM_011517095.2 XM_011517096.2 XM_011517097.2 XM_011517098.2 XM_017013518.1 XM_017013519.1 XM_017013520.1 XM_017013521.1 XM_017013522.1 XM_017013523.1 XM_017013524.1 XM_017013525.1 XM_017013526.1 XM_017013527.1 XM_017013528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767017 c.303A>C p.K101N Chr.8: 86488584-86488584 on Build GRCh38
1102190 c.335G>T p.S112I Chr.8: 86488552-86488552 on Build GRCh38
6269546 c.327C>A p.Y109* Chr.8: 86488560-86488560 on Build GRCh38
486726 c.290G>C p.G97A Chr.8: 86488597-86488597 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045253120 Chr.8: 86488497-86488497 on Build GRCh38
rs777969115 Chr.8: 86488560-86488561 on Build GRCh38
rs746258149 Chr.8: 86488657-86488658 on Build GRCh38
rs368241098 Chr.8: 86488598-86488598 on Build GRCh38
rs531975593 Chr.8: 86488205-86488205 on Build GRCh38
rs748991916 Chr.8: 86488575-86488575 on Build GRCh38
rs762808149 Chr.8: 86488535-86488535 on Build GRCh38
rs752618060 Chr.8: 86488656-86488656 on Build GRCh38
rs567832560 Chr.8: 86488459-86488459 on Build GRCh38
rs1032343353 Chr.8: 86488242-86488242 on Build GRCh38
rs757509509 Chr.8: 86488601-86488602 on Build GRCh38
rs747337179 Chr.8: 86488593-86488593 on Build GRCh38
rs553431189 Chr.8: 86488501-86488501 on Build GRCh38
rs199749886 Chr.8: 86488514-86488514 on Build GRCh38
rs201315294 Chr.8: 86488676-86488676 on Build GRCh38
rs182603700 Chr.8: 86488649-86488649 on Build GRCh38
rs777374497 Chr.8: 86488572-86488572 on Build GRCh38
rs142480071 Chr.8: 86488618-86488618 on Build GRCh38
rs1056664605 Chr.8: 86488420-86488420 on Build GRCh38
rs911228674 Chr.8: 86488351-86488351 on Build GRCh38
rs752205957 Chr.8: 86488566-86488566 on Build GRCh38
rs755599338 Chr.8: 86488663-86488663 on Build GRCh38
rs896618741 Chr.8: 86488447-86488447 on Build GRCh38
rs202111370 Chr.8: 86488640-86488640 on Build GRCh38
rs758335161 Chr.8: 86488404-86488404 on Build GRCh38
rs1023699747 Chr.8: 86488301-86488301 on Build GRCh38
rs781642599 Chr.8: 86488604-86488605 on Build GRCh38
rs985382058 Chr.8: 86488647-86488647 on Build GRCh38
rs768612519 Chr.8: 86488506-86488506 on Build GRCh38
rs1018238262 Chr.8: 86488659-86488659 on Build GRCh38
rs941308241 Chr.8: 86488356-86488356 on Build GRCh38
rs761909255 Chr.8: 86488609-86488609 on Build GRCh38
rs750701897 Chr.8: 86488522-86488522 on Build GRCh38
rs780462572 Chr.8: 86488590-86488590 on Build GRCh38
rs559917586 Chr.8: 86488200-86488200 on Build GRCh38
rs1033091032 Chr.8: 86488580-86488580 on Build GRCh38
rs34032915 Chr.8: 86488555-86488556 on Build GRCh38
rs112070246 Chr.8: 86488652-86488652 on Build GRCh38
rs539045891 Chr.8: 86488543-86488543 on Build GRCh38
rs536375686 Chr.8: 86488464-86488464 on Build GRCh38
rs970846180 Chr.8: 86488324-86488324 on Build GRCh38
rs1015095816 Chr.8: 86488438-86488438 on Build GRCh38
rs988684175 Chr.8: 86488272-86488272 on Build GRCh38
rs372789326 Chr.8: 86488538-86488538 on Build GRCh38
rs772413519 Chr.8: 86488580-86488580 on Build GRCh38
rs770387526 Chr.8: 86488665-86488666 on Build GRCh38
rs751450183 Chr.8: 86488539-86488539 on Build GRCh38
rs755611388 Chr.8: 86488571-86488571 on Build GRCh38
rs865942018 Chr.8: 86488275-86488275 on Build GRCh38
rs920265248 Chr.8: 86488470-86488470 on Build GRCh38
rs147377085 Chr.8: 86488415-86488421 on Build GRCh38
rs759120176 Chr.8: 86488237-86488237 on Build GRCh38
rs955329377 Chr.8: 86488287-86488287 on Build GRCh38
rs924480650 Chr.8: 86488585-86488585 on Build GRCh38
rs186063483 Chr.8: 86488457-86488457 on Build GRCh38
rs752243697 Chr.8: 86488388-86488388 on Build GRCh38
rs140258784 Chr.8: 86488554-86488554 on Build GRCh38
rs113621178 Chr.8: 86488285-86488285 on Build GRCh38
rs776705038 Chr.8: 86488511-86488511 on Build GRCh38
rs117026435 Chr.8: 86488243-86488243 on Build GRCh38
rs765505751 Chr.8: 86488516-86488516 on Build GRCh38
rs770029265 Chr.8: 86488617-86488617 on Build GRCh38
rs753810849 Chr.8: 86488504-86488504 on Build GRCh38
rs974101766 Chr.8: 86488467-86488467 on Build GRCh38
rs373720421 Chr.8: 86488249-86488249 on Build GRCh38
rs766715344 Chr.8: 86488631-86488631 on Build GRCh38
rs747196042 Chr.8: 86488579-86488580 on Build GRCh38
rs376892466 Chr.8: 86488599-86488599 on Build GRCh38
rs1032730270 Chr.8: 86488220-86488220 on Build GRCh38
rs551697873 Chr.8: 86488221-86488221 on Build GRCh38
rs758389019 Chr.8: 86488550-86488551 on Build GRCh38
rs918717679 Chr.8: 86488392-86488392 on Build GRCh38

View Full Product Details