Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90645021-90645514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM64
Entrez Gene ID
169200
Gene Name
transmembrane protein 64
Gene Aliases
-
UniGene
Hs.567759
Gene Chromosome Location
Chr.8: 90621995-90645905 on Build GRCh38
Transcripts
NM_001008495.3 NM_001146273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1458562 c.541C>T p.L181F Chr.8: 90645365-90645365 on Build GRCh38
4804757 c.395T>A p.L132Q Chr.8: 90645511-90645511 on Build GRCh38
3367423 c.131T>C p.V44A Chr.8: 90645193-90645193 on Build GRCh38
1458560 c.599T>C p.V200A Chr.8: 90645307-90645307 on Build GRCh38
4734594 c.786A>C p.A262A Chr.8: 90645120-90645120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557304485 Chr.8: 90645121-90645121 on Build GRCh38
rs769086295 Chr.8: 90645093-90645093 on Build GRCh38
rs759807350 Chr.8: 90645131-90645131 on Build GRCh38
rs374040300 Chr.8: 90645080-90645080 on Build GRCh38
rs752503818 Chr.8: 90645140-90645140 on Build GRCh38
rs766406618 Chr.8: 90645106-90645106 on Build GRCh38
rs948642463 Chr.8: 90645270-90645270 on Build GRCh38
rs756855360 Chr.8: 90645159-90645159 on Build GRCh38
rs112214008 Chr.8: 90645110-90645110 on Build GRCh38
rs367657591 Chr.8: 90645429-90645429 on Build GRCh38
rs528083855 Chr.8: 90645277-90645277 on Build GRCh38
rs777765372 Chr.8: 90645072-90645072 on Build GRCh38
rs868228432 Chr.8: 90645173-90645173 on Build GRCh38
rs765235284 Chr.8: 90645287-90645287 on Build GRCh38
rs773739088 Chr.8: 90645192-90645192 on Build GRCh38
rs1021911919 Chr.8: 90645052-90645052 on Build GRCh38
rs540089087 Chr.8: 90645345-90645345 on Build GRCh38
rs748518409 Chr.8: 90645189-90645189 on Build GRCh38
rs746210252 Chr.8: 90645237-90645239 on Build GRCh38
rs775719726 Chr.8: 90645254-90645254 on Build GRCh38
rs767692449 Chr.8: 90645250-90645250 on Build GRCh38
rs761968646 Chr.8: 90645180-90645180 on Build GRCh38
rs144669410 Chr.8: 90645139-90645139 on Build GRCh38
rs749624341 Chr.8: 90645351-90645351 on Build GRCh38
rs776909047 Chr.8: 90645094-90645094 on Build GRCh38
rs778709150 Chr.8: 90645089-90645089 on Build GRCh38
rs756459793 Chr.8: 90645332-90645332 on Build GRCh38
rs200134832 Chr.8: 90645371-90645371 on Build GRCh38
rs568979194 Chr.8: 90645453-90645453 on Build GRCh38
rs745474745 Chr.8: 90645091-90645091 on Build GRCh38
rs954256859 Chr.8: 90645059-90645059 on Build GRCh38
rs750714788 Chr.8: 90645297-90645297 on Build GRCh38
rs761693690 Chr.8: 90645482-90645482 on Build GRCh38
rs952687304 Chr.8: 90645228-90645228 on Build GRCh38
rs771497912 Chr.8: 90645352-90645352 on Build GRCh38
rs370150546 Chr.8: 90645160-90645160 on Build GRCh38
rs1008127054 Chr.8: 90645226-90645226 on Build GRCh38
rs774458105 Chr.8: 90645239-90645239 on Build GRCh38
rs773652938 Chr.8: 90645103-90645103 on Build GRCh38
rs763026957 Chr.8: 90645105-90645105 on Build GRCh38
rs770202439 Chr.8: 90645247-90645248 on Build GRCh38
rs774829564 Chr.8: 90645361-90645361 on Build GRCh38
rs753759014 Chr.8: 90645158-90645158 on Build GRCh38
rs974139139 Chr.8: 90645320-90645320 on Build GRCh38
rs763291928 Chr.8: 90645311-90645311 on Build GRCh38
rs895124093 Chr.8: 90645051-90645051 on Build GRCh38
rs759659301 Chr.8: 90645240-90645240 on Build GRCh38
rs771173189 Chr.8: 90645079-90645091 on Build GRCh38
rs963777160 Chr.8: 90645267-90645267 on Build GRCh38
rs746632910 Chr.8: 90645323-90645323 on Build GRCh38
rs545600360 Chr.8: 90645221-90645221 on Build GRCh38
rs908020228 Chr.8: 90645365-90645365 on Build GRCh38
rs779751595 Chr.8: 90645188-90645188 on Build GRCh38
rs776878174 Chr.8: 90645080-90645081 on Build GRCh38
rs1007519852 Chr.8: 90645439-90645439 on Build GRCh38
rs373056626 Chr.8: 90645191-90645191 on Build GRCh38
rs1042739726 Chr.8: 90645399-90645399 on Build GRCh38
rs1012642403 Chr.8: 90645088-90645088 on Build GRCh38
rs766779156 Chr.8: 90645438-90645438 on Build GRCh38
rs776607007 Chr.8: 90645461-90645461 on Build GRCh38
rs1007095918 Chr.8: 90645165-90645166 on Build GRCh38
rs890073654 Chr.8: 90645401-90645401 on Build GRCh38
rs753845236 Chr.8: 90645263-90645263 on Build GRCh38
rs756048599 Chr.8: 90645063-90645063 on Build GRCh38
rs777582971 Chr.8: 90645149-90645149 on Build GRCh38
rs765323164 Chr.8: 90645491-90645491 on Build GRCh38
rs1043843410 Chr.8: 90645104-90645104 on Build GRCh38
rs762204998 Chr.8: 90645101-90645101 on Build GRCh38
rs749746976 Chr.8: 90645211-90645211 on Build GRCh38
rs1019782390 Chr.8: 90645447-90645447 on Build GRCh38
rs745348330 Chr.8: 90645162-90645162 on Build GRCh38
rs750019142 Chr.8: 90645288-90645288 on Build GRCh38
rs757910770 Chr.8: 90645310-90645310 on Build GRCh38
rs140609855 Chr.8: 90645260-90645260 on Build GRCh38
rs563701639 Chr.8: 90645261-90645261 on Build GRCh38
rs370671668 Chr.8: 90645074-90645074 on Build GRCh38
rs200765950 Chr.8: 90645363-90645363 on Build GRCh38
rs947057723 Chr.8: 90645389-90645389 on Build GRCh38
rs992392051 Chr.8: 90645440-90645440 on Build GRCh38
rs550730991 Chr.8: 90645420-90645420 on Build GRCh38
rs891536640 Chr.8: 90645384-90645384 on Build GRCh38

View Full Product Details