Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168297391-168297890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX19
Entrez Gene ID
9095
Gene Name
T-box 19
Gene Aliases
TBS19, TPIT, dJ747L4.1
UniGene
Hs.716656
Gene Chromosome Location
Chr.1: 168281040-168314426 on Build GRCh38
Transcripts
NM_005149.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434823 c.655G>A p.A219T Chr.1: 168297775-168297775 on Build GRCh38
3863454 c.632C>T p.P211L Chr.1: 168297752-168297752 on Build GRCh38
5341725 c.112C>A p.L38I Chr.1: 168297735-168297735 on Build GRCh38
5889618 c.661G>A p.E221K Chr.1: 168297781-168297781 on Build GRCh38
5052788 c.665+2T>C p.? Chr.1: 168297787-168297787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750098406 Chr.1: 168297828-168297828 on Build GRCh38
rs543913611 Chr.1: 168297863-168297863 on Build GRCh38
rs937596824 Chr.1: 168297604-168297604 on Build GRCh38
rs748717639 Chr.1: 168297728-168297728 on Build GRCh38
rs747193913 Chr.1: 168297518-168297518 on Build GRCh38
rs568545240 Chr.1: 168297608-168297608 on Build GRCh38
rs773404822 Chr.1: 168297628-168297628 on Build GRCh38
rs752703804 Chr.1: 168297658-168297658 on Build GRCh38
rs1018233502 Chr.1: 168297758-168297758 on Build GRCh38
rs756970226 Chr.1: 168297807-168297807 on Build GRCh38
rs957931873 Chr.1: 168297727-168297727 on Build GRCh38
rs756989988 Chr.1: 168297693-168297693 on Build GRCh38
rs540505768 Chr.1: 168297631-168297631 on Build GRCh38
rs769216569 Chr.1: 168297602-168297602 on Build GRCh38
rs1025363737 Chr.1: 168297834-168297834 on Build GRCh38
rs778811027 Chr.1: 168297517-168297517 on Build GRCh38
rs1040495670 Chr.1: 168297476-168297476 on Build GRCh38
rs35031496 Chr.1: 168297854-168297854 on Build GRCh38
rs918356233 Chr.1: 168297582-168297582 on Build GRCh38
rs753730891 Chr.1: 168297806-168297806 on Build GRCh38
rs771549040 Chr.1: 168297717-168297717 on Build GRCh38
rs751759621 Chr.1: 168297638-168297638 on Build GRCh38
rs762213847 Chr.1: 168297616-168297616 on Build GRCh38
rs893129915 Chr.1: 168297613-168297613 on Build GRCh38
rs891005934 Chr.1: 168297688-168297688 on Build GRCh38
rs766499588 Chr.1: 168297633-168297633 on Build GRCh38
rs985086878 Chr.1: 168297502-168297502 on Build GRCh38
rs745367073 Chr.1: 168297599-168297599 on Build GRCh38
rs371332221 Chr.1: 168297657-168297657 on Build GRCh38
rs151125756 Chr.1: 168297791-168297791 on Build GRCh38
rs778409511 Chr.1: 168297598-168297598 on Build GRCh38
rs368837661 Chr.1: 168297830-168297830 on Build GRCh38
rs201143551 Chr.1: 168297620-168297620 on Build GRCh38
rs929253192 Chr.1: 168297555-168297555 on Build GRCh38
rs1047766358 Chr.1: 168297561-168297561 on Build GRCh38
rs537571823 Chr.1: 168297579-168297579 on Build GRCh38
rs753521546 Chr.1: 168297690-168297690 on Build GRCh38
rs746443767 Chr.1: 168297837-168297837 on Build GRCh38
rs201212830 Chr.1: 168297697-168297697 on Build GRCh38
rs999787264 Chr.1: 168297429-168297429 on Build GRCh38
rs374779946 Chr.1: 168297816-168297816 on Build GRCh38
rs759786679 Chr.1: 168297761-168297761 on Build GRCh38
rs746971970 Chr.1: 168297689-168297689 on Build GRCh38
rs16860323 Chr.1: 168297600-168297600 on Build GRCh38
rs542878204 Chr.1: 168297778-168297778 on Build GRCh38
rs973805505 Chr.1: 168297559-168297559 on Build GRCh38
rs779486584 Chr.1: 168297724-168297724 on Build GRCh38
rs745422254 Chr.1: 168297709-168297709 on Build GRCh38
rs764917947 Chr.1: 168297815-168297815 on Build GRCh38
rs372080768 Chr.1: 168297682-168297682 on Build GRCh38
rs763927465 Chr.1: 168297805-168297805 on Build GRCh38
rs369866828 Chr.1: 168297674-168297674 on Build GRCh38
rs771321846 Chr.1: 168297749-168297749 on Build GRCh38
rs1006805593 Chr.1: 168297742-168297742 on Build GRCh38
rs199994044 Chr.1: 168297729-168297729 on Build GRCh38
rs911082983 Chr.1: 168297422-168297422 on Build GRCh38
rs140528998 Chr.1: 168297747-168297747 on Build GRCh38
rs567049651 Chr.1: 168297572-168297572 on Build GRCh38
rs752758748 Chr.1: 168297790-168297790 on Build GRCh38
rs928867777 Chr.1: 168297601-168297601 on Build GRCh38
rs78189582 Chr.1: 168297663-168297663 on Build GRCh38
rs574766678 Chr.1: 168297460-168297460 on Build GRCh38
rs923316052 Chr.1: 168297859-168297859 on Build GRCh38
rs779539932 Chr.1: 168297836-168297836 on Build GRCh38
rs148991498 Chr.1: 168297480-168297480 on Build GRCh38
rs766232297 Chr.1: 168297594-168297594 on Build GRCh38
rs767736352 Chr.1: 168297775-168297775 on Build GRCh38
rs74123913 Chr.1: 168297627-168297627 on Build GRCh38

View Full Product Details