Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169854183-169854684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCYL3
Entrez Gene ID
57147
Gene Name
SCY1 like pseudokinase 3
Gene Aliases
PACE-1, PACE1
UniGene
Hs.435560
Gene Chromosome Location
Chr.1: 169853074-169894267 on Build GRCh38
Transcripts
NM_020423.6 NM_181093.3 XM_006711465.1 XM_011509801.1 XM_011509802.1 XM_011509803.1 XM_017001862.1 XM_017001863.1 XM_017001864.1 XM_017001865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024990 c.1928A>C p.Q643P Chr.1: 169854511-169854511 on Build GRCh38
3863494 c.1916C>T p.P639L Chr.1: 169854361-169854361 on Build GRCh38
6486146 c.1873A>G p.M625V Chr.1: 169854404-169854404 on Build GRCh38
4777976 c.1832A>G p.K611R Chr.1: 169854445-169854445 on Build GRCh38
3863493 c.2078C>T p.P693L Chr.1: 169854361-169854361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776120084 Chr.1: 169854451-169854451 on Build GRCh38
rs771372873 Chr.1: 169854439-169854439 on Build GRCh38
rs777930005 Chr.1: 169854508-169854508 on Build GRCh38
rs748191571 Chr.1: 169854388-169854388 on Build GRCh38
rs374585625 Chr.1: 169854545-169854545 on Build GRCh38
rs146581982 Chr.1: 169854417-169854417 on Build GRCh38
rs12143301 Chr.1: 169854649-169854649 on Build GRCh38
rs4656197 Chr.1: 169854577-169854577 on Build GRCh38
rs1043667543 Chr.1: 169854210-169854210 on Build GRCh38
rs772742115 Chr.1: 169854534-169854534 on Build GRCh38
rs747584587 Chr.1: 169854445-169854447 on Build GRCh38
rs374960229 Chr.1: 169854555-169854555 on Build GRCh38
rs1062976 Chr.1: 169854519-169854519 on Build GRCh38
rs779869723 Chr.1: 169854285-169854285 on Build GRCh38
rs765750536 Chr.1: 169854345-169854345 on Build GRCh38
rs754563724 Chr.1: 169854362-169854363 on Build GRCh38
rs769212971 Chr.1: 169854632-169854632 on Build GRCh38
rs746613991 Chr.1: 169854323-169854325 on Build GRCh38
rs754772695 Chr.1: 169854354-169854354 on Build GRCh38
rs770215041 Chr.1: 169854443-169854443 on Build GRCh38
rs750152645 Chr.1: 169854355-169854355 on Build GRCh38
rs149664958 Chr.1: 169854639-169854639 on Build GRCh38
rs746217601 Chr.1: 169854524-169854524 on Build GRCh38
rs1053231240 Chr.1: 169854584-169854584 on Build GRCh38
rs752749440 Chr.1: 169854504-169854504 on Build GRCh38
rs188968440 Chr.1: 169854258-169854258 on Build GRCh38
rs201351551 Chr.1: 169854302-169854302 on Build GRCh38
rs998979108 Chr.1: 169854227-169854227 on Build GRCh38
rs781556718 Chr.1: 169854581-169854581 on Build GRCh38
rs753638322 Chr.1: 169854489-169854489 on Build GRCh38
rs201750145 Chr.1: 169854630-169854630 on Build GRCh38
rs201275933 Chr.1: 169854650-169854650 on Build GRCh38
rs765072871 Chr.1: 169854495-169854495 on Build GRCh38
rs6702407 Chr.1: 169854529-169854529 on Build GRCh38
rs757633492 Chr.1: 169854265-169854266 on Build GRCh38
rs878978404 Chr.1: 169854444-169854444 on Build GRCh38
rs185704431 Chr.1: 169854254-169854254 on Build GRCh38
rs570451904 Chr.1: 169854533-169854533 on Build GRCh38
rs74371722 Chr.1: 169854455-169854455 on Build GRCh38
rs116366458 Chr.1: 169854237-169854237 on Build GRCh38
rs780486621 Chr.1: 169854612-169854612 on Build GRCh38
rs748364285 Chr.1: 169854229-169854229 on Build GRCh38
rs778884076 Chr.1: 169854219-169854219 on Build GRCh38
rs866459266 Chr.1: 169854574-169854574 on Build GRCh38
rs890601606 Chr.1: 169854595-169854595 on Build GRCh38
rs535561761 Chr.1: 169854560-169854560 on Build GRCh38
rs151324190 Chr.1: 169854521-169854521 on Build GRCh38
rs540873660 Chr.1: 169854352-169854352 on Build GRCh38
rs140613939 Chr.1: 169854654-169854654 on Build GRCh38
rs1027596296 Chr.1: 169854661-169854661 on Build GRCh38
rs575284885 Chr.1: 169854602-169854602 on Build GRCh38
rs772253566 Chr.1: 169854389-169854389 on Build GRCh38
rs974598692 Chr.1: 169854576-169854576 on Build GRCh38
rs192824290 Chr.1: 169854211-169854211 on Build GRCh38
rs758379234 Chr.1: 169854506-169854506 on Build GRCh38
rs776324841 Chr.1: 169854550-169854550 on Build GRCh38
rs201470077 Chr.1: 169854637-169854637 on Build GRCh38
rs756137846 Chr.1: 169854403-169854403 on Build GRCh38
rs920500419 Chr.1: 169854385-169854385 on Build GRCh38
rs568128308 Chr.1: 169854556-169854556 on Build GRCh38
rs751034965 Chr.1: 169854322-169854322 on Build GRCh38
rs56042723 Chr.1: 169854561-169854561 on Build GRCh38
rs1017088861 Chr.1: 169854217-169854217 on Build GRCh38
rs376335762 Chr.1: 169854566-169854566 on Build GRCh38
rs780791337 Chr.1: 169854388-169854389 on Build GRCh38
rs562214139 Chr.1: 169854367-169854367 on Build GRCh38
rs376716355 Chr.1: 169854440-169854440 on Build GRCh38
rs193246431 Chr.1: 169854551-169854551 on Build GRCh38
rs369110458 Chr.1: 169854330-169854330 on Build GRCh38
rs149349564 Chr.1: 169854470-169854470 on Build GRCh38
rs551344610 Chr.1: 169854528-169854528 on Build GRCh38
rs766647157 Chr.1: 169854216-169854216 on Build GRCh38
rs762265291 Chr.1: 169854309-169854309 on Build GRCh38
rs751779597 Chr.1: 169854275-169854275 on Build GRCh38
rs767488890 Chr.1: 169854368-169854368 on Build GRCh38
rs199637041 Chr.1: 169854494-169854494 on Build GRCh38
rs747276144 Chr.1: 169854433-169854433 on Build GRCh38
rs766126185 Chr.1: 169854487-169854487 on Build GRCh38
rs529395088 Chr.1: 169854370-169854370 on Build GRCh38
rs765122352 Chr.1: 169854553-169854553 on Build GRCh38
rs760141791 Chr.1: 169854313-169854313 on Build GRCh38
rs771489570 Chr.1: 169854280-169854280 on Build GRCh38
rs754941806 Chr.1: 169854383-169854383 on Build GRCh38
rs766036301 Chr.1: 169854546-169854546 on Build GRCh38
rs763655003 Chr.1: 169854316-169854316 on Build GRCh38
rs200694889 Chr.1: 169854472-169854472 on Build GRCh38
rs755867923 Chr.1: 169854359-169854359 on Build GRCh38
rs55788990 Chr.1: 169854380-169854380 on Build GRCh38
rs760345142 Chr.1: 169854478-169854478 on Build GRCh38
rs898654901 Chr.1: 169854497-169854497 on Build GRCh38
rs778988645 Chr.1: 169854384-169854384 on Build GRCh38
rs967853716 Chr.1: 169854624-169854624 on Build GRCh38
rs764109119 Chr.1: 169854659-169854659 on Build GRCh38

View Full Product Details