Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94494864-94495406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643661 c.4575T>C p.D1525D Chr.8: 94494926-94494926 on Build GRCh38
5803270 c.4545-1G>A p.? Chr.8: 94494957-94494957 on Build GRCh38
3925721 c.4604C>T p.P1535L Chr.8: 94494897-94494897 on Build GRCh38
4695647 c.4570A>G p.M1524V Chr.8: 94494931-94494931 on Build GRCh38
361057 c.4629A>G p.T1543T Chr.8: 94494872-94494872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970632768 Chr.8: 94495025-94495025 on Build GRCh38
rs1011547010 Chr.8: 94494998-94494998 on Build GRCh38
rs915832774 Chr.8: 94495170-94495170 on Build GRCh38
rs1052021794 Chr.8: 94495288-94495288 on Build GRCh38
rs542882654 Chr.8: 94495290-94495290 on Build GRCh38
rs112365062 Chr.8: 94495009-94495010 on Build GRCh38
rs997751215 Chr.8: 94495116-94495116 on Build GRCh38
rs763006732 Chr.8: 94494948-94494948 on Build GRCh38
rs566764755 Chr.8: 94495051-94495051 on Build GRCh38
rs919183827 Chr.8: 94495009-94495009 on Build GRCh38
rs58577954 Chr.8: 94495204-94495204 on Build GRCh38
rs922280898 Chr.8: 94495283-94495283 on Build GRCh38
rs1053450965 Chr.8: 94495059-94495059 on Build GRCh38
rs575318534 Chr.8: 94495151-94495151 on Build GRCh38
rs146145980 Chr.8: 94494910-94494910 on Build GRCh38
rs34769902 Chr.8: 94495327-94495328 on Build GRCh38
rs183286147 Chr.8: 94495093-94495093 on Build GRCh38
rs965756842 Chr.8: 94495087-94495087 on Build GRCh38
rs563210155 Chr.8: 94495361-94495361 on Build GRCh38
rs1016349694 Chr.8: 94495148-94495148 on Build GRCh38
rs1003663006 Chr.8: 94494893-94494893 on Build GRCh38
rs985077328 Chr.8: 94495157-94495157 on Build GRCh38
rs772881638 Chr.8: 94494947-94494947 on Build GRCh38
rs746143111 Chr.8: 94495064-94495064 on Build GRCh38
rs1052394243 Chr.8: 94495120-94495120 on Build GRCh38
rs4366055 Chr.8: 94495100-94495100 on Build GRCh38
rs1016299850 Chr.8: 94494905-94494905 on Build GRCh38
rs546664585 Chr.8: 94495030-94495030 on Build GRCh38
rs926473217 Chr.8: 94495178-94495178 on Build GRCh38
rs146640304 Chr.8: 94495216-94495216 on Build GRCh38
rs776603028 Chr.8: 94495289-94495289 on Build GRCh38
rs889274149 Chr.8: 94495123-94495123 on Build GRCh38
rs747907081 Chr.8: 94495301-94495301 on Build GRCh38
rs948656038 Chr.8: 94495177-94495178 on Build GRCh38
rs1042000961 Chr.8: 94495081-94495081 on Build GRCh38
rs560146488 Chr.8: 94495244-94495244 on Build GRCh38
rs982036206 Chr.8: 94495177-94495177 on Build GRCh38
rs138979789 Chr.8: 94494946-94494946 on Build GRCh38
rs777019768 Chr.8: 94494900-94494900 on Build GRCh38
rs1007689018 Chr.8: 94495146-94495146 on Build GRCh38
rs993501003 Chr.8: 94495168-94495168 on Build GRCh38
rs945088011 Chr.8: 94495258-94495258 on Build GRCh38
rs926448013 Chr.8: 94495053-94495053 on Build GRCh38
rs191340662 Chr.8: 94495241-94495241 on Build GRCh38
rs555579607 Chr.8: 94495109-94495109 on Build GRCh38
rs1007741117 Chr.8: 94495353-94495353 on Build GRCh38
rs766241488 Chr.8: 94494975-94494975 on Build GRCh38
rs774280486 Chr.8: 94494977-94494977 on Build GRCh38
rs748361563 Chr.8: 94494928-94494928 on Build GRCh38
rs547987039 Chr.8: 94495063-94495063 on Build GRCh38
rs376087934 Chr.8: 94494966-94494966 on Build GRCh38
rs963372947 Chr.8: 94494932-94494932 on Build GRCh38
rs1042053476 Chr.8: 94495263-94495263 on Build GRCh38
rs148784427 Chr.8: 94495284-94495284 on Build GRCh38
rs937786374 Chr.8: 94495054-94495054 on Build GRCh38
rs972009796 Chr.8: 94494959-94494959 on Build GRCh38
rs971064266 Chr.8: 94495172-94495172 on Build GRCh38
rs112174315 Chr.8: 94494970-94494970 on Build GRCh38
rs1010910542 Chr.8: 94495279-94495279 on Build GRCh38
rs966198459 Chr.8: 94495365-94495365 on Build GRCh38
rs538062965 Chr.8: 94495170-94495170 on Build GRCh38

View Full Product Details