Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94726065-94726569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L4
Entrez Gene ID
286148
Gene Name
dpy-19 like 4 (C. elegans)
Gene Aliases
-
UniGene
Hs.567828
Gene Chromosome Location
Chr.8: 94719708-94793848 on Build GRCh38
Transcripts
NM_181787.2 XM_005250894.4 XM_005250895.3 XM_011516983.2 XM_011516984.2 XM_017013351.1 XM_017013352.1 XM_017013353.1 XM_017013354.1 XM_017013355.1 XM_017013356.1 XM_017013357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
606778 c.116G>A p.R39K Chr.8: 94726430-94726430 on Build GRCh38
4830139 c.85G>A p.E29K Chr.8: 94726399-94726399 on Build GRCh38
1489487 c.93C>G p.I31M Chr.8: 94726407-94726407 on Build GRCh38
5482225 c.35G>A p.R12H Chr.8: 94726349-94726349 on Build GRCh38
1731515 c.42delA p.K16fs*49 Chr.8: 94726356-94726356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986752971 Chr.8: 94726130-94726130 on Build GRCh38
rs772898672 Chr.8: 94726476-94726476 on Build GRCh38
rs922453974 Chr.8: 94726092-94726092 on Build GRCh38
rs773027960 Chr.8: 94726333-94726333 on Build GRCh38
rs942344999 Chr.8: 94726178-94726178 on Build GRCh38
rs765178039 Chr.8: 94726397-94726402 on Build GRCh38
rs780030433 Chr.8: 94726426-94726426 on Build GRCh38
rs747254361 Chr.8: 94726298-94726298 on Build GRCh38
rs769835284 Chr.8: 94726312-94726312 on Build GRCh38
rs138373475 Chr.8: 94726334-94726334 on Build GRCh38
rs780124871 Chr.8: 94726281-94726281 on Build GRCh38
rs758889948 Chr.8: 94726423-94726423 on Build GRCh38
rs926155360 Chr.8: 94726101-94726101 on Build GRCh38
rs762747708 Chr.8: 94726442-94726442 on Build GRCh38
rs748303092 Chr.8: 94726446-94726446 on Build GRCh38
rs759107741 Chr.8: 94726493-94726493 on Build GRCh38
rs754314433 Chr.8: 94726363-94726363 on Build GRCh38
rs769489216 Chr.8: 94726460-94726460 on Build GRCh38
rs202036194 Chr.8: 94726358-94726358 on Build GRCh38
rs371964466 Chr.8: 94726307-94726307 on Build GRCh38
rs1037974080 Chr.8: 94726165-94726165 on Build GRCh38
rs755250249 Chr.8: 94726438-94726438 on Build GRCh38
rs749673091 Chr.8: 94726355-94726356 on Build GRCh38
rs771539476 Chr.8: 94726443-94726444 on Build GRCh38
rs555224417 Chr.8: 94726212-94726212 on Build GRCh38
rs957562205 Chr.8: 94726116-94726116 on Build GRCh38
rs759299136 Chr.8: 94726356-94726356 on Build GRCh38
rs7816188 Chr.8: 94726135-94726135 on Build GRCh38
rs575038715 Chr.8: 94726218-94726218 on Build GRCh38
rs780352450 Chr.8: 94726291-94726291 on Build GRCh38
rs758415720 Chr.8: 94726280-94726280 on Build GRCh38
rs535116985 Chr.8: 94726369-94726369 on Build GRCh38
rs770366232 Chr.8: 94726348-94726348 on Build GRCh38
rs566090301 Chr.8: 94726536-94726536 on Build GRCh38
rs139917646 Chr.8: 94726442-94726442 on Build GRCh38
rs768828213 Chr.8: 94726299-94726299 on Build GRCh38
rs141766630 Chr.8: 94726361-94726361 on Build GRCh38
rs781006393 Chr.8: 94726302-94726302 on Build GRCh38
rs7816645 Chr.8: 94726489-94726489 on Build GRCh38
rs760205503 Chr.8: 94726475-94726476 on Build GRCh38
rs375072866 Chr.8: 94726406-94726406 on Build GRCh38
rs751608947 Chr.8: 94726435-94726435 on Build GRCh38
rs773956154 Chr.8: 94726349-94726349 on Build GRCh38
rs933434000 Chr.8: 94726100-94726100 on Build GRCh38
rs373521471 Chr.8: 94726476-94726476 on Build GRCh38
rs539492112 Chr.8: 94726450-94726450 on Build GRCh38
rs146012143 Chr.8: 94726392-94726392 on Build GRCh38
rs111530837 Chr.8: 94726474-94726474 on Build GRCh38
rs770867798 Chr.8: 94726488-94726488 on Build GRCh38
rs527594523 Chr.8: 94726514-94726514 on Build GRCh38
rs34813981 Chr.8: 94726514-94726515 on Build GRCh38
rs777571577 Chr.8: 94726464-94726464 on Build GRCh38
rs1040710303 Chr.8: 94726179-94726179 on Build GRCh38
rs369139666 Chr.8: 94726411-94726411 on Build GRCh38
rs374439628 Chr.8: 94726205-94726205 on Build GRCh38
rs1051833147 Chr.8: 94726129-94726129 on Build GRCh38
rs940703273 Chr.8: 94726527-94726527 on Build GRCh38
rs747687299 Chr.8: 94726442-94726443 on Build GRCh38

View Full Product Details