Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94927934-94928443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFAF6
Entrez Gene ID
137682
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases
C8orf38
UniGene
Hs.729144
Gene Chromosome Location
Chr.8: 94895271-95118496 on Build GRCh38
Transcripts
XM_011516835.2 XM_017013028.1 XM_017013030.1 XM_017013031.1 XM_017013032.1 XM_017013035.1 XM_017013036.1 XM_017013039.1 NM_001135733.1 NM_033285.3 XM_011517386.2

Gene Symbol
TP53INP1
Entrez Gene ID
94241
Gene Name
tumor protein p53 inducible nuclear protein 1
Gene Aliases
SIP, TP53DINP1, TP53INP1A, TP53INP1B, Teap, p53DINP1
UniGene
Hs.492261
Gene Chromosome Location
Chr.8: 94925972-94949387 on Build GRCh38
Transcripts
XM_011516835.2 XM_017013028.1 XM_017013030.1 XM_017013031.1 XM_017013032.1 XM_017013035.1 XM_017013036.1 XM_017013039.1 NM_001135733.1 NM_033285.3 XM_011517386.2

SNP Detail

SNP ID Chromosome Location
rs184020969 Chr.8: 94927994-94927994 on Build GRCh38
rs913473810 Chr.8: 94928127-94928129 on Build GRCh38
rs200462206 Chr.8: 94928249-94928249 on Build GRCh38
rs780848561 Chr.8: 94928373-94928374 on Build GRCh38
rs959972891 Chr.8: 94928401-94928401 on Build GRCh38
rs2904889 Chr.8: 94928370-94928370 on Build GRCh38
rs398008863 Chr.8: 94928249-94928249 on Build GRCh38
rs896939238 Chr.8: 94928364-94928364 on Build GRCh38
rs970000187 Chr.8: 94928081-94928081 on Build GRCh38
rs912123517 Chr.8: 94928325-94928325 on Build GRCh38
rs969084801 Chr.8: 94928037-94928037 on Build GRCh38
rs988812046 Chr.8: 94928212-94928212 on Build GRCh38
rs553926404 Chr.8: 94928152-94928152 on Build GRCh38
rs1016567522 Chr.8: 94928011-94928011 on Build GRCh38
rs769844313 Chr.8: 94928365-94928365 on Build GRCh38
rs534158762 Chr.8: 94928075-94928075 on Build GRCh38
rs978940019 Chr.8: 94928272-94928272 on Build GRCh38
rs541541853 Chr.8: 94928194-94928194 on Build GRCh38
rs77789511 Chr.8: 94928243-94928243 on Build GRCh38
rs575136211 Chr.8: 94928299-94928299 on Build GRCh38
rs192136125 Chr.8: 94928328-94928328 on Build GRCh38
rs3832569 Chr.8: 94928243-94928243 on Build GRCh38
rs777119035 Chr.8: 94928339-94928339 on Build GRCh38
rs1034770897 Chr.8: 94928420-94928420 on Build GRCh38
rs763270548 Chr.8: 94928386-94928386 on Build GRCh38
rs769219116 Chr.8: 94928324-94928324 on Build GRCh38
rs980490407 Chr.8: 94928046-94928046 on Build GRCh38
rs564267444 Chr.8: 94928362-94928362 on Build GRCh38
rs772361096 Chr.8: 94928085-94928085 on Build GRCh38
rs1039782530 Chr.8: 94928304-94928304 on Build GRCh38
rs571899086 Chr.8: 94927998-94927998 on Build GRCh38
rs749705317 Chr.8: 94928267-94928267 on Build GRCh38
rs747286090 Chr.8: 94928159-94928159 on Build GRCh38
rs780496947 Chr.8: 94928145-94928145 on Build GRCh38
rs995367385 Chr.8: 94927967-94927967 on Build GRCh38
rs913349828 Chr.8: 94928229-94928229 on Build GRCh38
rs927416335 Chr.8: 94928068-94928068 on Build GRCh38
rs932118117 Chr.8: 94928301-94928301 on Build GRCh38
rs186819647 Chr.8: 94928158-94928158 on Build GRCh38
rs1001857039 Chr.8: 94928413-94928413 on Build GRCh38
rs1052459343 Chr.8: 94928311-94928311 on Build GRCh38
rs922106473 Chr.8: 94928280-94928280 on Build GRCh38
rs766466459 Chr.8: 94928394-94928394 on Build GRCh38
rs544195470 Chr.8: 94928361-94928361 on Build GRCh38
rs943724879 Chr.8: 94928345-94928345 on Build GRCh38
rs998009733 Chr.8: 94928346-94928346 on Build GRCh38
rs146707005 Chr.8: 94928270-94928270 on Build GRCh38
rs139349422 Chr.8: 94928395-94928395 on Build GRCh38

View Full Product Details