Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96299521-96300054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTDSS1
Entrez Gene ID
9791
Gene Name
phosphatidylserine synthase 1
Gene Aliases
LMHD, PSS1, PSSA
UniGene
Hs.292579
Gene Chromosome Location
Chr.8: 96261886-96334552 on Build GRCh38
Transcripts
NM_001290225.1 NM_014754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758340 c.619C>A p.L207I Chr.8: 96299712-96299712 on Build GRCh38
2792162 c.642G>T p.W214C Chr.8: 96299735-96299735 on Build GRCh38
1102772 c.706G>A p.V236I Chr.8: 96299799-96299799 on Build GRCh38
284347 c.601-6T>C p.? Chr.8: 96299688-96299688 on Build GRCh38
5433227 c.673T>C p.C225R Chr.8: 96299766-96299766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559780966 Chr.8: 96299568-96299568 on Build GRCh38
rs768915542 Chr.8: 96299923-96299923 on Build GRCh38
rs781389525 Chr.8: 96299850-96299850 on Build GRCh38
rs111556920 Chr.8: 96299726-96299726 on Build GRCh38
rs764528912 Chr.8: 96299661-96299661 on Build GRCh38
rs758567950 Chr.8: 96299806-96299806 on Build GRCh38
rs1046859072 Chr.8: 96299985-96299985 on Build GRCh38
rs563168646 Chr.8: 96299631-96299631 on Build GRCh38
rs747274555 Chr.8: 96299813-96299813 on Build GRCh38
rs929743313 Chr.8: 96299930-96299930 on Build GRCh38
rs537784423 Chr.8: 96300003-96300003 on Build GRCh38
rs557568888 Chr.8: 96300029-96300029 on Build GRCh38
rs926279613 Chr.8: 96299639-96299639 on Build GRCh38
rs73271885 Chr.8: 96299602-96299602 on Build GRCh38
rs528031546 Chr.8: 96299907-96299907 on Build GRCh38
rs751115254 Chr.8: 96299651-96299651 on Build GRCh38
rs200369557 Chr.8: 96299674-96299674 on Build GRCh38
rs956453834 Chr.8: 96299935-96299935 on Build GRCh38
rs922312996 Chr.8: 96299929-96299929 on Build GRCh38
rs976445025 Chr.8: 96299807-96299807 on Build GRCh38
rs754291059 Chr.8: 96299793-96299793 on Build GRCh38
rs780067819 Chr.8: 96299706-96299706 on Build GRCh38
rs778071629 Chr.8: 96299940-96299940 on Build GRCh38
rs375896685 Chr.8: 96299774-96299774 on Build GRCh38
rs780471479 Chr.8: 96299607-96299607 on Build GRCh38
rs747976755 Chr.8: 96299885-96299885 on Build GRCh38
rs895126306 Chr.8: 96299576-96299576 on Build GRCh38
rs549546808 Chr.8: 96299623-96299623 on Build GRCh38
rs999665192 Chr.8: 96299642-96299642 on Build GRCh38
rs371518710 Chr.8: 96299759-96299759 on Build GRCh38
rs748373115 Chr.8: 96299768-96299768 on Build GRCh38
rs755007403 Chr.8: 96299843-96299843 on Build GRCh38
rs766157225 Chr.8: 96299775-96299775 on Build GRCh38
rs769774179 Chr.8: 96299893-96299893 on Build GRCh38
rs368785037 Chr.8: 96299799-96299799 on Build GRCh38
rs964046382 Chr.8: 96299805-96299805 on Build GRCh38
rs573114388 Chr.8: 96299587-96299587 on Build GRCh38
rs758944053 Chr.8: 96299784-96299784 on Build GRCh38
rs1017730495 Chr.8: 96299831-96299831 on Build GRCh38
rs897699454 Chr.8: 96299703-96299703 on Build GRCh38
rs530017132 Chr.8: 96299606-96299606 on Build GRCh38
rs375073869 Chr.8: 96299719-96299719 on Build GRCh38
rs201256591 Chr.8: 96299647-96299647 on Build GRCh38
rs911510872 Chr.8: 96299804-96299804 on Build GRCh38
rs767123710 Chr.8: 96299791-96299791 on Build GRCh38
rs776404022 Chr.8: 96299727-96299727 on Build GRCh38
rs1055663547 Chr.8: 96299543-96299543 on Build GRCh38
rs148193783 Chr.8: 96299990-96299990 on Build GRCh38
rs758628059 Chr.8: 96299699-96299699 on Build GRCh38
rs188668457 Chr.8: 96299597-96299597 on Build GRCh38
rs746035975 Chr.8: 96299677-96299677 on Build GRCh38
rs765524296 Chr.8: 96299798-96299798 on Build GRCh38
rs956229826 Chr.8: 96299579-96299579 on Build GRCh38
rs1037579579 Chr.8: 96299689-96299689 on Build GRCh38
rs756351104 Chr.8: 96299870-96299870 on Build GRCh38
rs568627570 Chr.8: 96300000-96300000 on Build GRCh38
rs756600447 Chr.8: 96299657-96299657 on Build GRCh38
rs1000235770 Chr.8: 96299926-96299926 on Build GRCh38
rs773154204 Chr.8: 96299762-96299762 on Build GRCh38
rs748182224 Chr.8: 96299729-96299729 on Build GRCh38
rs769462686 Chr.8: 96299753-96299753 on Build GRCh38
rs773947622 Chr.8: 96299777-96299777 on Build GRCh38
rs529347484 Chr.8: 96299732-96299732 on Build GRCh38

View Full Product Details