Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100158253-100158747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAG1
Entrez Gene ID
6674
Gene Name
sperm associated antigen 1
Gene Aliases
CILD28, CT140, HEL-S-268, HSD-3.8, SP75, TPIS
UniGene
Hs.591866
Gene Chromosome Location
Chr.8: 100157869-100241904 on Build GRCh38
Transcripts
NM_003114.4 NM_172218.2 XM_011517240.2 XM_011517241.2 XM_011517242.2 XM_011517245.2 XM_017013754.1 XM_017013755.1

SNP Detail

SNP ID Chromosome Location
rs949604509 Chr.8: 100158273-100158273 on Build GRCh38
rs995337261 Chr.8: 100158511-100158511 on Build GRCh38
rs949605844 Chr.8: 100158438-100158438 on Build GRCh38
rs1017091189 Chr.8: 100158490-100158490 on Build GRCh38
rs942483218 Chr.8: 100158351-100158351 on Build GRCh38
rs747149246 Chr.8: 100158286-100158286 on Build GRCh38
rs988338379 Chr.8: 100158723-100158723 on Build GRCh38
rs747443083 Chr.8: 100158527-100158527 on Build GRCh38
rs560773546 Chr.8: 100158287-100158287 on Build GRCh38
rs549505222 Chr.8: 100158465-100158465 on Build GRCh38
rs73279005 Chr.8: 100158333-100158333 on Build GRCh38
rs866494495 Chr.8: 100158694-100158694 on Build GRCh38
rs887033585 Chr.8: 100158497-100158497 on Build GRCh38
rs372363619 Chr.8: 100158446-100158446 on Build GRCh38
rs73279006 Chr.8: 100158692-100158692 on Build GRCh38
rs951295699 Chr.8: 100158299-100158299 on Build GRCh38
rs1025477565 Chr.8: 100158411-100158411 on Build GRCh38
rs995263694 Chr.8: 100158280-100158280 on Build GRCh38
rs1046861043 Chr.8: 100158534-100158534 on Build GRCh38
rs78139635 Chr.8: 100158614-100158614 on Build GRCh38
rs924205055 Chr.8: 100158313-100158313 on Build GRCh38
rs773465950 Chr.8: 100158685-100158685 on Build GRCh38
rs963717840 Chr.8: 100158332-100158332 on Build GRCh38
rs73696158 Chr.8: 100158577-100158577 on Build GRCh38
rs893546414 Chr.8: 100158350-100158350 on Build GRCh38
rs988307132 Chr.8: 100158416-100158416 on Build GRCh38
rs778631868 Chr.8: 100158526-100158526 on Build GRCh38
rs1025549480 Chr.8: 100158285-100158285 on Build GRCh38
rs997818673 Chr.8: 100158401-100158401 on Build GRCh38
rs1019684483 Chr.8: 100158323-100158323 on Build GRCh38
rs528962367 Chr.8: 100158645-100158645 on Build GRCh38
rs781363925 Chr.8: 100158594-100158594 on Build GRCh38
rs1005376853 Chr.8: 100158564-100158564 on Build GRCh38
rs867343876 Chr.8: 100158664-100158664 on Build GRCh38
rs768821614 Chr.8: 100158338-100158338 on Build GRCh38
rs1032547019 Chr.8: 100158672-100158672 on Build GRCh38
rs562597106 Chr.8: 100158489-100158489 on Build GRCh38
rs914181735 Chr.8: 100158427-100158427 on Build GRCh38
rs755557679 Chr.8: 100158591-100158591 on Build GRCh38
rs368259804 Chr.8: 100158642-100158642 on Build GRCh38
rs777324361 Chr.8: 100158389-100158389 on Build GRCh38
rs887053962 Chr.8: 100158552-100158552 on Build GRCh38
rs756812711 Chr.8: 100158501-100158501 on Build GRCh38
rs1007871047 Chr.8: 100158320-100158320 on Build GRCh38
rs1039326219 Chr.8: 100158363-100158363 on Build GRCh38
rs924181582 Chr.8: 100158670-100158670 on Build GRCh38
rs1038197750 Chr.8: 100158468-100158468 on Build GRCh38
rs1005589198 Chr.8: 100158450-100158450 on Build GRCh38
rs753723809 Chr.8: 100158306-100158306 on Build GRCh38
rs1023740143 Chr.8: 100158601-100158601 on Build GRCh38
rs900875144 Chr.8: 100158396-100158396 on Build GRCh38

View Full Product Details