Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100919675-100920205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAZ
Entrez Gene ID
7534
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
UniGene
Hs.492407
Gene Chromosome Location
Chr.8: 100918576-100954068 on Build GRCh38
Transcripts
NM_001135699.1 NM_001135700.1 NM_001135701.1 NM_001135702.1 NM_003406.3 NM_145690.2 XM_005251061.3 XM_005251063.3 XM_017013810.1 XM_017013811.1

SNP Detail

SNP ID Chromosome Location
rs369444018 Chr.8: 100919869-100919870 on Build GRCh38
rs918650233 Chr.8: 100919904-100919904 on Build GRCh38
rs140806054 Chr.8: 100920129-100920130 on Build GRCh38
rs3921614 Chr.8: 100919898-100919898 on Build GRCh38
rs568595016 Chr.8: 100919949-100919949 on Build GRCh38
rs770981479 Chr.8: 100919883-100919884 on Build GRCh38
rs941462794 Chr.8: 100919901-100919902 on Build GRCh38
rs183659599 Chr.8: 100919819-100919819 on Build GRCh38
rs1013459575 Chr.8: 100920049-100920050 on Build GRCh38
rs918715841 Chr.8: 100920118-100920118 on Build GRCh38
rs142007434 Chr.8: 100919870-100919871 on Build GRCh38
rs1040002020 Chr.8: 100920164-100920164 on Build GRCh38
rs869207861 Chr.8: 100919876-100919879 on Build GRCh38
rs903495322 Chr.8: 100919993-100919993 on Build GRCh38
rs942948451 Chr.8: 100920152-100920152 on Build GRCh38
rs1051695009 Chr.8: 100919953-100919953 on Build GRCh38
rs969286537 Chr.8: 100920062-100920062 on Build GRCh38
rs930190713 Chr.8: 100919952-100919952 on Build GRCh38
rs879002446 Chr.8: 100920132-100920132 on Build GRCh38
rs565389081 Chr.8: 100920127-100920127 on Build GRCh38
rs570983777 Chr.8: 100919744-100919744 on Build GRCh38
rs554084123 Chr.8: 100920169-100920169 on Build GRCh38
rs912692387 Chr.8: 100920143-100920143 on Build GRCh38
rs974469560 Chr.8: 100920109-100920109 on Build GRCh38
rs1038396886 Chr.8: 100919902-100919902 on Build GRCh38
rs551539483 Chr.8: 100920106-100920106 on Build GRCh38
rs866800209 Chr.8: 100919883-100919883 on Build GRCh38
rs866388383 Chr.8: 100919869-100919869 on Build GRCh38
rs904090601 Chr.8: 100920167-100920167 on Build GRCh38
rs962709599 Chr.8: 100920108-100920108 on Build GRCh38
rs531619161 Chr.8: 100919968-100919968 on Build GRCh38
rs758882848 Chr.8: 100920030-100920030 on Build GRCh38
rs969709394 Chr.8: 100919817-100919821 on Build GRCh38
rs917874228 Chr.8: 100919785-100919785 on Build GRCh38
rs956590334 Chr.8: 100919769-100919769 on Build GRCh38
rs891378705 Chr.8: 100919967-100919967 on Build GRCh38
rs193223230 Chr.8: 100919774-100919774 on Build GRCh38
rs1039398730 Chr.8: 100919980-100919980 on Build GRCh38
rs927263604 Chr.8: 100919870-100919870 on Build GRCh38
rs866960160 Chr.8: 100919765-100919765 on Build GRCh38
rs980628006 Chr.8: 100920077-100920077 on Build GRCh38
rs1000586295 Chr.8: 100919743-100919743 on Build GRCh38
rs4085567 Chr.8: 100919828-100919828 on Build GRCh38
rs868532393 Chr.8: 100919884-100919884 on Build GRCh38
rs535786363 Chr.8: 100919954-100919954 on Build GRCh38
rs986992388 Chr.8: 100920136-100920136 on Build GRCh38
rs58344290 Chr.8: 100919882-100919883 on Build GRCh38
rs4003432 Chr.8: 100919959-100919959 on Build GRCh38
rs570522311 Chr.8: 100920038-100920042 on Build GRCh38
rs1827729 Chr.8: 100919779-100919779 on Build GRCh38
rs1025190009 Chr.8: 100920061-100920061 on Build GRCh38
rs1000514429 Chr.8: 100920027-100920027 on Build GRCh38

View Full Product Details