Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109241506-109242050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDCD1
Entrez Gene ID
84955
Gene Name
NudC domain containing 1
Gene Aliases
CML66, OVA66
UniGene
Hs.380291
Gene Chromosome Location
Chr.8: 109240919-109334205 on Build GRCh38
Transcripts
NM_001128211.1 NM_032869.3 XM_017013910.1 XM_017013911.1

SNP Detail

SNP ID Chromosome Location
rs138607977 Chr.8: 109241621-109241621 on Build GRCh38
rs66767511 Chr.8: 109241702-109241703 on Build GRCh38
rs984180574 Chr.8: 109241551-109241551 on Build GRCh38
rs1053262343 Chr.8: 109241763-109241763 on Build GRCh38
rs779721767 Chr.8: 109241786-109241786 on Build GRCh38
rs904706260 Chr.8: 109241674-109241674 on Build GRCh38
rs754610086 Chr.8: 109241858-109241858 on Build GRCh38
rs1046521757 Chr.8: 109241942-109241942 on Build GRCh38
rs183347407 Chr.8: 109241716-109241716 on Build GRCh38
rs67919153 Chr.8: 109241928-109241928 on Build GRCh38
rs192205359 Chr.8: 109241587-109241587 on Build GRCh38
rs926146290 Chr.8: 109241882-109241882 on Build GRCh38
rs71564038 Chr.8: 109241701-109241702 on Build GRCh38
rs927866006 Chr.8: 109241968-109241968 on Build GRCh38
rs201346435 Chr.8: 109241698-109241698 on Build GRCh38
rs553339982 Chr.8: 109241835-109241835 on Build GRCh38
rs936461973 Chr.8: 109241570-109241570 on Build GRCh38
rs781565185 Chr.8: 109241626-109241626 on Build GRCh38
rs79117529 Chr.8: 109241656-109241656 on Build GRCh38
rs1028579490 Chr.8: 109241532-109241532 on Build GRCh38
rs1548083 Chr.8: 109241650-109241650 on Build GRCh38
rs922862652 Chr.8: 109241534-109241534 on Build GRCh38
rs369500673 Chr.8: 109241852-109241852 on Build GRCh38
rs374987106 Chr.8: 109241709-109241709 on Build GRCh38
rs892251062 Chr.8: 109241781-109241781 on Build GRCh38
rs906452575 Chr.8: 109241602-109241602 on Build GRCh38
rs947712783 Chr.8: 109241941-109241941 on Build GRCh38
rs929903344 Chr.8: 109241995-109241995 on Build GRCh38
rs957769903 Chr.8: 109241884-109241884 on Build GRCh38
rs993996167 Chr.8: 109241620-109241620 on Build GRCh38
rs867768447 Chr.8: 109241579-109241579 on Build GRCh38
rs917241411 Chr.8: 109242024-109242024 on Build GRCh38
rs760865579 Chr.8: 109241681-109241681 on Build GRCh38
rs1041150427 Chr.8: 109241788-109241788 on Build GRCh38
rs977566928 Chr.8: 109241554-109241554 on Build GRCh38
rs377135321 Chr.8: 109241701-109241701 on Build GRCh38
rs539453615 Chr.8: 109241670-109241670 on Build GRCh38
rs72682627 Chr.8: 109241630-109241630 on Build GRCh38
rs947731559 Chr.8: 109241589-109241589 on Build GRCh38
rs543093649 Chr.8: 109241822-109241822 on Build GRCh38
rs992923606 Chr.8: 109241944-109241944 on Build GRCh38
rs868077964 Chr.8: 109241837-109241837 on Build GRCh38
rs34259822 Chr.8: 109241706-109241707 on Build GRCh38
rs923683045 Chr.8: 109241559-109241559 on Build GRCh38
rs386413654 Chr.8: 109241704-109241705 on Build GRCh38
rs1046273609 Chr.8: 109241595-109241595 on Build GRCh38
rs572724224 Chr.8: 109241946-109241946 on Build GRCh38
rs5893946 Chr.8: 109241707-109241708 on Build GRCh38
rs548333158 Chr.8: 109241597-109241597 on Build GRCh38
rs570158210 Chr.8: 109241553-109241553 on Build GRCh38
rs916319296 Chr.8: 109241931-109241931 on Build GRCh38
rs778676252 Chr.8: 109241869-109241869 on Build GRCh38

View Full Product Details