Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109364452-109364997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKHD1L1
Entrez Gene ID
93035
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
Gene Aliases
PKHDL1
UniGene
Hs.170128
Gene Chromosome Location
Chr.8: 109362477-109536809 on Build GRCh38
Transcripts
NM_177531.4 XM_011517371.2 XM_017013969.1 XM_017013970.1 XM_017013971.1 XM_017013972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4976169 c.80C>T p.S27F Chr.8: 109364553-109364553 on Build GRCh38
326162 c.85A>T p.I29L Chr.8: 109364558-109364558 on Build GRCh38
125398 c.140C>A p.T47K Chr.8: 109364613-109364613 on Build GRCh38
3271680 c.90C>T p.I30I Chr.8: 109364563-109364563 on Build GRCh38
1094833 c.119A>G p.Y40C Chr.8: 109364592-109364592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042539247 Chr.8: 109364961-109364961 on Build GRCh38
rs752873795 Chr.8: 109364674-109364674 on Build GRCh38
rs750469571 Chr.8: 109364664-109364665 on Build GRCh38
rs765434737 Chr.8: 109364669-109364669 on Build GRCh38
rs911488289 Chr.8: 109364818-109364818 on Build GRCh38
rs778283728 Chr.8: 109364558-109364558 on Build GRCh38
rs778493904 Chr.8: 109364616-109364616 on Build GRCh38
rs765237223 Chr.8: 109364646-109364647 on Build GRCh38
rs766480817 Chr.8: 109364527-109364527 on Build GRCh38
rs565247810 Chr.8: 109364891-109364891 on Build GRCh38
rs77973025 Chr.8: 109364878-109364878 on Build GRCh38
rs779386774 Chr.8: 109364680-109364680 on Build GRCh38
rs766532172 Chr.8: 109364665-109364679 on Build GRCh38
rs759680709 Chr.8: 109364656-109364656 on Build GRCh38
rs1020698594 Chr.8: 109364536-109364536 on Build GRCh38
rs748852355 Chr.8: 109364596-109364596 on Build GRCh38
rs992611337 Chr.8: 109364763-109364763 on Build GRCh38
rs752029351 Chr.8: 109364572-109364572 on Build GRCh38
rs755374884 Chr.8: 109364674-109364679 on Build GRCh38
rs201083009 Chr.8: 109364642-109364642 on Build GRCh38
rs935967293 Chr.8: 109364904-109364904 on Build GRCh38
rs370157617 Chr.8: 109364531-109364531 on Build GRCh38
rs753103239 Chr.8: 109364545-109364545 on Build GRCh38
rs753960366 Chr.8: 109364528-109364528 on Build GRCh38
rs770525586 Chr.8: 109364647-109364648 on Build GRCh38
rs970548013 Chr.8: 109364691-109364691 on Build GRCh38
rs565296426 Chr.8: 109364758-109364758 on Build GRCh38
rs796184362 Chr.8: 109364664-109364665 on Build GRCh38
rs183558972 Chr.8: 109364848-109364848 on Build GRCh38
rs901337871 Chr.8: 109364505-109364505 on Build GRCh38
rs765437609 Chr.8: 109364533-109364533 on Build GRCh38
rs918646916 Chr.8: 109364739-109364739 on Build GRCh38
rs1015725061 Chr.8: 109364690-109364690 on Build GRCh38
rs776679409 Chr.8: 109364645-109364645 on Build GRCh38
rs779675720 Chr.8: 109364586-109364586 on Build GRCh38
rs554313501 Chr.8: 109364753-109364753 on Build GRCh38
rs60902563 Chr.8: 109364663-109364664 on Build GRCh38
rs760700522 Chr.8: 109364519-109364519 on Build GRCh38
rs773165419 Chr.8: 109364637-109364637 on Build GRCh38
rs746608602 Chr.8: 109364645-109364646 on Build GRCh38
rs946227315 Chr.8: 109364896-109364896 on Build GRCh38
rs970238768 Chr.8: 109364786-109364786 on Build GRCh38
rs942984724 Chr.8: 109364858-109364858 on Build GRCh38
rs973263978 Chr.8: 109364706-109364706 on Build GRCh38
rs773038542 Chr.8: 109364518-109364518 on Build GRCh38
rs574167063 Chr.8: 109364835-109364835 on Build GRCh38
rs755645795 Chr.8: 109364585-109364585 on Build GRCh38
rs902220670 Chr.8: 109364962-109364962 on Build GRCh38
rs936035148 Chr.8: 109364496-109364496 on Build GRCh38
rs776438150 Chr.8: 109364647-109364647 on Build GRCh38
rs772613805 Chr.8: 109364539-109364539 on Build GRCh38
rs961119632 Chr.8: 109364713-109364714 on Build GRCh38
rs767483127 Chr.8: 109364498-109364498 on Build GRCh38
rs868630554 Chr.8: 109364880-109364880 on Build GRCh38
rs879186893 Chr.8: 109364646-109364646 on Build GRCh38
rs931311108 Chr.8: 109364774-109364774 on Build GRCh38
rs7008687 Chr.8: 109364632-109364632 on Build GRCh38
rs984116024 Chr.8: 109364779-109364779 on Build GRCh38
rs1032674311 Chr.8: 109364624-109364624 on Build GRCh38
rs767573075 Chr.8: 109364650-109364651 on Build GRCh38
rs926240648 Chr.8: 109364886-109364886 on Build GRCh38
rs758692922 Chr.8: 109364548-109364548 on Build GRCh38
rs747921989 Chr.8: 109364617-109364617 on Build GRCh38
rs530768459 Chr.8: 109364917-109364917 on Build GRCh38
rs775638541 Chr.8: 109364647-109364652 on Build GRCh38
rs760578258 Chr.8: 109364641-109364641 on Build GRCh38
rs759454281 Chr.8: 109364647-109364653 on Build GRCh38
rs760906805 Chr.8: 109364665-109364671 on Build GRCh38
rs901388568 Chr.8: 109364883-109364883 on Build GRCh38

View Full Product Details