Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109968524-109969031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6179437 c.1046T>C p.L349P Chr.8: 109968545-109968545 on Build GRCh38
4587734 c.1020C>T p.T340T Chr.8: 109968571-109968571 on Build GRCh38
6889220 c.1039G>T p.G347C Chr.8: 109968552-109968552 on Build GRCh38
748743 c.997C>A p.R333S Chr.8: 109968594-109968594 on Build GRCh38
3644128 c.1061C>T p.S354F Chr.8: 109968530-109968530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182361247 Chr.8: 109968800-109968800 on Build GRCh38
rs372723430 Chr.8: 109968666-109968666 on Build GRCh38
rs1013980642 Chr.8: 109968719-109968719 on Build GRCh38
rs754470123 Chr.8: 109968562-109968562 on Build GRCh38
rs924752395 Chr.8: 109968997-109968997 on Build GRCh38
rs535764021 Chr.8: 109968882-109968882 on Build GRCh38
rs537666413 Chr.8: 109968623-109968623 on Build GRCh38
rs896582189 Chr.8: 109968688-109968688 on Build GRCh38
rs538457181 Chr.8: 109968970-109968970 on Build GRCh38
rs542448308 Chr.8: 109968926-109968926 on Build GRCh38
rs562126186 Chr.8: 109968951-109968951 on Build GRCh38
rs778526971 Chr.8: 109968574-109968574 on Build GRCh38
rs762914494 Chr.8: 109968638-109968638 on Build GRCh38
rs761982695 Chr.8: 109968650-109968650 on Build GRCh38
rs757979003 Chr.8: 109968549-109968549 on Build GRCh38
rs551457160 Chr.8: 109968589-109968589 on Build GRCh38
rs202237668 Chr.8: 109968647-109968647 on Build GRCh38
rs531911056 Chr.8: 109968887-109968887 on Build GRCh38
rs769648442 Chr.8: 109968619-109968619 on Build GRCh38
rs140828945 Chr.8: 109968864-109968864 on Build GRCh38
rs771779153 Chr.8: 109968760-109968760 on Build GRCh38
rs1052773498 Chr.8: 109968720-109968720 on Build GRCh38
rs759617731 Chr.8: 109968994-109968994 on Build GRCh38
rs957156311 Chr.8: 109968818-109968818 on Build GRCh38
rs747735274 Chr.8: 109968604-109968604 on Build GRCh38
rs962948341 Chr.8: 109968912-109968912 on Build GRCh38
rs767420033 Chr.8: 109968661-109968661 on Build GRCh38
rs1017593674 Chr.8: 109968910-109968910 on Build GRCh38
rs932221954 Chr.8: 109968874-109968874 on Build GRCh38
rs549654569 Chr.8: 109968965-109968965 on Build GRCh38
rs563210153 Chr.8: 109968872-109968872 on Build GRCh38
rs975522259 Chr.8: 109968921-109968921 on Build GRCh38
rs868751450 Chr.8: 109968561-109968561 on Build GRCh38
rs780932230 Chr.8: 109968578-109968578 on Build GRCh38
rs1036507252 Chr.8: 109968941-109968941 on Build GRCh38
rs1003302818 Chr.8: 109968854-109968854 on Build GRCh38
rs934088954 Chr.8: 109968603-109968603 on Build GRCh38
rs6980482 Chr.8: 109968831-109968831 on Build GRCh38
rs1032845088 Chr.8: 109968795-109968795 on Build GRCh38
rs574203182 Chr.8: 109968722-109968722 on Build GRCh38
rs569312220 Chr.8: 109968966-109968966 on Build GRCh38
rs1010726249 Chr.8: 109968744-109968744 on Build GRCh38
rs988654186 Chr.8: 109968827-109968827 on Build GRCh38
rs1036371726 Chr.8: 109968686-109968686 on Build GRCh38
rs372536889 Chr.8: 109968983-109968983 on Build GRCh38
rs373862719 Chr.8: 109968582-109968582 on Build GRCh38
rs746808311 Chr.8: 109968889-109968889 on Build GRCh38
rs576799149 Chr.8: 109968851-109968851 on Build GRCh38
rs768697423 Chr.8: 109968645-109968645 on Build GRCh38

View Full Product Details