Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120536380-120536896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNTB1
Entrez Gene ID
6641
Gene Name
syntrophin beta 1
Gene Aliases
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43
UniGene
Hs.46701
Gene Chromosome Location
Chr.8: 120535745-120812069 on Build GRCh38
Transcripts
NM_021021.3

SNP Detail

SNP ID Chromosome Location
rs949737573 Chr.8: 120536579-120536579 on Build GRCh38
rs889101921 Chr.8: 120536804-120536804 on Build GRCh38
rs950193152 Chr.8: 120536744-120536744 on Build GRCh38
rs1019060140 Chr.8: 120536563-120536565 on Build GRCh38
rs950419353 Chr.8: 120536522-120536522 on Build GRCh38
rs1054754055 Chr.8: 120536795-120536795 on Build GRCh38
rs1018024573 Chr.8: 120536777-120536777 on Build GRCh38
rs566386721 Chr.8: 120536801-120536801 on Build GRCh38
rs547010084 Chr.8: 120536745-120536745 on Build GRCh38
rs950882726 Chr.8: 120536845-120536845 on Build GRCh38
rs986211947 Chr.8: 120536541-120536541 on Build GRCh38
rs768907936 Chr.8: 120536449-120536449 on Build GRCh38
rs903447597 Chr.8: 120536451-120536451 on Build GRCh38
rs1028255539 Chr.8: 120536513-120536513 on Build GRCh38
rs558427823 Chr.8: 120536627-120536631 on Build GRCh38
rs532890025 Chr.8: 120536679-120536679 on Build GRCh38
rs767280189 Chr.8: 120536742-120536742 on Build GRCh38
rs898942854 Chr.8: 120536810-120536810 on Build GRCh38
rs1007592369 Chr.8: 120536836-120536836 on Build GRCh38
rs1027811053 Chr.8: 120536800-120536800 on Build GRCh38
rs551354252 Chr.8: 120536709-120536709 on Build GRCh38
rs937719866 Chr.8: 120536681-120536684 on Build GRCh38
rs995510828 Chr.8: 120536787-120536787 on Build GRCh38
rs980417554 Chr.8: 120536599-120536599 on Build GRCh38
rs953438791 Chr.8: 120536533-120536533 on Build GRCh38
rs1056123503 Chr.8: 120536711-120536711 on Build GRCh38
rs965340375 Chr.8: 120536489-120536489 on Build GRCh38
rs952060044 Chr.8: 120536651-120536651 on Build GRCh38
rs945444976 Chr.8: 120536733-120536733 on Build GRCh38
rs774541286 Chr.8: 120536569-120536569 on Build GRCh38
rs983195061 Chr.8: 120536848-120536848 on Build GRCh38
rs973556246 Chr.8: 120536493-120536493 on Build GRCh38
rs772637590 Chr.8: 120536718-120536719 on Build GRCh38
rs567547602 Chr.8: 120536564-120536564 on Build GRCh38
rs927195533 Chr.8: 120536621-120536621 on Build GRCh38
rs903399778 Chr.8: 120536772-120536775 on Build GRCh38
rs1020523163 Chr.8: 120536405-120536405 on Build GRCh38
rs141703987 Chr.8: 120536866-120536866 on Build GRCh38
rs900728927 Chr.8: 120536779-120536779 on Build GRCh38
rs534952774 Chr.8: 120536666-120536666 on Build GRCh38
rs1017890685 Chr.8: 120536479-120536479 on Build GRCh38
rs1042489968 Chr.8: 120536741-120536741 on Build GRCh38
rs891661430 Chr.8: 120536683-120536683 on Build GRCh38
rs1000847398 Chr.8: 120536793-120536795 on Build GRCh38

View Full Product Details