Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123014547-123015043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL1
Entrez Gene ID
79139
Gene Name
derlin 1
Gene Aliases
DER-1, DER1
UniGene
Hs.241576
Gene Chromosome Location
Chr.8: 123013164-123042423 on Build GRCh38
Transcripts
NM_001134671.2 NM_024295.5 XM_006716657.1 XM_011517299.1

SNP Detail

SNP ID Chromosome Location
rs943520827 Chr.8: 123014648-123014648 on Build GRCh38
rs112674565 Chr.8: 123014906-123014906 on Build GRCh38
rs1059588 Chr.8: 123014660-123014660 on Build GRCh38
rs939151270 Chr.8: 123014825-123014825 on Build GRCh38
rs1008943889 Chr.8: 123014943-123014943 on Build GRCh38
rs527988779 Chr.8: 123014683-123014683 on Build GRCh38
rs971651500 Chr.8: 123014728-123014728 on Build GRCh38
rs536616453 Chr.8: 123014846-123014846 on Build GRCh38
rs887520903 Chr.8: 123014677-123014677 on Build GRCh38
rs1040296404 Chr.8: 123014871-123014871 on Build GRCh38
rs868637062 Chr.8: 123014580-123014580 on Build GRCh38
rs1012640841 Chr.8: 123014924-123014924 on Build GRCh38
rs145083623 Chr.8: 123014574-123014576 on Build GRCh38
rs568892297 Chr.8: 123014638-123014638 on Build GRCh38
rs950852973 Chr.8: 123014620-123014620 on Build GRCh38
rs189232241 Chr.8: 123014898-123014898 on Build GRCh38
rs946353470 Chr.8: 123014676-123014676 on Build GRCh38
rs950854280 Chr.8: 123014941-123014941 on Build GRCh38
rs1027439638 Chr.8: 123014675-123014675 on Build GRCh38
rs548099966 Chr.8: 123014685-123014685 on Build GRCh38
rs995227737 Chr.8: 123014921-123014921 on Build GRCh38
rs985439098 Chr.8: 123014845-123014845 on Build GRCh38
rs549931976 Chr.8: 123014881-123014881 on Build GRCh38
rs577459804 Chr.8: 123014968-123014968 on Build GRCh38
rs191870872 Chr.8: 123014590-123014590 on Build GRCh38
rs571453607 Chr.8: 123014954-123014954 on Build GRCh38
rs1008739825 Chr.8: 123014574-123014574 on Build GRCh38
rs760530776 Chr.8: 123014741-123014741 on Build GRCh38
rs556695276 Chr.8: 123014625-123014625 on Build GRCh38
rs749321459 Chr.8: 123014834-123014834 on Build GRCh38
rs376617688 Chr.8: 123014850-123014850 on Build GRCh38
rs762625165 Chr.8: 123014694-123014694 on Build GRCh38
rs183576997 Chr.8: 123014743-123014743 on Build GRCh38
rs1031916242 Chr.8: 123014778-123014778 on Build GRCh38
rs932694054 Chr.8: 123014842-123014842 on Build GRCh38
rs944024018 Chr.8: 123014861-123014861 on Build GRCh38
rs988032022 Chr.8: 123014627-123014627 on Build GRCh38
rs551964270 Chr.8: 123014653-123014653 on Build GRCh38
rs990723314 Chr.8: 123014833-123014833 on Build GRCh38
rs981665736 Chr.8: 123014750-123014750 on Build GRCh38
rs1026343325 Chr.8: 123014936-123014936 on Build GRCh38
rs1016776863 Chr.8: 123014889-123014889 on Build GRCh38
rs897727360 Chr.8: 123014880-123014880 on Build GRCh38
rs974957939 Chr.8: 123014659-123014659 on Build GRCh38
rs1049330323 Chr.8: 123014888-123014888 on Build GRCh38
rs956267558 Chr.8: 123014828-123014828 on Build GRCh38

View Full Product Details