Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142879484-142879978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B1
Entrez Gene ID
1584
Gene Name
cytochrome P450 family 11 subfamily B member 1
Gene Aliases
CPN1, CYP11B, FHI, P450C11
UniGene
Hs.184927
Gene Chromosome Location
Chr.8: 142872354-142879839 on Build GRCh38
Transcripts
NM_000497.3 NM_001026213.1 XM_011516874.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866756 c.74G>A p.G25D Chr.8: 142879740-142879740 on Build GRCh38
268973 c.203T>C p.V68A Chr.8: 142879611-142879611 on Build GRCh38
3924770 c.66G>A p.Q22Q Chr.8: 142879748-142879748 on Build GRCh38
1455294 c.77C>T p.T26M Chr.8: 142879737-142879737 on Build GRCh38
5358702 c.154C>T p.L52L Chr.8: 142879660-142879660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768007725 Chr.8: 142879718-142879718 on Build GRCh38
rs754860366 Chr.8: 142879842-142879842 on Build GRCh38
rs889717829 Chr.8: 142879901-142879901 on Build GRCh38
rs757776355 Chr.8: 142879528-142879528 on Build GRCh38
rs906336753 Chr.8: 142879730-142879730 on Build GRCh38
rs776199363 Chr.8: 142879824-142879824 on Build GRCh38
rs771206231 Chr.8: 142879741-142879741 on Build GRCh38
rs370830923 Chr.8: 142879549-142879549 on Build GRCh38
rs373827432 Chr.8: 142879833-142879833 on Build GRCh38
rs397831740 Chr.8: 142879703-142879703 on Build GRCh38
rs777199047 Chr.8: 142879807-142879807 on Build GRCh38
rs1049635142 Chr.8: 142879887-142879887 on Build GRCh38
rs763195324 Chr.8: 142879759-142879759 on Build GRCh38
rs745709636 Chr.8: 142879660-142879660 on Build GRCh38
rs748062304 Chr.8: 142879803-142879803 on Build GRCh38
rs746696910 Chr.8: 142879810-142879810 on Build GRCh38
rs756034651 Chr.8: 142879728-142879728 on Build GRCh38
rs759984716 Chr.8: 142879751-142879751 on Build GRCh38
rs6405 Chr.8: 142879785-142879785 on Build GRCh38
rs753712322 Chr.8: 142879775-142879775 on Build GRCh38
rs749188499 Chr.8: 142879672-142879672 on Build GRCh38
rs147788769 Chr.8: 142879736-142879736 on Build GRCh38
rs1042033744 Chr.8: 142879875-142879875 on Build GRCh38
rs760261276 Chr.8: 142879739-142879739 on Build GRCh38
rs754364294 Chr.8: 142879641-142879641 on Build GRCh38
rs201103987 Chr.8: 142879725-142879725 on Build GRCh38
rs750604556 Chr.8: 142879840-142879840 on Build GRCh38
rs778976411 Chr.8: 142879786-142879786 on Build GRCh38
rs775480247 Chr.8: 142879613-142879613 on Build GRCh38
rs762693458 Chr.8: 142879545-142879545 on Build GRCh38
rs773886880 Chr.8: 142879624-142879624 on Build GRCh38
rs768052570 Chr.8: 142879676-142879676 on Build GRCh38
rs985479574 Chr.8: 142879876-142879876 on Build GRCh38
rs748867146 Chr.8: 142879667-142879667 on Build GRCh38
rs763658052 Chr.8: 142879618-142879618 on Build GRCh38
rs368794827 Chr.8: 142879530-142879530 on Build GRCh38
rs751833268 Chr.8: 142879765-142879765 on Build GRCh38
rs779731828 Chr.8: 142879774-142879774 on Build GRCh38
rs369941128 Chr.8: 142879705-142879705 on Build GRCh38
rs757512473 Chr.8: 142879772-142879772 on Build GRCh38
rs765826051 Chr.8: 142879753-142879753 on Build GRCh38
rs539054150 Chr.8: 142879839-142879839 on Build GRCh38
rs768585386 Chr.8: 142879734-142879734 on Build GRCh38
rs928735201 Chr.8: 142879915-142879915 on Build GRCh38
rs751195976 Chr.8: 142879553-142879553 on Build GRCh38
rs759989671 Chr.8: 142879820-142879820 on Build GRCh38
rs981515612 Chr.8: 142879902-142879902 on Build GRCh38
rs776502555 Chr.8: 142879607-142879607 on Build GRCh38
rs750733169 Chr.8: 142879631-142879631 on Build GRCh38
rs933656667 Chr.8: 142879882-142879882 on Build GRCh38
rs142591816 Chr.8: 142879764-142879764 on Build GRCh38
rs770569014 Chr.8: 142879816-142879816 on Build GRCh38
rs370524170 Chr.8: 142879509-142879509 on Build GRCh38
rs372954252 Chr.8: 142879847-142879847 on Build GRCh38
rs200651334 Chr.8: 142879536-142879536 on Build GRCh38
rs766911727 Chr.8: 142879639-142879639 on Build GRCh38
rs755069041 Chr.8: 142879642-142879642 on Build GRCh38
rs1038641306 Chr.8: 142879936-142879936 on Build GRCh38
rs759489306 Chr.8: 142879532-142879532 on Build GRCh38
rs750931191 Chr.8: 142879585-142879585 on Build GRCh38
rs760426236 Chr.8: 142879524-142879524 on Build GRCh38
rs1026508780 Chr.8: 142879855-142879855 on Build GRCh38
rs762414385 Chr.8: 142879615-142879615 on Build GRCh38
rs780130248 Chr.8: 142879586-142879586 on Build GRCh38
rs769810247 Chr.8: 142879612-142879612 on Build GRCh38
rs139569725 Chr.8: 142879737-142879737 on Build GRCh38
rs761472886 Chr.8: 142879684-142879684 on Build GRCh38
rs140274628 Chr.8: 142879726-142879726 on Build GRCh38
rs774466832 Chr.8: 142879866-142879866 on Build GRCh38
rs752773985 Chr.8: 142879720-142879720 on Build GRCh38
rs150163594 Chr.8: 142879789-142879789 on Build GRCh38
rs754316010 Chr.8: 142879702-142879702 on Build GRCh38
rs780398462 Chr.8: 142879615-142879615 on Build GRCh38
rs754138797 Chr.8: 142879512-142879512 on Build GRCh38
rs61751134 Chr.8: 142879760-142879760 on Build GRCh38
rs4534 Chr.8: 142879686-142879686 on Build GRCh38
rs754243278 Chr.8: 142879616-142879616 on Build GRCh38
rs796443111 Chr.8: 142879535-142879535 on Build GRCh38
rs892400238 Chr.8: 142879761-142879761 on Build GRCh38
rs777288901 Chr.8: 142879856-142879856 on Build GRCh38
rs1005365266 Chr.8: 142879932-142879932 on Build GRCh38
rs61751135 Chr.8: 142879657-142879657 on Build GRCh38
rs755448048 Chr.8: 142879707-142879707 on Build GRCh38
rs764297218 Chr.8: 142879829-142879829 on Build GRCh38
rs371662064 Chr.8: 142879596-142879596 on Build GRCh38
rs104894069 Chr.8: 142879690-142879690 on Build GRCh38
rs6410 Chr.8: 142879589-142879589 on Build GRCh38
rs756448189 Chr.8: 142879637-142879637 on Build GRCh38
rs6389 Chr.8: 142879815-142879815 on Build GRCh38
rs778994889 Chr.8: 142879647-142879647 on Build GRCh38
rs774074506 Chr.8: 142879789-142879789 on Build GRCh38
rs566126580 Chr.8: 142879940-142879940 on Build GRCh38
rs758653314 Chr.8: 142879655-142879655 on Build GRCh38
rs6388 Chr.8: 142879555-142879555 on Build GRCh38
rs200952801 Chr.8: 142879683-142879683 on Build GRCh38
rs5280 Chr.8: 142879877-142879877 on Build GRCh38
rs747743740 Chr.8: 142879823-142879823 on Build GRCh38
rs763499747 Chr.8: 142879826-142879826 on Build GRCh38
rs770053556 Chr.8: 142879823-142879823 on Build GRCh38
rs6402 Chr.8: 142879562-142879562 on Build GRCh38
rs752633749 Chr.8: 142879843-142879843 on Build GRCh38
rs201951316 Chr.8: 142879710-142879710 on Build GRCh38
rs745832184 Chr.8: 142879610-142879610 on Build GRCh38
rs372674759 Chr.8: 142879537-142879537 on Build GRCh38
rs753228911 Chr.8: 142879757-142879757 on Build GRCh38
rs200096159 Chr.8: 142879592-142879592 on Build GRCh38
rs747434849 Chr.8: 142879783-142879783 on Build GRCh38
rs749551361 Chr.8: 142879514-142879514 on Build GRCh38
rs749211518 Chr.8: 142879731-142879731 on Build GRCh38
rs139381647 Chr.8: 142879628-142879628 on Build GRCh38
rs748324186 Chr.8: 142879649-142879649 on Build GRCh38
rs777014524 Chr.8: 142879742-142879742 on Build GRCh38
rs776126642 Chr.8: 142879818-142879818 on Build GRCh38
rs758533779 Chr.8: 142879724-142879724 on Build GRCh38
rs774609209 Chr.8: 142879518-142879518 on Build GRCh38
rs150983560 Chr.8: 142879770-142879770 on Build GRCh38
rs747287245 Chr.8: 142879608-142879608 on Build GRCh38
rs769684367 Chr.8: 142879661-142879661 on Build GRCh38
rs369213890 Chr.8: 142879687-142879687 on Build GRCh38
rs764476381 Chr.8: 142879508-142879508 on Build GRCh38
rs757622420 Chr.8: 142879505-142879505 on Build GRCh38
rs193922538 Chr.8: 142879689-142879689 on Build GRCh38
rs775358429 Chr.8: 142879662-142879662 on Build GRCh38
rs200581194 Chr.8: 142879805-142879805 on Build GRCh38
rs144224988 Chr.8: 142879729-142879729 on Build GRCh38
rs5282 Chr.8: 142879627-142879627 on Build GRCh38

View Full Product Details