Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143295982-143296471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF696
Entrez Gene ID
79943
Gene Name
zinc finger protein 696
Gene Aliases
-
UniGene
Hs.512740
Gene Chromosome Location
Chr.8: 143291389-143299952 on Build GRCh38
Transcripts
NM_030895.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2867957 c.418G>A p.A140T Chr.8: 143296093-143296093 on Build GRCh38
6199093 c.459C>T p.Y153Y Chr.8: 143296134-143296134 on Build GRCh38
5951284 c.694A>G p.T232A Chr.8: 143296369-143296369 on Build GRCh38
233836 c.775C>T p.H259Y Chr.8: 143296450-143296450 on Build GRCh38
4744745 c.477G>A p.G159G Chr.8: 143296152-143296152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561321739 Chr.8: 143296057-143296057 on Build GRCh38
rs767531884 Chr.8: 143296148-143296148 on Build GRCh38
rs767718898 Chr.8: 143296407-143296407 on Build GRCh38
rs569378410 Chr.8: 143296230-143296230 on Build GRCh38
rs11991521 Chr.8: 143296155-143296155 on Build GRCh38
rs370322824 Chr.8: 143296139-143296139 on Build GRCh38
rs933909096 Chr.8: 143296085-143296085 on Build GRCh38
rs754654180 Chr.8: 143296167-143296167 on Build GRCh38
rs762264249 Chr.8: 143296399-143296399 on Build GRCh38
rs763972581 Chr.8: 143296015-143296015 on Build GRCh38
rs746139536 Chr.8: 143296207-143296207 on Build GRCh38
rs776187507 Chr.8: 143296238-143296238 on Build GRCh38
rs367990848 Chr.8: 143296253-143296253 on Build GRCh38
rs925655216 Chr.8: 143296266-143296266 on Build GRCh38
rs749931528 Chr.8: 143296102-143296102 on Build GRCh38
rs773246191 Chr.8: 143296150-143296150 on Build GRCh38
rs770952230 Chr.8: 143296137-143296137 on Build GRCh38
rs776044783 Chr.8: 143296311-143296311 on Build GRCh38
rs773272149 Chr.8: 143296076-143296076 on Build GRCh38
rs1039379194 Chr.8: 143296117-143296117 on Build GRCh38
rs772143902 Chr.8: 143296068-143296068 on Build GRCh38
rs765080475 Chr.8: 143296013-143296013 on Build GRCh38
rs781179297 Chr.8: 143296264-143296264 on Build GRCh38
rs966485578 Chr.8: 143296040-143296040 on Build GRCh38
rs755310310 Chr.8: 143296119-143296119 on Build GRCh38
rs779308003 Chr.8: 143296060-143296060 on Build GRCh38
rs781332118 Chr.8: 143296206-143296206 on Build GRCh38
rs771947080 Chr.8: 143296223-143296223 on Build GRCh38
rs750486605 Chr.8: 143296267-143296267 on Build GRCh38
rs542019617 Chr.8: 143296431-143296431 on Build GRCh38
rs759156535 Chr.8: 143296242-143296242 on Build GRCh38
rs773436307 Chr.8: 143296011-143296011 on Build GRCh38
rs748665746 Chr.8: 143296129-143296129 on Build GRCh38
rs199703934 Chr.8: 143296331-143296331 on Build GRCh38
rs757381438 Chr.8: 143296113-143296113 on Build GRCh38
rs750583425 Chr.8: 143296395-143296396 on Build GRCh38
rs750848818 Chr.8: 143296337-143296337 on Build GRCh38
rs748629891 Chr.8: 143296062-143296062 on Build GRCh38
rs558259005 Chr.8: 143296299-143296299 on Build GRCh38
rs755873115 Chr.8: 143296104-143296104 on Build GRCh38
rs758885660 Chr.8: 143296130-143296130 on Build GRCh38
rs372249835 Chr.8: 143296280-143296280 on Build GRCh38
rs780442451 Chr.8: 143296414-143296415 on Build GRCh38
rs576760839 Chr.8: 143296314-143296314 on Build GRCh38
rs771996263 Chr.8: 143296291-143296291 on Build GRCh38
rs778614333 Chr.8: 143296218-143296218 on Build GRCh38
rs552952180 Chr.8: 143296374-143296374 on Build GRCh38
rs753437636 Chr.8: 143296160-143296160 on Build GRCh38
rs201673017 Chr.8: 143296156-143296156 on Build GRCh38
rs201561096 Chr.8: 143296330-143296330 on Build GRCh38
rs779270066 Chr.8: 143296123-143296123 on Build GRCh38
rs556036385 Chr.8: 143296362-143296362 on Build GRCh38
rs992766377 Chr.8: 143296211-143296211 on Build GRCh38
rs941008100 Chr.8: 143296302-143296302 on Build GRCh38
rs757336518 Chr.8: 143296204-143296204 on Build GRCh38
rs762681691 Chr.8: 143296089-143296089 on Build GRCh38
rs201628392 Chr.8: 143296053-143296053 on Build GRCh38
rs781210571 Chr.8: 143296371-143296371 on Build GRCh38
rs1038100234 Chr.8: 143296126-143296126 on Build GRCh38
rs745876743 Chr.8: 143296436-143296436 on Build GRCh38
rs772090698 Chr.8: 143296009-143296009 on Build GRCh38
rs893111554 Chr.8: 143296224-143296224 on Build GRCh38
rs1002578785 Chr.8: 143296202-143296202 on Build GRCh38
rs904284660 Chr.8: 143296101-143296101 on Build GRCh38
rs776694704 Chr.8: 143296007-143296007 on Build GRCh38
rs142905790 Chr.8: 143296184-143296184 on Build GRCh38
rs775392529 Chr.8: 143296086-143296086 on Build GRCh38
rs752130687 Chr.8: 143296262-143296262 on Build GRCh38
rs146124944 Chr.8: 143296233-143296233 on Build GRCh38
rs200788851 Chr.8: 143296335-143296335 on Build GRCh38
rs762590919 Chr.8: 143296319-143296319 on Build GRCh38
rs372712936 Chr.8: 143296386-143296386 on Build GRCh38
rs765305546 Chr.8: 143296295-143296295 on Build GRCh38
rs756234415 Chr.8: 143296348-143296348 on Build GRCh38
rs1042856605 Chr.8: 143296066-143296066 on Build GRCh38
rs779580375 Chr.8: 143296441-143296441 on Build GRCh38
rs760815415 Chr.8: 143296012-143296012 on Build GRCh38
rs748989959 Chr.8: 143296388-143296388 on Build GRCh38
rs765854878 Chr.8: 143296159-143296159 on Build GRCh38
rs780616301 Chr.8: 143296052-143296052 on Build GRCh38
rs754177164 Chr.8: 143296350-143296350 on Build GRCh38
rs773957383 Chr.8: 143296393-143296393 on Build GRCh38
rs61729401 Chr.8: 143296360-143296360 on Build GRCh38
rs201961006 Chr.8: 143296338-143296338 on Build GRCh38
rs778280872 Chr.8: 143296193-143296193 on Build GRCh38
rs756024838 Chr.8: 143296438-143296438 on Build GRCh38
rs866346211 Chr.8: 143296382-143296382 on Build GRCh38
rs937593961 Chr.8: 143296413-143296413 on Build GRCh38
rs767194477 Chr.8: 143296094-143296094 on Build GRCh38
rs374725649 Chr.8: 143296213-143296213 on Build GRCh38
rs531866086 Chr.8: 143296080-143296080 on Build GRCh38
rs753479290 Chr.8: 143296118-143296118 on Build GRCh38
rs369129290 Chr.8: 143296442-143296442 on Build GRCh38
rs376939193 Chr.8: 143296059-143296059 on Build GRCh38
rs777876903 Chr.8: 143296131-143296131 on Build GRCh38
rs768271527 Chr.8: 143296392-143296392 on Build GRCh38
rs749462125 Chr.8: 143296286-143296286 on Build GRCh38
rs780786184 Chr.8: 143296034-143296034 on Build GRCh38
rs775129940 Chr.8: 143296317-143296317 on Build GRCh38
rs747459332 Chr.8: 143296197-143296197 on Build GRCh38
rs536397793 Chr.8: 143296190-143296190 on Build GRCh38
rs887307137 Chr.8: 143296154-143296154 on Build GRCh38
rs998432953 Chr.8: 143296134-143296134 on Build GRCh38
rs368453184 Chr.8: 143296358-143296358 on Build GRCh38
rs562692809 Chr.8: 143296342-143296342 on Build GRCh38
rs755290412 Chr.8: 143296429-143296429 on Build GRCh38
rs774068714 Chr.8: 143296448-143296448 on Build GRCh38
rs199744108 Chr.8: 143296143-143296143 on Build GRCh38
rs753942655 Chr.8: 143296424-143296424 on Build GRCh38
rs1034765279 Chr.8: 143296221-143296221 on Build GRCh38
rs770879602 Chr.8: 143296309-143296309 on Build GRCh38
rs1040794281 Chr.8: 143296208-143296208 on Build GRCh38
rs530763271 Chr.8: 143296444-143296444 on Build GRCh38
rs543928353 Chr.8: 143296449-143296449 on Build GRCh38
rs755815688 Chr.8: 143296045-143296045 on Build GRCh38
rs766621028 Chr.8: 143296415-143296415 on Build GRCh38
rs755233259 Chr.8: 143296353-143296353 on Build GRCh38
rs746996370 Chr.8: 143296304-143296304 on Build GRCh38
rs2235115 Chr.8: 143296014-143296014 on Build GRCh38
rs747057957 Chr.8: 143296133-143296133 on Build GRCh38
rs764378449 Chr.8: 143296260-143296260 on Build GRCh38
rs750168288 Chr.8: 143296039-143296039 on Build GRCh38
rs1045938933 Chr.8: 143296387-143296387 on Build GRCh38
rs773293902 Chr.8: 143296227-143296227 on Build GRCh38
rs780213336 Chr.8: 143296349-143296349 on Build GRCh38
rs61729412 Chr.8: 143296107-143296107 on Build GRCh38
rs116460889 Chr.8: 143296146-143296146 on Build GRCh38
rs769135621 Chr.8: 143296316-143296316 on Build GRCh38
rs777560842 Chr.8: 143296118-143296118 on Build GRCh38
rs1020394009 Chr.8: 143296245-143296245 on Build GRCh38
rs373620784 Chr.8: 143296030-143296030 on Build GRCh38
rs1035058880 Chr.8: 143296203-143296203 on Build GRCh38
rs745728675 Chr.8: 143296373-143296373 on Build GRCh38
rs780846059 Chr.8: 143296435-143296435 on Build GRCh38
rs945759627 Chr.8: 143296064-143296064 on Build GRCh38
rs768103302 Chr.8: 143296333-143296333 on Build GRCh38
rs757773639 Chr.8: 143296263-143296263 on Build GRCh38
rs761842581 Chr.8: 143296092-143296092 on Build GRCh38
rs758940971 Chr.8: 143296191-143296191 on Build GRCh38
rs756235962 Chr.8: 143296402-143296402 on Build GRCh38
rs139921932 Chr.8: 143296259-143296259 on Build GRCh38
rs969504442 Chr.8: 143296194-143296194 on Build GRCh38
rs61729428 Chr.8: 143296254-143296254 on Build GRCh38
rs907330096 Chr.8: 143296409-143296409 on Build GRCh38
rs752812203 Chr.8: 143296432-143296432 on Build GRCh38
rs200159338 Chr.8: 143296325-143296325 on Build GRCh38
rs757086398 Chr.8: 143296434-143296434 on Build GRCh38
rs375958494 Chr.8: 143296408-143296408 on Build GRCh38
rs756108453 Chr.8: 143296270-143296270 on Build GRCh38
rs913106655 Chr.8: 143296044-143296044 on Build GRCh38
rs747285069 Chr.8: 143296077-143296077 on Build GRCh38
rs751468050 Chr.8: 143296023-143296023 on Build GRCh38
rs574682133 Chr.8: 143296410-143296410 on Build GRCh38
rs554762611 Chr.8: 143296219-143296219 on Build GRCh38
rs774163214 Chr.8: 143296090-143296090 on Build GRCh38

View Full Product Details