Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143651999-143652544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF623
Entrez Gene ID
9831
Gene Name
zinc finger protein 623
Gene Aliases
-
UniGene
Hs.43133
Gene Chromosome Location
Chr.8: 143636013-143653730 on Build GRCh38
Transcripts
NM_001082480.2 NM_001261843.1 NM_014789.3 XM_006716708.3

SNP Detail

SNP ID Chromosome Location
rs188820479 Chr.8: 143652491-143652491 on Build GRCh38
rs955708321 Chr.8: 143652370-143652370 on Build GRCh38
rs948763391 Chr.8: 143652404-143652404 on Build GRCh38
rs190073679 Chr.8: 143652409-143652409 on Build GRCh38
rs551166152 Chr.8: 143652324-143652324 on Build GRCh38
rs145833382 Chr.8: 143652349-143652349 on Build GRCh38
rs967992328 Chr.8: 143652283-143652283 on Build GRCh38
rs920147608 Chr.8: 143652051-143652051 on Build GRCh38
rs896296879 Chr.8: 143652238-143652238 on Build GRCh38
rs561686899 Chr.8: 143652221-143652221 on Build GRCh38
rs139206799 Chr.8: 143652450-143652451 on Build GRCh38
rs1057189918 Chr.8: 143652084-143652084 on Build GRCh38
rs938747979 Chr.8: 143652083-143652083 on Build GRCh38
rs922736644 Chr.8: 143652151-143652151 on Build GRCh38
rs748807740 Chr.8: 143652440-143652440 on Build GRCh38
rs1035231882 Chr.8: 143652062-143652062 on Build GRCh38
rs61473404 Chr.8: 143652213-143652213 on Build GRCh38
rs904875821 Chr.8: 143652221-143652222 on Build GRCh38
rs199676905 Chr.8: 143652459-143652459 on Build GRCh38
rs761850441 Chr.8: 143652230-143652230 on Build GRCh38
rs566292834 Chr.8: 143652345-143652345 on Build GRCh38
rs138712751 Chr.8: 143652371-143652371 on Build GRCh38
rs894468692 Chr.8: 143652157-143652157 on Build GRCh38
rs975528849 Chr.8: 143652150-143652150 on Build GRCh38
rs942747291 Chr.8: 143652132-143652132 on Build GRCh38
rs73369357 Chr.8: 143652036-143652036 on Build GRCh38
rs937721979 Chr.8: 143652497-143652497 on Build GRCh38
rs570859347 Chr.8: 143652273-143652273 on Build GRCh38
rs764011278 Chr.8: 143652193-143652193 on Build GRCh38
rs754758840 Chr.8: 143652303-143652303 on Build GRCh38
rs575880077 Chr.8: 143652039-143652039 on Build GRCh38
rs1056230654 Chr.8: 143652234-143652234 on Build GRCh38
rs1000714527 Chr.8: 143652186-143652186 on Build GRCh38
rs766275788 Chr.8: 143652255-143652255 on Build GRCh38
rs181813544 Chr.8: 143652469-143652469 on Build GRCh38
rs903741419 Chr.8: 143652175-143652175 on Build GRCh38
rs964290662 Chr.8: 143652038-143652038 on Build GRCh38
rs528906965 Chr.8: 143652225-143652225 on Build GRCh38
rs779558508 Chr.8: 143652501-143652501 on Build GRCh38
rs1047637689 Chr.8: 143652065-143652065 on Build GRCh38
rs200620650 Chr.8: 143652466-143652466 on Build GRCh38
rs370123860 Chr.8: 143652496-143652496 on Build GRCh38
rs915975871 Chr.8: 143652377-143652377 on Build GRCh38
rs562678913 Chr.8: 143652307-143652307 on Build GRCh38
rs533255860 Chr.8: 143652314-143652314 on Build GRCh38
rs926020947 Chr.8: 143652130-143652130 on Build GRCh38
rs184039859 Chr.8: 143652477-143652477 on Build GRCh38
rs879469416 Chr.8: 143652507-143652507 on Build GRCh38
rs760961814 Chr.8: 143652164-143652164 on Build GRCh38
rs141981344 Chr.8: 143652159-143652159 on Build GRCh38
rs908632573 Chr.8: 143652078-143652078 on Build GRCh38
rs887381991 Chr.8: 143652168-143652168 on Build GRCh38
rs1030789000 Chr.8: 143652195-143652195 on Build GRCh38
rs546466240 Chr.8: 143652063-143652063 on Build GRCh38
rs983012098 Chr.8: 143652342-143652342 on Build GRCh38
rs938779074 Chr.8: 143652367-143652367 on Build GRCh38
rs772457993 Chr.8: 143652121-143652121 on Build GRCh38
rs978799116 Chr.8: 143652097-143652097 on Build GRCh38
rs777097040 Chr.8: 143652422-143652422 on Build GRCh38
rs200845831 Chr.8: 143652081-143652090 on Build GRCh38
rs564622348 Chr.8: 143652082-143652082 on Build GRCh38

View Full Product Details