Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33917270-33917773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2R2
Entrez Gene ID
54926
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
CDC34B, E2-CDC34B, UBC3B
UniGene
Hs.643648
Gene Chromosome Location
Chr.9: 33817164-33920403 on Build GRCh38
Transcripts
NM_017811.3 XM_005251496.2 XM_011517949.1 XM_017014860.1

SNP Detail

SNP ID Chromosome Location
rs1041100358 Chr.9: 33917417-33917418 on Build GRCh38
rs78156020 Chr.9: 33917716-33917717 on Build GRCh38
rs921364912 Chr.9: 33917692-33917695 on Build GRCh38
rs878858580 Chr.9: 33917703-33917703 on Build GRCh38
rs947386770 Chr.9: 33917641-33917641 on Build GRCh38
rs964557168 Chr.9: 33917516-33917516 on Build GRCh38
rs763991401 Chr.9: 33917488-33917488 on Build GRCh38
rs141897963 Chr.9: 33917351-33917352 on Build GRCh38
rs769447791 Chr.9: 33917589-33917589 on Build GRCh38
rs113151747 Chr.9: 33917352-33917352 on Build GRCh38
rs566584796 Chr.9: 33917696-33917696 on Build GRCh38
rs939988005 Chr.9: 33917715-33917718 on Build GRCh38
rs370898349 Chr.9: 33917426-33917426 on Build GRCh38
rs538594040 Chr.9: 33917728-33917728 on Build GRCh38
rs574969860 Chr.9: 33917318-33917318 on Build GRCh38
rs1040266366 Chr.9: 33917304-33917304 on Build GRCh38
rs546663437 Chr.9: 33917671-33917671 on Build GRCh38
rs192377967 Chr.9: 33917511-33917511 on Build GRCh38
rs138622833 Chr.9: 33917641-33917643 on Build GRCh38
rs1053143661 Chr.9: 33917357-33917357 on Build GRCh38
rs943206337 Chr.9: 33917298-33917298 on Build GRCh38
rs4817 Chr.9: 33917500-33917500 on Build GRCh38
rs989174576 Chr.9: 33917526-33917526 on Build GRCh38
rs1011619809 Chr.9: 33917369-33917369 on Build GRCh38
rs923175408 Chr.9: 33917563-33917563 on Build GRCh38
rs544936741 Chr.9: 33917398-33917398 on Build GRCh38
rs3184789 Chr.9: 33917407-33917407 on Build GRCh38
rs961637529 Chr.9: 33917438-33917438 on Build GRCh38
rs899891967 Chr.9: 33917420-33917420 on Build GRCh38
rs113888596 Chr.9: 33917355-33917355 on Build GRCh38
rs893220800 Chr.9: 33917364-33917364 on Build GRCh38
rs924763442 Chr.9: 33917382-33917382 on Build GRCh38
rs560428091 Chr.9: 33917340-33917340 on Build GRCh38
rs762212106 Chr.9: 33917700-33917700 on Build GRCh38
rs768337761 Chr.9: 33917373-33917373 on Build GRCh38
rs976349049 Chr.9: 33917555-33917555 on Build GRCh38
rs1044463440 Chr.9: 33917659-33917659 on Build GRCh38
rs969019781 Chr.9: 33917562-33917562 on Build GRCh38
rs1033431567 Chr.9: 33917501-33917501 on Build GRCh38
rs1004519363 Chr.9: 33917475-33917475 on Build GRCh38
rs1044452 Chr.9: 33917306-33917306 on Build GRCh38
rs3832626 Chr.9: 33917358-33917359 on Build GRCh38
rs1015469181 Chr.9: 33917479-33917482 on Build GRCh38
rs550633223 Chr.9: 33917504-33917504 on Build GRCh38
rs1057268307 Chr.9: 33917750-33917750 on Build GRCh38
rs59827470 Chr.9: 33917717-33917718 on Build GRCh38
rs1022365765 Chr.9: 33917549-33917549 on Build GRCh38
rs564808746 Chr.9: 33917493-33917493 on Build GRCh38
rs909872659 Chr.9: 33917714-33917718 on Build GRCh38
rs914540822 Chr.9: 33917620-33917620 on Build GRCh38
rs944216911 Chr.9: 33917646-33917646 on Build GRCh38
rs3824461 Chr.9: 33917717-33917717 on Build GRCh38
rs905968488 Chr.9: 33917733-33917733 on Build GRCh38
rs369140439 Chr.9: 33917320-33917320 on Build GRCh38
rs747854717 Chr.9: 33917487-33917490 on Build GRCh38
rs150855520 Chr.9: 33917322-33917322 on Build GRCh38
rs529892435 Chr.9: 33917601-33917601 on Build GRCh38
rs1036959164 Chr.9: 33917724-33917724 on Build GRCh38
rs1048416701 Chr.9: 33917703-33917706 on Build GRCh38
rs971697688 Chr.9: 33917463-33917463 on Build GRCh38
rs1003032496 Chr.9: 33917740-33917740 on Build GRCh38
rs959087894 Chr.9: 33917523-33917523 on Build GRCh38
rs934635394 Chr.9: 33917343-33917343 on Build GRCh38
rs576785644 Chr.9: 33917341-33917341 on Build GRCh38
rs924814164 Chr.9: 33917598-33917599 on Build GRCh38
rs768526470 Chr.9: 33917702-33917703 on Build GRCh38
rs11542250 Chr.9: 33917703-33917703 on Build GRCh38
rs1053025214 Chr.9: 33917600-33917600 on Build GRCh38
rs1035887471 Chr.9: 33917430-33917430 on Build GRCh38
rs976847124 Chr.9: 33917669-33917669 on Build GRCh38
rs959035413 Chr.9: 33917315-33917315 on Build GRCh38

View Full Product Details