Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34329415-34329907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF24
Entrez Gene ID
347240
Gene Name
kinesin family member 24
Gene Aliases
C9orf48, bA571F15.4
UniGene
Hs.710246
Gene Chromosome Location
Chr.9: 34252380-34332972 on Build GRCh38
Transcripts
XM_011517863.2 NM_001161.4 NM_001244390.1 NM_147172.2 NM_147173.2

Gene Symbol
NUDT2
Entrez Gene ID
318
Gene Name
nudix hydrolase 2
Gene Aliases
APAH1
UniGene
Hs.493767
Gene Chromosome Location
Chr.9: 34329506-34343713 on Build GRCh38
Transcripts
XM_011517863.2 NM_001161.4 NM_001244390.1 NM_147172.2 NM_147173.2

SNP Detail

SNP ID Chromosome Location
rs1002152947 Chr.9: 34329797-34329797 on Build GRCh38
rs117117451 Chr.9: 34329795-34329795 on Build GRCh38
rs142195219 Chr.9: 34329687-34329687 on Build GRCh38
rs557134319 Chr.9: 34329467-34329467 on Build GRCh38
rs983906469 Chr.9: 34329620-34329620 on Build GRCh38
rs930815535 Chr.9: 34329668-34329668 on Build GRCh38
rs545326668 Chr.9: 34329648-34329648 on Build GRCh38
rs566892871 Chr.9: 34329823-34329823 on Build GRCh38
rs991796946 Chr.9: 34329484-34329484 on Build GRCh38
rs947859824 Chr.9: 34329471-34329471 on Build GRCh38
rs200833156 Chr.9: 34329858-34329859 on Build GRCh38
rs199994105 Chr.9: 34329464-34329464 on Build GRCh38
rs897964599 Chr.9: 34329814-34329814 on Build GRCh38
rs952444598 Chr.9: 34329612-34329612 on Build GRCh38
rs972203362 Chr.9: 34329647-34329647 on Build GRCh38
rs916289777 Chr.9: 34329630-34329630 on Build GRCh38
rs1006981742 Chr.9: 34329500-34329500 on Build GRCh38
rs919393792 Chr.9: 34329653-34329653 on Build GRCh38
rs1048115403 Chr.9: 34329685-34329685 on Build GRCh38
rs2039344 Chr.9: 34329704-34329704 on Build GRCh38
rs565213484 Chr.9: 34329660-34329660 on Build GRCh38
rs140851024 Chr.9: 34329853-34329854 on Build GRCh38
rs564845556 Chr.9: 34329546-34329546 on Build GRCh38
rs1010956228 Chr.9: 34329588-34329588 on Build GRCh38
rs980761891 Chr.9: 34329600-34329600 on Build GRCh38
rs1009466176 Chr.9: 34329543-34329543 on Build GRCh38
rs1010992612 Chr.9: 34329517-34329517 on Build GRCh38
rs2039345 Chr.9: 34329841-34329841 on Build GRCh38
rs942327343 Chr.9: 34329693-34329693 on Build GRCh38
rs1052788346 Chr.9: 34329724-34329724 on Build GRCh38
rs1022813339 Chr.9: 34329590-34329590 on Build GRCh38
rs543352552 Chr.9: 34329504-34329504 on Build GRCh38
rs1052309595 Chr.9: 34329702-34329702 on Build GRCh38
rs367668823 Chr.9: 34329631-34329631 on Build GRCh38
rs1011172558 Chr.9: 34329763-34329763 on Build GRCh38
rs1050561745 Chr.9: 34329729-34329729 on Build GRCh38
rs929781419 Chr.9: 34329480-34329480 on Build GRCh38
rs1046325581 Chr.9: 34329479-34329479 on Build GRCh38
rs993714776 Chr.9: 34329827-34329827 on Build GRCh38
rs969392194 Chr.9: 34329592-34329592 on Build GRCh38
rs879257538 Chr.9: 34329825-34329825 on Build GRCh38
rs998061491 Chr.9: 34329713-34329713 on Build GRCh38
rs777287325 Chr.9: 34329483-34329483 on Build GRCh38
rs561230896 Chr.9: 34329709-34329709 on Build GRCh38
rs530183871 Chr.9: 34329770-34329770 on Build GRCh38
rs573639422 Chr.9: 34329491-34329491 on Build GRCh38
rs1023420896 Chr.9: 34329865-34329865 on Build GRCh38
rs956747042 Chr.9: 34329569-34329569 on Build GRCh38
rs1030877097 Chr.9: 34329555-34329555 on Build GRCh38
rs1005619720 Chr.9: 34329859-34329859 on Build GRCh38
rs16924260 Chr.9: 34329607-34329607 on Build GRCh38
rs746448261 Chr.9: 34329577-34329579 on Build GRCh38
rs918323543 Chr.9: 34329461-34329461 on Build GRCh38
rs561363922 Chr.9: 34329873-34329873 on Build GRCh38
rs770069948 Chr.9: 34329525-34329525 on Build GRCh38
rs982349072 Chr.9: 34329651-34329651 on Build GRCh38
rs931057834 Chr.9: 34329460-34329460 on Build GRCh38
rs962266720 Chr.9: 34329564-34329564 on Build GRCh38
rs368488735 Chr.9: 34329634-34329634 on Build GRCh38
rs750958853 Chr.9: 34329563-34329563 on Build GRCh38
rs1019063750 Chr.9: 34329802-34329803 on Build GRCh38
rs866201744 Chr.9: 34329774-34329774 on Build GRCh38
rs767843487 Chr.9: 34329749-34329749 on Build GRCh38
rs185212525 Chr.9: 34329847-34329847 on Build GRCh38

View Full Product Details