Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34648026-34648509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALT
Entrez Gene ID
2592
Gene Name
galactose-1-phosphate uridylyltransferase
Gene Aliases
-
UniGene
Hs.522090
Gene Chromosome Location
Chr.9: 34646589-34650598 on Build GRCh38
Transcripts
NM_000155.3 NM_001258332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625705 c.565-1G>A p.? Chr.9: 34648333-34648333 on Build GRCh38
1108504 c.602G>A p.R201H Chr.9: 34648371-34648371 on Build GRCh38
6901095 c.546C>T p.N182N Chr.9: 34648153-34648153 on Build GRCh38
5160332 c.566T>C p.V189A Chr.9: 34648335-34648335 on Build GRCh38
6669198 c.538T>G p.C180G Chr.9: 34648145-34648145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111033743 Chr.9: 34648388-34648388 on Build GRCh38
rs398123181 Chr.9: 34648114-34648114 on Build GRCh38
rs111033722 Chr.9: 34648161-34648161 on Build GRCh38
rs111033744 Chr.9: 34648395-34648395 on Build GRCh38
rs111033712 Chr.9: 34648104-34648104 on Build GRCh38
rs552820931 Chr.9: 34648283-34648283 on Build GRCh38
rs1057113544 Chr.9: 34648389-34648389 on Build GRCh38
rs964746854 Chr.9: 34648136-34648136 on Build GRCh38
rs111033727 Chr.9: 34648352-34648352 on Build GRCh38
rs111033730 Chr.9: 34648364-34648364 on Build GRCh38
rs111033728 Chr.9: 34648353-34648353 on Build GRCh38
rs753958275 Chr.9: 34648423-34648423 on Build GRCh38
rs111033828 Chr.9: 34648148-34648148 on Build GRCh38
rs111033715 Chr.9: 34648119-34648119 on Build GRCh38
rs111033846 Chr.9: 34648449-34648449 on Build GRCh38
rs866391530 Chr.9: 34648155-34648155 on Build GRCh38
rs747870898 Chr.9: 34648166-34648166 on Build GRCh38
rs111033753 Chr.9: 34648456-34648456 on Build GRCh38
rs753832879 Chr.9: 34648389-34648390 on Build GRCh38
rs111033737 Chr.9: 34648379-34648379 on Build GRCh38
rs771840709 Chr.9: 34648327-34648327 on Build GRCh38
rs770786546 Chr.9: 34648258-34648258 on Build GRCh38
rs749283916 Chr.9: 34648299-34648299 on Build GRCh38
rs12000481 Chr.9: 34648098-34648098 on Build GRCh38
rs937733171 Chr.9: 34648277-34648277 on Build GRCh38
rs886042097 Chr.9: 34648165-34648165 on Build GRCh38
rs111033830 Chr.9: 34648343-34648343 on Build GRCh38
rs750603435 Chr.9: 34648288-34648288 on Build GRCh38
rs553769637 Chr.9: 34648437-34648437 on Build GRCh38
rs777577533 Chr.9: 34648447-34648447 on Build GRCh38
rs745373322 Chr.9: 34648473-34648473 on Build GRCh38
rs111033844 Chr.9: 34648146-34648146 on Build GRCh38
rs111033717 Chr.9: 34648159-34648159 on Build GRCh38
rs367543259 Chr.9: 34648149-34648149 on Build GRCh38
rs117998880 Chr.9: 34648465-34648465 on Build GRCh38
rs780156428 Chr.9: 34648121-34648121 on Build GRCh38
rs754930588 Chr.9: 34648306-34648306 on Build GRCh38
rs569393869 Chr.9: 34648326-34648326 on Build GRCh38
rs193922250 Chr.9: 34648457-34648457 on Build GRCh38
rs111033747 Chr.9: 34648427-34648427 on Build GRCh38
rs111033729 Chr.9: 34648363-34648363 on Build GRCh38
rs367543260 Chr.9: 34648399-34648399 on Build GRCh38
rs111033742 Chr.9: 34648421-34648421 on Build GRCh38
rs763403230 Chr.9: 34648194-34648194 on Build GRCh38
rs781377278 Chr.9: 34648112-34648112 on Build GRCh38
rs398123184 Chr.9: 34648337-34648337 on Build GRCh38
rs538993858 Chr.9: 34648268-34648268 on Build GRCh38
rs111033833 Chr.9: 34648404-34648404 on Build GRCh38
rs111033741 Chr.9: 34648419-34648419 on Build GRCh38
rs760662473 Chr.9: 34648090-34648091 on Build GRCh38
rs111033748 Chr.9: 34648458-34648458 on Build GRCh38
rs756379314 Chr.9: 34648414-34648414 on Build GRCh38
rs111033732 Chr.9: 34648186-34648186 on Build GRCh38
rs111033750 Chr.9: 34648436-34648436 on Build GRCh38
rs111033734 Chr.9: 34648344-34648344 on Build GRCh38
rs946336792 Chr.9: 34648251-34648251 on Build GRCh38
rs760101824 Chr.9: 34648089-34648089 on Build GRCh38
rs886044409 Chr.9: 34648120-34648120 on Build GRCh38
rs111033735 Chr.9: 34648371-34648371 on Build GRCh38
rs111033751 Chr.9: 34648445-34648445 on Build GRCh38
rs794727838 Chr.9: 34648341-34648341 on Build GRCh38
rs61735984 Chr.9: 34648117-34648117 on Build GRCh38
rs896091797 Chr.9: 34648397-34648397 on Build GRCh38
rs756291248 Chr.9: 34648296-34648296 on Build GRCh38
rs750693939 Chr.9: 34648413-34648413 on Build GRCh38
rs554104080 Chr.9: 34648450-34648450 on Build GRCh38
rs111033726 Chr.9: 34648349-34648349 on Build GRCh38
rs111033714 Chr.9: 34648110-34648110 on Build GRCh38
rs111033746 Chr.9: 34648403-34648403 on Build GRCh38
rs755961073 Chr.9: 34648190-34648190 on Build GRCh38
rs766219666 Chr.9: 34648307-34648309 on Build GRCh38
rs111033718 Chr.9: 34648131-34648131 on Build GRCh38
rs749869608 Chr.9: 34648078-34648078 on Build GRCh38
rs111033736 Chr.9: 34648376-34648376 on Build GRCh38
rs767099109 Chr.9: 34648067-34648067 on Build GRCh38
rs111033725 Chr.9: 34648163-34648163 on Build GRCh38
rs927326631 Chr.9: 34648291-34648291 on Build GRCh38
rs754928397 Chr.9: 34648466-34648467 on Build GRCh38
rs1057516720 Chr.9: 34648396-34648396 on Build GRCh38
rs111033825 Chr.9: 34648427-34648428 on Build GRCh38
rs368787765 Chr.9: 34648351-34648351 on Build GRCh38
rs767859611 Chr.9: 34648408-34648408 on Build GRCh38
rs1006449714 Chr.9: 34648383-34648383 on Build GRCh38
rs1057516883 Chr.9: 34648439-34648439 on Build GRCh38
rs41274867 Chr.9: 34648091-34648091 on Build GRCh38
rs886042066 Chr.9: 34648356-34648356 on Build GRCh38
rs111033752 Chr.9: 34648446-34648446 on Build GRCh38
rs75391579 Chr.9: 34648170-34648170 on Build GRCh38
rs755000603 Chr.9: 34648432-34648432 on Build GRCh38
rs111033739 Chr.9: 34648370-34648370 on Build GRCh38
rs778835377 Chr.9: 34648324-34648324 on Build GRCh38
rs111033720 Chr.9: 34648143-34648143 on Build GRCh38
rs111033733 Chr.9: 34648203-34648203 on Build GRCh38
rs111033826 Chr.9: 34648160-34648160 on Build GRCh38
rs111033731 Chr.9: 34648332-34648332 on Build GRCh38
rs111033719 Chr.9: 34648135-34648136 on Build GRCh38
rs111033706 Chr.9: 34648077-34648077 on Build GRCh38
rs772835481 Chr.9: 34648174-34648174 on Build GRCh38
rs769294519 Chr.9: 34648482-34648482 on Build GRCh38
rs111033716 Chr.9: 34648157-34648157 on Build GRCh38
rs760189807 Chr.9: 34648178-34648178 on Build GRCh38
rs727503947 Chr.9: 34648103-34648103 on Build GRCh38
rs770451714 Chr.9: 34648378-34648378 on Build GRCh38
rs774080916 Chr.9: 34648066-34648066 on Build GRCh38
rs111033721 Chr.9: 34648154-34648154 on Build GRCh38
rs111033738 Chr.9: 34648367-34648367 on Build GRCh38
rs111033740 Chr.9: 34648380-34648380 on Build GRCh38
rs373048062 Chr.9: 34648452-34648452 on Build GRCh38
rs3824553 Chr.9: 34648401-34648401 on Build GRCh38
rs111033711 Chr.9: 34648086-34648086 on Build GRCh38
rs886042062 Chr.9: 34648374-34648374 on Build GRCh38
rs929470918 Chr.9: 34648357-34648357 on Build GRCh38
rs757569550 Chr.9: 34648223-34648223 on Build GRCh38
rs780230448 Chr.9: 34648298-34648298 on Build GRCh38
rs398123182 Chr.9: 34648169-34648169 on Build GRCh38
rs377589017 Chr.9: 34648200-34648200 on Build GRCh38
rs2070075 Chr.9: 34648421-34648421 on Build GRCh38
rs373302496 Chr.9: 34648180-34648180 on Build GRCh38
rs111033723 Chr.9: 34648172-34648172 on Build GRCh38
rs545007743 Chr.9: 34648483-34648483 on Build GRCh38
rs111033839 Chr.9: 34648116-34648116 on Build GRCh38

View Full Product Details