Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36091049-36091566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECK
Entrez Gene ID
8434
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Gene Aliases
ST15
UniGene
Hs.388918
Gene Chromosome Location
Chr.9: 36036905-36124455 on Build GRCh38
Transcripts
NM_001316345.1 NM_021111.2 XM_017015207.1 XM_017015208.1 XM_017015209.1 XM_017015210.1 XM_017015211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6764661 c.958C>A p.Q320K Chr.9: 36091216-36091216 on Build GRCh38
150606 c.1002C>T p.S334S Chr.9: 36091260-36091260 on Build GRCh38
6764662 c.959A>G p.Q320R Chr.9: 36091217-36091217 on Build GRCh38
6007500 c.980A>T p.E327V Chr.9: 36091238-36091238 on Build GRCh38
4799807 c.1001C>G p.S334C Chr.9: 36091259-36091259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994589223 Chr.9: 36091447-36091447 on Build GRCh38
rs944542924 Chr.9: 36091079-36091079 on Build GRCh38
rs923980255 Chr.9: 36091174-36091174 on Build GRCh38
rs746312531 Chr.9: 36091380-36091380 on Build GRCh38
rs999589347 Chr.9: 36091109-36091109 on Build GRCh38
rs761385396 Chr.9: 36091230-36091230 on Build GRCh38
rs755081820 Chr.9: 36091149-36091149 on Build GRCh38
rs936745299 Chr.9: 36091211-36091211 on Build GRCh38
rs773124487 Chr.9: 36091101-36091101 on Build GRCh38
rs541880054 Chr.9: 36091465-36091465 on Build GRCh38
rs1052007926 Chr.9: 36091277-36091277 on Build GRCh38
rs986242559 Chr.9: 36091362-36091362 on Build GRCh38
rs772836458 Chr.9: 36091258-36091258 on Build GRCh38
rs561543275 Chr.9: 36091529-36091529 on Build GRCh38
rs958383044 Chr.9: 36091526-36091526 on Build GRCh38
rs764950900 Chr.9: 36091122-36091122 on Build GRCh38
rs749822134 Chr.9: 36091187-36091187 on Build GRCh38
rs917180014 Chr.9: 36091086-36091086 on Build GRCh38
rs773795118 Chr.9: 36091477-36091477 on Build GRCh38
rs372033961 Chr.9: 36091262-36091262 on Build GRCh38
rs1054299359 Chr.9: 36091130-36091130 on Build GRCh38
rs1053866439 Chr.9: 36091517-36091517 on Build GRCh38
rs759149260 Chr.9: 36091361-36091361 on Build GRCh38
rs778957417 Chr.9: 36091377-36091377 on Build GRCh38
rs755167457 Chr.9: 36091287-36091287 on Build GRCh38
rs369149597 Chr.9: 36091360-36091360 on Build GRCh38
rs1041493482 Chr.9: 36091090-36091090 on Build GRCh38
rs774889274 Chr.9: 36091189-36091189 on Build GRCh38
rs12235235 Chr.9: 36091136-36091136 on Build GRCh38
rs944592204 Chr.9: 36091383-36091383 on Build GRCh38
rs576082158 Chr.9: 36091368-36091368 on Build GRCh38
rs781628813 Chr.9: 36091154-36091154 on Build GRCh38
rs911563575 Chr.9: 36091377-36091377 on Build GRCh38
rs767648280 Chr.9: 36091309-36091309 on Build GRCh38
rs935857735 Chr.9: 36091482-36091482 on Build GRCh38
rs751642879 Chr.9: 36091141-36091141 on Build GRCh38
rs1012334902 Chr.9: 36091541-36091541 on Build GRCh38
rs376008783 Chr.9: 36091252-36091252 on Build GRCh38
rs748369529 Chr.9: 36091175-36091175 on Build GRCh38
rs756366882 Chr.9: 36091328-36091328 on Build GRCh38
rs1012695108 Chr.9: 36091153-36091153 on Build GRCh38
rs924425735 Chr.9: 36091474-36091474 on Build GRCh38
rs1041630190 Chr.9: 36091435-36091435 on Build GRCh38
rs749653933 Chr.9: 36091359-36091359 on Build GRCh38
rs777935241 Chr.9: 36091185-36091185 on Build GRCh38
rs746588440 Chr.9: 36091193-36091193 on Build GRCh38
rs770557269 Chr.9: 36091460-36091460 on Build GRCh38
rs372625092 Chr.9: 36091181-36091181 on Build GRCh38
rs374907660 Chr.9: 36091286-36091286 on Build GRCh38
rs992758612 Chr.9: 36091082-36091082 on Build GRCh38
rs374823318 Chr.9: 36091349-36091349 on Build GRCh38
rs372176214 Chr.9: 36091315-36091315 on Build GRCh38
rs771390084 Chr.9: 36091188-36091188 on Build GRCh38
rs758231132 Chr.9: 36091135-36091135 on Build GRCh38
rs748985090 Chr.9: 36091214-36091214 on Build GRCh38
rs920368789 Chr.9: 36091314-36091314 on Build GRCh38
rs768155414 Chr.9: 36091392-36091392 on Build GRCh38
rs546462273 Chr.9: 36091229-36091229 on Build GRCh38
rs768136800 Chr.9: 36091215-36091215 on Build GRCh38
rs3736884 Chr.9: 36091260-36091260 on Build GRCh38
rs766270335 Chr.9: 36091274-36091274 on Build GRCh38
rs367769417 Chr.9: 36091146-36091146 on Build GRCh38
rs750377303 Chr.9: 36091131-36091131 on Build GRCh38
rs778712519 Chr.9: 36091514-36091519 on Build GRCh38

View Full Product Details