Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113157435-113157933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC31A2
Entrez Gene ID
1318
Gene Name
solute carrier family 31 member 2
Gene Aliases
COPT2, CTR2, hCTR2
UniGene
Hs.24030
Gene Chromosome Location
Chr.9: 113150958-113164142 on Build GRCh38
Transcripts
NM_001860.2 XM_011518216.1 XM_017014267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
270167 c.73delG p.G25fs*32 Chr.9: 113157793-113157793 on Build GRCh38
6582184 c.7-3C>G p.? Chr.9: 113157724-113157724 on Build GRCh38
3170447 c.52T>C p.W18R Chr.9: 113157772-113157772 on Build GRCh38
6182028 c.23C>G p.S8* Chr.9: 113157743-113157743 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897446236 Chr.9: 113157630-113157630 on Build GRCh38
rs993393747 Chr.9: 113157659-113157659 on Build GRCh38
rs368951803 Chr.9: 113157701-113157701 on Build GRCh38
rs951945719 Chr.9: 113157697-113157697 on Build GRCh38
rs928763809 Chr.9: 113157853-113157853 on Build GRCh38
rs976531088 Chr.9: 113157731-113157731 on Build GRCh38
rs377133770 Chr.9: 113157595-113157595 on Build GRCh38
rs781004195 Chr.9: 113157721-113157721 on Build GRCh38
rs4344146 Chr.9: 113157819-113157819 on Build GRCh38
rs898078473 Chr.9: 113157510-113157510 on Build GRCh38
rs971002379 Chr.9: 113157675-113157675 on Build GRCh38
rs370049098 Chr.9: 113157738-113157738 on Build GRCh38
rs934929256 Chr.9: 113157520-113157520 on Build GRCh38
rs890644548 Chr.9: 113157460-113157460 on Build GRCh38
rs199568431 Chr.9: 113157729-113157729 on Build GRCh38
rs966161909 Chr.9: 113157728-113157728 on Build GRCh38
rs770597660 Chr.9: 113157753-113157753 on Build GRCh38
rs540278111 Chr.9: 113157617-113157617 on Build GRCh38
rs1023370045 Chr.9: 113157640-113157640 on Build GRCh38
rs545864751 Chr.9: 113157881-113157881 on Build GRCh38
rs3934969 Chr.9: 113157502-113157502 on Build GRCh38
rs981965551 Chr.9: 113157676-113157676 on Build GRCh38
rs199500515 Chr.9: 113157757-113157757 on Build GRCh38
rs79528407 Chr.9: 113157709-113157709 on Build GRCh38
rs1054688001 Chr.9: 113157544-113157544 on Build GRCh38
rs986192687 Chr.9: 113157482-113157482 on Build GRCh38
rs10122151 Chr.9: 113157605-113157605 on Build GRCh38
rs12379157 Chr.9: 113157718-113157718 on Build GRCh38
rs758680456 Chr.9: 113157685-113157685 on Build GRCh38
rs758010107 Chr.9: 113157629-113157629 on Build GRCh38
rs565476785 Chr.9: 113157874-113157874 on Build GRCh38
rs777444514 Chr.9: 113157690-113157690 on Build GRCh38
rs780386864 Chr.9: 113157858-113157858 on Build GRCh38
rs879510000 Chr.9: 113157876-113157876 on Build GRCh38
rs976142300 Chr.9: 113157513-113157513 on Build GRCh38
rs924572618 Chr.9: 113157514-113157514 on Build GRCh38
rs764897006 Chr.9: 113157746-113157746 on Build GRCh38
rs1011974443 Chr.9: 113157631-113157631 on Build GRCh38
rs1048709686 Chr.9: 113157904-113157904 on Build GRCh38
rs1003698561 Chr.9: 113157614-113157614 on Build GRCh38
rs748845352 Chr.9: 113157752-113157752 on Build GRCh38
rs754882608 Chr.9: 113157716-113157716 on Build GRCh38
rs1056594279 Chr.9: 113157878-113157878 on Build GRCh38
rs993152440 Chr.9: 113157903-113157903 on Build GRCh38
rs1027500764 Chr.9: 113157679-113157679 on Build GRCh38
rs781258711 Chr.9: 113157726-113157726 on Build GRCh38
rs966523187 Chr.9: 113157507-113157507 on Build GRCh38
rs200827546 Chr.9: 113157801-113157801 on Build GRCh38
rs1044620037 Chr.9: 113157831-113157831 on Build GRCh38
rs1044568270 Chr.9: 113157596-113157596 on Build GRCh38
rs995014735 Chr.9: 113157516-113157516 on Build GRCh38
rs752195492 Chr.9: 113157622-113157622 on Build GRCh38
rs377320489 Chr.9: 113157772-113157772 on Build GRCh38
rs914755450 Chr.9: 113157587-113157587 on Build GRCh38

View Full Product Details