Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114592314-114592823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G1
Entrez Gene ID
9550
Gene Name
ATPase H+ transporting V1 subunit G1
Gene Aliases
ATP6G, ATP6G1, ATP6GL, ATP6J, Vma10
UniGene
Hs.388654
Gene Chromosome Location
Chr.9: 114587714-114598872 on Build GRCh38
Transcripts
NM_004888.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1496988 c.116A>C p.E39A Chr.9: 114592585-114592585 on Build GRCh38
1104422 c.160G>T p.E54* Chr.9: 114592629-114592629 on Build GRCh38
5793053 c.133G>C p.E45Q Chr.9: 114592602-114592602 on Build GRCh38
1104423 c.165C>A p.F55L Chr.9: 114592634-114592634 on Build GRCh38
6006280 c.110_112delAAG p.E39delE Chr.9: 114592579-114592581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891203453 Chr.9: 114592427-114592427 on Build GRCh38
rs151190698 Chr.9: 114592424-114592424 on Build GRCh38
rs1032971690 Chr.9: 114592530-114592530 on Build GRCh38
rs765667517 Chr.9: 114592612-114592612 on Build GRCh38
rs543507200 Chr.9: 114592445-114592445 on Build GRCh38
rs1037996267 Chr.9: 114592417-114592417 on Build GRCh38
rs756109590 Chr.9: 114592537-114592537 on Build GRCh38
rs2274595 Chr.9: 114592663-114592663 on Build GRCh38
rs539464851 Chr.9: 114592703-114592703 on Build GRCh38
rs763039774 Chr.9: 114592647-114592647 on Build GRCh38
rs924336580 Chr.9: 114592401-114592401 on Build GRCh38
rs756121855 Chr.9: 114592671-114592671 on Build GRCh38
rs72752535 Chr.9: 114592585-114592585 on Build GRCh38
rs141971687 Chr.9: 114592725-114592725 on Build GRCh38
rs141470687 Chr.9: 114592434-114592434 on Build GRCh38
rs973097717 Chr.9: 114592681-114592681 on Build GRCh38
rs780561945 Chr.9: 114592715-114592715 on Build GRCh38
rs1013595923 Chr.9: 114592626-114592626 on Build GRCh38
rs572739437 Chr.9: 114592359-114592359 on Build GRCh38
rs868346980 Chr.9: 114592572-114592572 on Build GRCh38
rs867712129 Chr.9: 114592651-114592651 on Build GRCh38
rs890458770 Chr.9: 114592458-114592458 on Build GRCh38
rs540664809 Chr.9: 114592402-114592402 on Build GRCh38
rs991751473 Chr.9: 114592338-114592338 on Build GRCh38
rs900436518 Chr.9: 114592422-114592422 on Build GRCh38
rs902487753 Chr.9: 114592441-114592441 on Build GRCh38
rs549413854 Chr.9: 114592486-114592486 on Build GRCh38
rs373215827 Chr.9: 114592513-114592513 on Build GRCh38
rs781107001 Chr.9: 114592517-114592517 on Build GRCh38
rs564466398 Chr.9: 114592473-114592473 on Build GRCh38
rs752533840 Chr.9: 114592664-114592664 on Build GRCh38
rs977173876 Chr.9: 114592377-114592377 on Build GRCh38
rs150139392 Chr.9: 114592611-114592611 on Build GRCh38
rs745966925 Chr.9: 114592660-114592660 on Build GRCh38
rs150952974 Chr.9: 114592580-114592580 on Build GRCh38
rs1008758009 Chr.9: 114592428-114592428 on Build GRCh38
rs371695769 Chr.9: 114592667-114592667 on Build GRCh38
rs748762523 Chr.9: 114592560-114592560 on Build GRCh38
rs766544680 Chr.9: 114592652-114592652 on Build GRCh38
rs990418839 Chr.9: 114592407-114592407 on Build GRCh38
rs113631225 Chr.9: 114592551-114592551 on Build GRCh38
rs758347722 Chr.9: 114592747-114592747 on Build GRCh38
rs547366971 Chr.9: 114592662-114592662 on Build GRCh38
rs777704361 Chr.9: 114592694-114592694 on Build GRCh38
rs751850924 Chr.9: 114592656-114592656 on Build GRCh38
rs1051535026 Chr.9: 114592756-114592756 on Build GRCh38
rs921850064 Chr.9: 114592696-114592696 on Build GRCh38
rs761151940 Chr.9: 114592425-114592425 on Build GRCh38
rs769047044 Chr.9: 114592606-114592606 on Build GRCh38
rs147009891 Chr.9: 114592535-114592535 on Build GRCh38
rs563395005 Chr.9: 114592484-114592484 on Build GRCh38
rs76694082 Chr.9: 114592658-114592658 on Build GRCh38
rs565826777 Chr.9: 114592634-114592634 on Build GRCh38
rs747363699 Chr.9: 114592601-114592601 on Build GRCh38
rs971019315 Chr.9: 114592360-114592360 on Build GRCh38
rs754230587 Chr.9: 114592454-114592454 on Build GRCh38

View Full Product Details