Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120760312-120760843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW2
Entrez Gene ID
26190
Gene Name
F-box and WD repeat domain containing 2
Gene Aliases
FBW2, Fwd2, Md6
UniGene
Hs.494985
Gene Chromosome Location
Chr.9: 120756976-120793471 on Build GRCh38
Transcripts
NM_012164.3 XM_005251910.3 XM_005251913.2 XM_006717049.3 XM_006717051.2 XM_011518518.1 XM_017014615.1 XM_017014616.1 XM_017014617.1

SNP Detail

SNP ID Chromosome Location
rs535046192 Chr.9: 120760355-120760355 on Build GRCh38
rs962008014 Chr.9: 120760419-120760419 on Build GRCh38
rs922975298 Chr.9: 120760446-120760446 on Build GRCh38
rs554962699 Chr.9: 120760475-120760475 on Build GRCh38
rs568523742 Chr.9: 120760577-120760577 on Build GRCh38
rs988254797 Chr.9: 120760661-120760661 on Build GRCh38
rs978645574 Chr.9: 120760613-120760613 on Build GRCh38
rs369320275 Chr.9: 120760553-120760553 on Build GRCh38
rs947166989 Chr.9: 120760773-120760773 on Build GRCh38
rs759324304 Chr.9: 120760691-120760691 on Build GRCh38
rs891323858 Chr.9: 120760488-120760488 on Build GRCh38
rs1023179782 Chr.9: 120760585-120760585 on Build GRCh38
rs996125941 Chr.9: 120760501-120760501 on Build GRCh38
rs773930634 Chr.9: 120760771-120760771 on Build GRCh38
rs899968113 Chr.9: 120760728-120760728 on Build GRCh38
rs569888558 Chr.9: 120760670-120760670 on Build GRCh38
rs954193351 Chr.9: 120760417-120760417 on Build GRCh38
rs1012805359 Chr.9: 120760364-120760364 on Build GRCh38
rs112739848 Chr.9: 120760640-120760640 on Build GRCh38
rs867322094 Chr.9: 120760353-120760353 on Build GRCh38
rs111912967 Chr.9: 120760402-120760402 on Build GRCh38
rs915819919 Chr.9: 120760381-120760381 on Build GRCh38
rs565973088 Chr.9: 120760807-120760807 on Build GRCh38
rs751286582 Chr.9: 120760676-120760676 on Build GRCh38
rs772209768 Chr.9: 120760352-120760352 on Build GRCh38
rs531226077 Chr.9: 120760412-120760412 on Build GRCh38
rs563685237 Chr.9: 120760380-120760380 on Build GRCh38
rs558545552 Chr.9: 120760754-120760754 on Build GRCh38
rs866768879 Chr.9: 120760742-120760742 on Build GRCh38
rs543879418 Chr.9: 120760794-120760794 on Build GRCh38
rs1056569702 Chr.9: 120760575-120760575 on Build GRCh38
rs1030098157 Chr.9: 120760694-120760694 on Build GRCh38
rs1006462831 Chr.9: 120760655-120760656 on Build GRCh38
rs896533659 Chr.9: 120760621-120760622 on Build GRCh38
rs557925335 Chr.9: 120760644-120760644 on Build GRCh38
rs147993269 Chr.9: 120760498-120760498 on Build GRCh38
rs773188385 Chr.9: 120760558-120760558 on Build GRCh38
rs563807554 Chr.9: 120760804-120760804 on Build GRCh38
rs886749715 Chr.9: 120760538-120760538 on Build GRCh38
rs985862889 Chr.9: 120760744-120760747 on Build GRCh38
rs900081097 Chr.9: 120760499-120760499 on Build GRCh38
rs762869308 Chr.9: 120760584-120760584 on Build GRCh38
rs550988546 Chr.9: 120760660-120760660 on Build GRCh38
rs975865578 Chr.9: 120760436-120760436 on Build GRCh38
rs570642837 Chr.9: 120760767-120760767 on Build GRCh38
rs968671696 Chr.9: 120760592-120760592 on Build GRCh38
rs547130009 Chr.9: 120760576-120760576 on Build GRCh38
rs760753626 Chr.9: 120760457-120760457 on Build GRCh38
rs537332691 Chr.9: 120760712-120760712 on Build GRCh38
rs925239087 Chr.9: 120760428-120760428 on Build GRCh38
rs931853942 Chr.9: 120760458-120760458 on Build GRCh38
rs200433020 Chr.9: 120760636-120760637 on Build GRCh38
rs528647574 Chr.9: 120760545-120760545 on Build GRCh38
rs902521669 Chr.9: 120760552-120760552 on Build GRCh38
rs577810927 Chr.9: 120760747-120760747 on Build GRCh38
rs907586340 Chr.9: 120760413-120760413 on Build GRCh38
rs991841415 Chr.9: 120760368-120760368 on Build GRCh38
rs191883544 Chr.9: 120760489-120760489 on Build GRCh38
rs1026617965 Chr.9: 120760505-120760505 on Build GRCh38
rs752326612 Chr.9: 120760797-120760797 on Build GRCh38
rs867733145 Chr.9: 120760483-120760483 on Build GRCh38

View Full Product Details