Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122675422-122675929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1L3
Entrez Gene ID
26735
Gene Name
olfactory receptor family 1 subfamily L member 3
Gene Aliases
OR9-28, OR9-D
UniGene
Hs.626839
Gene Chromosome Location
Chr.9: 122675130-122676104 on Build GRCh38
Transcripts
NM_001005234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3903833 c.431T>C p.L144P Chr.9: 122675560-122675560 on Build GRCh38
3323591 c.698G>A p.G233E Chr.9: 122675827-122675827 on Build GRCh38
109572 c.555G>A p.L185L Chr.9: 122675684-122675684 on Build GRCh38
3323583 c.495G>A p.R165R Chr.9: 122675624-122675624 on Build GRCh38
1104881 c.664C>T p.L222F Chr.9: 122675793-122675793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748456792 Chr.9: 122675746-122675746 on Build GRCh38
rs778260658 Chr.9: 122675737-122675737 on Build GRCh38
rs566876630 Chr.9: 122675829-122675829 on Build GRCh38
rs112274623 Chr.9: 122675593-122675593 on Build GRCh38
rs773493784 Chr.9: 122675754-122675754 on Build GRCh38
rs747640614 Chr.9: 122675462-122675462 on Build GRCh38
rs369112358 Chr.9: 122675702-122675702 on Build GRCh38
rs765338104 Chr.9: 122675588-122675588 on Build GRCh38
rs779741989 Chr.9: 122675502-122675502 on Build GRCh38
rs764831378 Chr.9: 122675844-122675844 on Build GRCh38
rs61480152 Chr.9: 122675541-122675541 on Build GRCh38
rs762051708 Chr.9: 122675640-122675640 on Build GRCh38
rs16912096 Chr.9: 122675446-122675446 on Build GRCh38
rs772440091 Chr.9: 122675790-122675791 on Build GRCh38
rs888676600 Chr.9: 122675644-122675644 on Build GRCh38
rs778092653 Chr.9: 122675898-122675898 on Build GRCh38
rs957874146 Chr.9: 122675536-122675536 on Build GRCh38
rs751319483 Chr.9: 122675605-122675605 on Build GRCh38
rs549128498 Chr.9: 122675652-122675654 on Build GRCh38
rs775893466 Chr.9: 122675478-122675478 on Build GRCh38
rs895765775 Chr.9: 122675730-122675730 on Build GRCh38
rs372859325 Chr.9: 122675736-122675736 on Build GRCh38
rs754602118 Chr.9: 122675514-122675514 on Build GRCh38
rs750460042 Chr.9: 122675591-122675591 on Build GRCh38
rs376814057 Chr.9: 122675610-122675610 on Build GRCh38
rs756825056 Chr.9: 122675839-122675839 on Build GRCh38
rs753536771 Chr.9: 122675713-122675713 on Build GRCh38
rs372271162 Chr.9: 122675542-122675542 on Build GRCh38
rs567688158 Chr.9: 122675656-122675657 on Build GRCh38
rs548653902 Chr.9: 122675584-122675584 on Build GRCh38
rs532843459 Chr.9: 122675873-122675873 on Build GRCh38
rs781555758 Chr.9: 122675743-122675743 on Build GRCh38
rs139225553 Chr.9: 122675629-122675629 on Build GRCh38
rs746677060 Chr.9: 122675449-122675449 on Build GRCh38
rs772803259 Chr.9: 122675792-122675814 on Build GRCh38
rs367886607 Chr.9: 122675492-122675492 on Build GRCh38
rs768317684 Chr.9: 122675457-122675457 on Build GRCh38
rs780210812 Chr.9: 122675457-122675460 on Build GRCh38
rs895954254 Chr.9: 122675698-122675698 on Build GRCh38
rs763196837 Chr.9: 122675882-122675882 on Build GRCh38
rs79304977 Chr.9: 122675783-122675783 on Build GRCh38
rs146579093 Chr.9: 122675845-122675845 on Build GRCh38
rs200356415 Chr.9: 122675551-122675552 on Build GRCh38
rs546730825 Chr.9: 122675494-122675494 on Build GRCh38
rs764316288 Chr.9: 122675488-122675488 on Build GRCh38
rs935482399 Chr.9: 122675596-122675596 on Build GRCh38
rs1013740201 Chr.9: 122675482-122675482 on Build GRCh38
rs771020185 Chr.9: 122675904-122675904 on Build GRCh38
rs141603415 Chr.9: 122675684-122675684 on Build GRCh38
rs774514841 Chr.9: 122675905-122675905 on Build GRCh38
rs148399372 Chr.9: 122675513-122675513 on Build GRCh38
rs867065384 Chr.9: 122675623-122675623 on Build GRCh38
rs76088588 Chr.9: 122675740-122675740 on Build GRCh38
rs368393655 Chr.9: 122675538-122675538 on Build GRCh38
rs750405135 Chr.9: 122675495-122675495 on Build GRCh38
rs760827970 Chr.9: 122675486-122675486 on Build GRCh38
rs777498475 Chr.9: 122675710-122675710 on Build GRCh38
rs757393563 Chr.9: 122675493-122675493 on Build GRCh38
rs772828607 Chr.9: 122675540-122675540 on Build GRCh38
rs199763256 Chr.9: 122675665-122675665 on Build GRCh38
rs776965522 Chr.9: 122675561-122675561 on Build GRCh38
rs780678535 Chr.9: 122675620-122675620 on Build GRCh38
rs770009817 Chr.9: 122675752-122675752 on Build GRCh38
rs780834268 Chr.9: 122675521-122675521 on Build GRCh38
rs754872664 Chr.9: 122675607-122675607 on Build GRCh38
rs759551045 Chr.9: 122675770-122675770 on Build GRCh38
rs748652885 Chr.9: 122675896-122675896 on Build GRCh38
rs372999173 Chr.9: 122675615-122675615 on Build GRCh38
rs16912099 Chr.9: 122675490-122675490 on Build GRCh38
rs773296831 Chr.9: 122675655-122675655 on Build GRCh38
rs576705691 Chr.9: 122675608-122675608 on Build GRCh38
rs964399850 Chr.9: 122675543-122675543 on Build GRCh38
rs117289796 Chr.9: 122675477-122675477 on Build GRCh38
rs142794411 Chr.9: 122675622-122675622 on Build GRCh38
rs761996558 Chr.9: 122675583-122675583 on Build GRCh38
rs746657512 Chr.9: 122675728-122675728 on Build GRCh38
rs866315429 Chr.9: 122675715-122675715 on Build GRCh38
rs777895439 Chr.9: 122675468-122675468 on Build GRCh38
rs368933686 Chr.9: 122675612-122675612 on Build GRCh38
rs776030536 Chr.9: 122675458-122675458 on Build GRCh38
rs758297685 Chr.9: 122675498-122675498 on Build GRCh38
rs374999670 Chr.9: 122675675-122675675 on Build GRCh38
rs774264975 Chr.9: 122675769-122675769 on Build GRCh38
rs766285174 Chr.9: 122675595-122675595 on Build GRCh38
rs752501356 Chr.9: 122675704-122675704 on Build GRCh38
rs976943090 Chr.9: 122675559-122675559 on Build GRCh38
rs781615022 Chr.9: 122675891-122675891 on Build GRCh38
rs780347027 Chr.9: 122675646-122675646 on Build GRCh38
rs760107014 Chr.9: 122675476-122675476 on Build GRCh38
rs147172743 Chr.9: 122675549-122675549 on Build GRCh38
rs749565366 Chr.9: 122675900-122675900 on Build GRCh38
rs747801263 Chr.9: 122675525-122675525 on Build GRCh38
rs78972606 Chr.9: 122675553-122675553 on Build GRCh38
rs145723911 Chr.9: 122675778-122675778 on Build GRCh38
rs751576666 Chr.9: 122675690-122675690 on Build GRCh38
rs144421636 Chr.9: 122675613-122675613 on Build GRCh38
rs1020822873 Chr.9: 122675899-122675899 on Build GRCh38
rs766370067 Chr.9: 122675677-122675677 on Build GRCh38
rs550125655 Chr.9: 122675813-122675813 on Build GRCh38
rs776945389 Chr.9: 122675638-122675638 on Build GRCh38
rs16912101 Chr.9: 122675798-122675798 on Build GRCh38
rs780262120 Chr.9: 122675669-122675669 on Build GRCh38
rs768238452 Chr.9: 122675739-122675739 on Build GRCh38
rs763075537 Chr.9: 122675757-122675757 on Build GRCh38
rs775729997 Chr.9: 122675545-122675545 on Build GRCh38
rs200264223 Chr.9: 122675799-122675799 on Build GRCh38
rs779249829 Chr.9: 122675657-122675657 on Build GRCh38

View Full Product Details