Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14607050-14607556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA2
Entrez Gene ID
2742
Gene Name
glycine receptor alpha 2
Gene Aliases
GLR
UniGene
Hs.2700
Gene Chromosome Location
Chr.X: 14448779-14731812 on Build GRCh38
Transcripts
NM_001118885.1 NM_001118886.1 NM_001171942.1 NM_002063.3 XM_006724487.3 XM_011545495.2 XM_011545496.2 XM_017029427.1 XM_017029428.1 XM_017029429.1

SNP Detail

SNP ID Chromosome Location
rs746619310 Chr.X: 14607280-14607281 on Build GRCh38
rs1036725727 Chr.X: 14607138-14607138 on Build GRCh38
rs386416661 Chr.X: 14607278-14607279 on Build GRCh38
rs112633985 Chr.X: 14607348-14607348 on Build GRCh38
rs763300855 Chr.X: 14607273-14607274 on Build GRCh38
rs372409623 Chr.X: 14607277-14607277 on Build GRCh38
rs759202145 Chr.X: 14607313-14607313 on Build GRCh38
rs758474224 Chr.X: 14607443-14607443 on Build GRCh38
rs779386789 Chr.X: 14607252-14607252 on Build GRCh38
rs752994665 Chr.X: 14607088-14607088 on Build GRCh38
rs778899369 Chr.X: 14607361-14607361 on Build GRCh38
rs765533199 Chr.X: 14607081-14607081 on Build GRCh38
rs769156407 Chr.X: 14607257-14607257 on Build GRCh38
rs767536117 Chr.X: 14607314-14607314 on Build GRCh38
rs774317933 Chr.X: 14607297-14607297 on Build GRCh38
rs193245219 Chr.X: 14607455-14607455 on Build GRCh38
rs372265920 Chr.X: 14607277-14607278 on Build GRCh38
rs750637461 Chr.X: 14607096-14607096 on Build GRCh38
rs386419266 Chr.X: 14607292-14607293 on Build GRCh38
rs372099149 Chr.X: 14607250-14607250 on Build GRCh38
rs113271440 Chr.X: 14607141-14607141 on Build GRCh38
rs746441897 Chr.X: 14607512-14607512 on Build GRCh38
rs773068338 Chr.X: 14607270-14607270 on Build GRCh38
rs770639274 Chr.X: 14607292-14607292 on Build GRCh38
rs748960440 Chr.X: 14607272-14607272 on Build GRCh38
rs1020091905 Chr.X: 14607098-14607098 on Build GRCh38
rs770447310 Chr.X: 14607290-14607291 on Build GRCh38
rs142647740 Chr.X: 14607392-14607392 on Build GRCh38
rs796319692 Chr.X: 14607278-14607279 on Build GRCh38
rs982989224 Chr.X: 14607414-14607414 on Build GRCh38
rs6630684 Chr.X: 14607480-14607480 on Build GRCh38
rs775257374 Chr.X: 14607315-14607315 on Build GRCh38
rs975933581 Chr.X: 14607373-14607373 on Build GRCh38
rs908809012 Chr.X: 14607445-14607445 on Build GRCh38
rs755545025 Chr.X: 14607147-14607147 on Build GRCh38
rs931050981 Chr.X: 14607180-14607180 on Build GRCh38
rs1048192928 Chr.X: 14607448-14607448 on Build GRCh38
rs949068438 Chr.X: 14607121-14607121 on Build GRCh38
rs756970298 Chr.X: 14607220-14607220 on Build GRCh38
rs777940399 Chr.X: 14607278-14607280 on Build GRCh38
rs747150969 Chr.X: 14607342-14607342 on Build GRCh38
rs377763484 Chr.X: 14607277-14607277 on Build GRCh38
rs750370767 Chr.X: 14607404-14607404 on Build GRCh38
rs199920383 Chr.X: 14607104-14607104 on Build GRCh38
rs184557224 Chr.X: 14607485-14607485 on Build GRCh38
rs778811116 Chr.X: 14607278-14607278 on Build GRCh38
rs1006736663 Chr.X: 14607484-14607484 on Build GRCh38

View Full Product Details