Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18240856-18241382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCML2
Entrez Gene ID
10389
Gene Name
sex comb on midleg-like 2 (Drosophila)
Gene Aliases
-
UniGene
Hs.495774
Gene Chromosome Location
Chr.X: 18239313-18355089 on Build GRCh38
Transcripts
NM_006089.2 NR_033717.1 XM_006724458.1 XM_017029219.1 XM_017029220.1 XM_017029221.1 XM_017029222.1

SNP Detail

SNP ID Chromosome Location
rs1037076724 Chr.X: 18241229-18241229 on Build GRCh38
rs745937223 Chr.X: 18241356-18241356 on Build GRCh38
rs141270670 Chr.X: 18240953-18240953 on Build GRCh38
rs746829529 Chr.X: 18241256-18241256 on Build GRCh38
rs749577518 Chr.X: 18241304-18241304 on Build GRCh38
rs760113781 Chr.X: 18241105-18241106 on Build GRCh38
rs779767255 Chr.X: 18241244-18241244 on Build GRCh38
rs959487267 Chr.X: 18241355-18241355 on Build GRCh38
rs1029225973 Chr.X: 18241095-18241095 on Build GRCh38
rs41309701 Chr.X: 18241021-18241021 on Build GRCh38
rs547537233 Chr.X: 18240893-18240893 on Build GRCh38
rs968083586 Chr.X: 18241061-18241061 on Build GRCh38
rs774636835 Chr.X: 18241329-18241329 on Build GRCh38
rs35441337 Chr.X: 18240904-18240905 on Build GRCh38
rs779044208 Chr.X: 18241142-18241142 on Build GRCh38
rs777830526 Chr.X: 18241268-18241268 on Build GRCh38
rs944461299 Chr.X: 18240915-18240915 on Build GRCh38
rs750358127 Chr.X: 18241225-18241225 on Build GRCh38
rs770905698 Chr.X: 18241327-18241327 on Build GRCh38
rs756690275 Chr.X: 18241215-18241215 on Build GRCh38
rs956872257 Chr.X: 18241015-18241015 on Build GRCh38
rs757476375 Chr.X: 18241092-18241095 on Build GRCh38
rs374539361 Chr.X: 18241353-18241353 on Build GRCh38
rs1049101398 Chr.X: 18241126-18241126 on Build GRCh38
rs755995920 Chr.X: 18240983-18240983 on Build GRCh38
rs888730791 Chr.X: 18241149-18241149 on Build GRCh38
rs906578755 Chr.X: 18241286-18241286 on Build GRCh38
rs200260668 Chr.X: 18241357-18241357 on Build GRCh38
rs763875532 Chr.X: 18241226-18241226 on Build GRCh38
rs180990036 Chr.X: 18241040-18241040 on Build GRCh38
rs976374836 Chr.X: 18241066-18241066 on Build GRCh38
rs911638936 Chr.X: 18240894-18240894 on Build GRCh38
rs1019112113 Chr.X: 18241000-18241000 on Build GRCh38
rs1009298156 Chr.X: 18241036-18241036 on Build GRCh38
rs746714168 Chr.X: 18241246-18241246 on Build GRCh38
rs79231433 Chr.X: 18241252-18241252 on Build GRCh38
rs764850013 Chr.X: 18241224-18241224 on Build GRCh38
rs777686432 Chr.X: 18241245-18241246 on Build GRCh38
rs980454734 Chr.X: 18241135-18241135 on Build GRCh38
rs78642840 Chr.X: 18241254-18241254 on Build GRCh38
rs753348424 Chr.X: 18241207-18241207 on Build GRCh38
rs372965508 Chr.X: 18241047-18241047 on Build GRCh38
rs746307395 Chr.X: 18241348-18241348 on Build GRCh38
rs756324398 Chr.X: 18241260-18241260 on Build GRCh38

View Full Product Details