Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21754168-21754672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPX
Entrez Gene ID
23676
Gene Name
small muscle protein, X-linked
Gene Aliases
DFN6, DFNX4
UniGene
Hs.734084
Gene Chromosome Location
Chr.X: 21705972-21758160 on Build GRCh38
Transcripts
NM_014332.2 NR_045617.1

SNP Detail

SNP ID Chromosome Location
rs940052562 Chr.X: 21754372-21754372 on Build GRCh38
rs150242820 Chr.X: 21754381-21754381 on Build GRCh38
rs748590318 Chr.X: 21754240-21754240 on Build GRCh38
rs769334610 Chr.X: 21754219-21754219 on Build GRCh38
rs779036885 Chr.X: 21754200-21754200 on Build GRCh38
rs375612194 Chr.X: 21754279-21754279 on Build GRCh38
rs762689770 Chr.X: 21754305-21754305 on Build GRCh38
rs1013809656 Chr.X: 21754592-21754592 on Build GRCh38
rs966030271 Chr.X: 21754416-21754416 on Build GRCh38
rs1055569199 Chr.X: 21754635-21754635 on Build GRCh38
rs983333031 Chr.X: 21754223-21754223 on Build GRCh38
rs113770939 Chr.X: 21754465-21754465 on Build GRCh38
rs926760512 Chr.X: 21754528-21754528 on Build GRCh38
rs937104865 Chr.X: 21754534-21754534 on Build GRCh38
rs748282018 Chr.X: 21754304-21754305 on Build GRCh38
rs747441182 Chr.X: 21754218-21754218 on Build GRCh38
rs3213453 Chr.X: 21754636-21754636 on Build GRCh38
rs946983012 Chr.X: 21754496-21754496 on Build GRCh38
rs980048059 Chr.X: 21754519-21754519 on Build GRCh38
rs914142249 Chr.X: 21754487-21754487 on Build GRCh38
rs762363215 Chr.X: 21754472-21754472 on Build GRCh38
rs571727006 Chr.X: 21754367-21754367 on Build GRCh38
rs770308935 Chr.X: 21754332-21754332 on Build GRCh38
rs745557355 Chr.X: 21754193-21754193 on Build GRCh38
rs770287561 Chr.X: 21754272-21754272 on Build GRCh38
rs898598566 Chr.X: 21754445-21754445 on Build GRCh38
rs781679413 Chr.X: 21754231-21754231 on Build GRCh38
rs780210941 Chr.X: 21754212-21754212 on Build GRCh38
rs750668242 Chr.X: 21754210-21754210 on Build GRCh38
rs896675044 Chr.X: 21754578-21754578 on Build GRCh38
rs35015961 Chr.X: 21754437-21754438 on Build GRCh38
rs908507026 Chr.X: 21754242-21754242 on Build GRCh38

View Full Product Details