Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 34630242-34630789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM47
Entrez Gene ID
83604
Gene Name
transmembrane protein 47
Gene Aliases
BCMP1, TM4SF10
UniGene
Hs.8769
Gene Chromosome Location
Chr.X: 34627064-34657288 on Build GRCh38
Transcripts
NM_031442.3

SNP Detail

SNP ID Chromosome Location
rs373107619 Chr.X: 34630517-34630517 on Build GRCh38
rs903030211 Chr.X: 34630356-34630356 on Build GRCh38
rs781465346 Chr.X: 34630497-34630497 on Build GRCh38
rs889707220 Chr.X: 34630712-34630712 on Build GRCh38
rs768980591 Chr.X: 34630470-34630470 on Build GRCh38
rs773491398 Chr.X: 34630368-34630368 on Build GRCh38
rs938191452 Chr.X: 34630536-34630536 on Build GRCh38
rs777297767 Chr.X: 34630510-34630510 on Build GRCh38
rs770870666 Chr.X: 34630529-34630529 on Build GRCh38
rs759097079 Chr.X: 34630702-34630705 on Build GRCh38
rs145024985 Chr.X: 34630718-34630718 on Build GRCh38
rs1023601286 Chr.X: 34630653-34630653 on Build GRCh38
rs775061241 Chr.X: 34630400-34630400 on Build GRCh38
rs770388301 Chr.X: 34630334-34630334 on Build GRCh38
rs538844607 Chr.X: 34630709-34630709 on Build GRCh38
rs762226871 Chr.X: 34630523-34630523 on Build GRCh38
rs769538642 Chr.X: 34630315-34630315 on Build GRCh38
rs186215242 Chr.X: 34630322-34630322 on Build GRCh38
rs760878087 Chr.X: 34630469-34630469 on Build GRCh38
rs773875631 Chr.X: 34630362-34630362 on Build GRCh38
rs868197582 Chr.X: 34630339-34630339 on Build GRCh38
rs767266648 Chr.X: 34630714-34630714 on Build GRCh38
rs778329651 Chr.X: 34630532-34630532 on Build GRCh38
rs747973639 Chr.X: 34630309-34630309 on Build GRCh38
rs931969300 Chr.X: 34630592-34630592 on Build GRCh38
rs780786653 Chr.X: 34630299-34630299 on Build GRCh38
rs1006279674 Chr.X: 34630698-34630698 on Build GRCh38
rs267606443 Chr.X: 34630371-34630371 on Build GRCh38
rs762215328 Chr.X: 34630436-34630436 on Build GRCh38
rs746279596 Chr.X: 34630676-34630676 on Build GRCh38
rs748371531 Chr.X: 34630508-34630508 on Build GRCh38
rs755083281 Chr.X: 34630482-34630482 on Build GRCh38
rs749264967 Chr.X: 34630326-34630326 on Build GRCh38
rs755910956 Chr.X: 34630509-34630509 on Build GRCh38
rs760874321 Chr.X: 34630757-34630757 on Build GRCh38
rs750916621 Chr.X: 34630458-34630458 on Build GRCh38
rs755261595 Chr.X: 34630287-34630287 on Build GRCh38
rs375573490 Chr.X: 34630569-34630569 on Build GRCh38
rs775423495 Chr.X: 34630457-34630457 on Build GRCh38
rs1003149977 Chr.X: 34630357-34630357 on Build GRCh38
rs894424082 Chr.X: 34630573-34630573 on Build GRCh38
rs1012243241 Chr.X: 34630642-34630642 on Build GRCh38
rs901874475 Chr.X: 34630579-34630579 on Build GRCh38
rs750863403 Chr.X: 34630682-34630682 on Build GRCh38
rs1057032678 Chr.X: 34630562-34630562 on Build GRCh38
rs751690455 Chr.X: 34630475-34630475 on Build GRCh38
rs765661991 Chr.X: 34630585-34630585 on Build GRCh38
rs767192945 Chr.X: 34630370-34630370 on Build GRCh38

View Full Product Details