Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45162902-45163406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_024689.2 NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs149302589 Chr.X: 45163336-45163336 on Build GRCh38
rs1012178396 Chr.X: 45163211-45163211 on Build GRCh38
rs979819065 Chr.X: 45163263-45163263 on Build GRCh38
rs931140420 Chr.X: 45163075-45163075 on Build GRCh38
rs963447327 Chr.X: 45163254-45163254 on Build GRCh38
rs919852717 Chr.X: 45163360-45163360 on Build GRCh38
rs768110743 Chr.X: 45163362-45163362 on Build GRCh38
rs893360904 Chr.X: 45163210-45163210 on Build GRCh38
rs1025824614 Chr.X: 45163165-45163165 on Build GRCh38
rs946144458 Chr.X: 45162957-45162957 on Build GRCh38
rs368812641 Chr.X: 45163264-45163264 on Build GRCh38
rs952582313 Chr.X: 45163370-45163370 on Build GRCh38
rs972355601 Chr.X: 45163313-45163313 on Build GRCh38
rs1015988318 Chr.X: 45163246-45163246 on Build GRCh38
rs926949527 Chr.X: 45163284-45163284 on Build GRCh38
rs28619177 Chr.X: 45163262-45163262 on Build GRCh38
rs1053308019 Chr.X: 45163181-45163181 on Build GRCh38
rs999102036 Chr.X: 45163155-45163155 on Build GRCh38
rs758072172 Chr.X: 45162952-45162952 on Build GRCh38
rs1040805888 Chr.X: 45163142-45163142 on Build GRCh38
rs970242449 Chr.X: 45163233-45163233 on Build GRCh38
rs927264239 Chr.X: 45162945-45162945 on Build GRCh38
rs146169230 Chr.X: 45162956-45162956 on Build GRCh38
rs992671303 Chr.X: 45163309-45163309 on Build GRCh38
rs779921100 Chr.X: 45162953-45162953 on Build GRCh38
rs901843361 Chr.X: 45163150-45163150 on Build GRCh38
rs775218213 Chr.X: 45163037-45163037 on Build GRCh38
rs1035805320 Chr.X: 45163291-45163291 on Build GRCh38
rs904534290 Chr.X: 45162972-45162972 on Build GRCh38
rs1049549651 Chr.X: 45163094-45163094 on Build GRCh38
rs972645321 Chr.X: 45162979-45162979 on Build GRCh38
rs371833171 Chr.X: 45163102-45163102 on Build GRCh38
rs762443248 Chr.X: 45163219-45163219 on Build GRCh38
rs866226819 Chr.X: 45163359-45163359 on Build GRCh38
rs143570142 Chr.X: 45163375-45163375 on Build GRCh38
rs943448045 Chr.X: 45163132-45163132 on Build GRCh38
rs1043084355 Chr.X: 45162967-45162967 on Build GRCh38
rs1043267588 Chr.X: 45163382-45163382 on Build GRCh38
rs961037138 Chr.X: 45163307-45163307 on Build GRCh38

View Full Product Details