Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47482322-47482827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01560
Entrez Gene ID
203414
Gene Name
long intergenic non-protein coding RNA 1560
Gene Aliases
CXorf24
UniGene
Hs.597617
Gene Chromosome Location
Chr.X: 47482716-47485227 on Build GRCh38
Transcripts
NR_126059.1 NM_001324139.1 NM_001324140.1 NM_001324141.1 NM_001324142.1 NM_001324143.1 NM_001324144.1 NM_001324145.1 NM_001324147.1 NM_001324148.1 NM_001324149.1 NM_001324150.1 NM_001324151.1 NM_001324152.1 NM_001324153.1 NM_007130.3 NM_153380.3 XM_006724550.3 XM_006724555.3 XM_017029810.1 XM_017029811.1 XM_017029812.1 XM_017029814.1 XM_017029815.1

Gene Symbol
ZNF41
Entrez Gene ID
7592
Gene Name
zinc finger protein 41
Gene Aliases
MRX89
UniGene
Hs.496074
Gene Chromosome Location
Chr.X: 47444691-47483671 on Build GRCh38
Transcripts
NR_126059.1 NM_001324139.1 NM_001324140.1 NM_001324141.1 NM_001324142.1 NM_001324143.1 NM_001324144.1 NM_001324145.1 NM_001324147.1 NM_001324148.1 NM_001324149.1 NM_001324150.1 NM_001324151.1 NM_001324152.1 NM_001324153.1 NM_007130.3 NM_153380.3 XM_006724550.3 XM_006724555.3 XM_017029810.1 XM_017029811.1 XM_017029812.1 XM_017029814.1 XM_017029815.1

SNP Detail

SNP ID Chromosome Location
rs1057515905 Chr.X: 47482726-47482726 on Build GRCh38
rs386825111 Chr.X: 47482381-47482383 on Build GRCh38
rs962014618 Chr.X: 47482657-47482657 on Build GRCh38
rs989561890 Chr.X: 47482747-47482747 on Build GRCh38
rs911770372 Chr.X: 47482520-47482520 on Build GRCh38
rs986051569 Chr.X: 47482499-47482499 on Build GRCh38
rs1041010002 Chr.X: 47482783-47482783 on Build GRCh38
rs750623553 Chr.X: 47482710-47482710 on Build GRCh38
rs1022573831 Chr.X: 47482395-47482395 on Build GRCh38
rs761263565 Chr.X: 47482539-47482539 on Build GRCh38
rs200168272 Chr.X: 47482617-47482618 on Build GRCh38
rs35384257 Chr.X: 47482799-47482800 on Build GRCh38
rs796332298 Chr.X: 47482609-47482610 on Build GRCh38
rs906764532 Chr.X: 47482739-47482739 on Build GRCh38
rs926429255 Chr.X: 47482742-47482742 on Build GRCh38
rs1057515904 Chr.X: 47482693-47482693 on Build GRCh38
rs754233603 Chr.X: 47482709-47482709 on Build GRCh38
rs2071780 Chr.X: 47482422-47482422 on Build GRCh38
rs867592831 Chr.X: 47482356-47482356 on Build GRCh38
rs930074243 Chr.X: 47482681-47482681 on Build GRCh38
rs181467155 Chr.X: 47482482-47482482 on Build GRCh38
rs980619364 Chr.X: 47482726-47482726 on Build GRCh38
rs370201848 Chr.X: 47482552-47482552 on Build GRCh38
rs756461751 Chr.X: 47482730-47482730 on Build GRCh38
rs201628942 Chr.X: 47482604-47482605 on Build GRCh38
rs5905609 Chr.X: 47482381-47482381 on Build GRCh38
rs993462852 Chr.X: 47482687-47482687 on Build GRCh38
rs1015957271 Chr.X: 47482637-47482637 on Build GRCh38
rs189121794 Chr.X: 47482434-47482434 on Build GRCh38
rs759437743 Chr.X: 47482550-47482550 on Build GRCh38
rs371358051 Chr.X: 47482610-47482610 on Build GRCh38
rs945375290 Chr.X: 47482768-47482768 on Build GRCh38
rs945686753 Chr.X: 47482461-47482461 on Build GRCh38
rs754568556 Chr.X: 47482609-47482610 on Build GRCh38
rs2071779 Chr.X: 47482486-47482486 on Build GRCh38
rs953238438 Chr.X: 47482479-47482479 on Build GRCh38
rs996972516 Chr.X: 47482514-47482514 on Build GRCh38
rs5905610 Chr.X: 47482386-47482386 on Build GRCh38
rs369917096 Chr.X: 47482616-47482616 on Build GRCh38
rs1005915083 Chr.X: 47482598-47482598 on Build GRCh38
rs200484529 Chr.X: 47482386-47482387 on Build GRCh38
rs965387028 Chr.X: 47482521-47482521 on Build GRCh38
rs922861198 Chr.X: 47482794-47482794 on Build GRCh38
rs910074669 Chr.X: 47482689-47482689 on Build GRCh38
rs958131302 Chr.X: 47482744-47482744 on Build GRCh38
rs764227140 Chr.X: 47482585-47482585 on Build GRCh38
rs976718788 Chr.X: 47482525-47482525 on Build GRCh38
rs372112867 Chr.X: 47482600-47482601 on Build GRCh38
rs1025319944 Chr.X: 47482693-47482693 on Build GRCh38
rs918577163 Chr.X: 47482561-47482561 on Build GRCh38
rs757307096 Chr.X: 47482601-47482601 on Build GRCh38
rs913935594 Chr.X: 47482753-47482753 on Build GRCh38
rs866722869 Chr.X: 47482596-47482596 on Build GRCh38
rs375324077 Chr.X: 47482614-47482614 on Build GRCh38
rs767640889 Chr.X: 47482696-47482696 on Build GRCh38
rs12859544 Chr.X: 47482600-47482600 on Build GRCh38
rs866647762 Chr.X: 47482618-47482618 on Build GRCh38

View Full Product Details