Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47571923-47572427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYN1
Entrez Gene ID
6853
Gene Name
synapsin I
Gene Aliases
SYN1a, SYN1b, SYNI
UniGene
Hs.225936
Gene Chromosome Location
Chr.X: 47571901-47619857 on Build GRCh38
Transcripts
NM_006950.3 NM_133499.2

SNP Detail

SNP ID Chromosome Location
rs113508989 Chr.X: 47572358-47572358 on Build GRCh38
rs945364620 Chr.X: 47571990-47571990 on Build GRCh38
rs903967580 Chr.X: 47572018-47572018 on Build GRCh38
rs1015907093 Chr.X: 47572207-47572207 on Build GRCh38
rs938518393 Chr.X: 47572398-47572398 on Build GRCh38
rs975057665 Chr.X: 47572010-47572010 on Build GRCh38
rs2855136 Chr.X: 47572302-47572302 on Build GRCh38
rs10066 Chr.X: 47572140-47572140 on Build GRCh38
rs35136843 Chr.X: 47572240-47572241 on Build GRCh38
rs756961959 Chr.X: 47572245-47572245 on Build GRCh38
rs185676793 Chr.X: 47572160-47572160 on Build GRCh38
rs1057492789 Chr.X: 47572132-47572132 on Build GRCh38
rs1048181931 Chr.X: 47572061-47572061 on Build GRCh38
rs988418210 Chr.X: 47571960-47571960 on Build GRCh38
rs900632321 Chr.X: 47572107-47572107 on Build GRCh38
rs1030581385 Chr.X: 47572147-47572147 on Build GRCh38
rs1000930549 Chr.X: 47572048-47572048 on Build GRCh38
rs759484232 Chr.X: 47572000-47572000 on Build GRCh38
rs896523752 Chr.X: 47571958-47571958 on Build GRCh38
rs148814895 Chr.X: 47572070-47572070 on Build GRCh38
rs1013601221 Chr.X: 47572242-47572242 on Build GRCh38
rs912472821 Chr.X: 47571982-47571982 on Build GRCh38
rs940203026 Chr.X: 47572101-47572101 on Build GRCh38
rs376126952 Chr.X: 47572245-47572245 on Build GRCh38
rs764637755 Chr.X: 47572221-47572221 on Build GRCh38
rs983098341 Chr.X: 47572207-47572208 on Build GRCh38
rs767782270 Chr.X: 47572060-47572060 on Build GRCh38
rs927157839 Chr.X: 47572336-47572336 on Build GRCh38
rs1057045746 Chr.X: 47571957-47571957 on Build GRCh38
rs778746358 Chr.X: 47572254-47572254 on Build GRCh38
rs751893264 Chr.X: 47572243-47572243 on Build GRCh38
rs765367783 Chr.X: 47572052-47572052 on Build GRCh38
rs920949940 Chr.X: 47572054-47572054 on Build GRCh38

View Full Product Details