Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47624082-47624483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFP
Entrez Gene ID
5199
Gene Name
complement factor properdin
Gene Aliases
BFD, PFC, PFD, PROPERDIN
UniGene
Hs.53155
Gene Chromosome Location
Chr.X: 47624213-47630305 on Build GRCh38
Transcripts
NM_001145252.1 NM_002621.2 XM_017029575.1

SNP Detail

SNP ID Chromosome Location
rs752645314 Chr.X: 47624451-47624451 on Build GRCh38
rs766164302 Chr.X: 47624380-47624380 on Build GRCh38
rs746293085 Chr.X: 47624290-47624290 on Build GRCh38
rs373434543 Chr.X: 47624416-47624416 on Build GRCh38
rs776785115 Chr.X: 47624342-47624342 on Build GRCh38
rs767295636 Chr.X: 47624393-47624393 on Build GRCh38
rs371401758 Chr.X: 47624262-47624262 on Build GRCh38
rs367914849 Chr.X: 47624251-47624251 on Build GRCh38
rs759534525 Chr.X: 47624390-47624390 on Build GRCh38
rs1048118 Chr.X: 47624401-47624401 on Build GRCh38
rs138490907 Chr.X: 47624273-47624273 on Build GRCh38
rs769210799 Chr.X: 47624454-47624454 on Build GRCh38
rs370014336 Chr.X: 47624311-47624311 on Build GRCh38
rs1024583450 Chr.X: 47624445-47624445 on Build GRCh38
rs747581281 Chr.X: 47624319-47624319 on Build GRCh38
rs1051092806 Chr.X: 47624131-47624131 on Build GRCh38
rs137980601 Chr.X: 47624426-47624426 on Build GRCh38
rs777563861 Chr.X: 47624287-47624287 on Build GRCh38
rs879218443 Chr.X: 47624460-47624460 on Build GRCh38
rs765005275 Chr.X: 47624276-47624277 on Build GRCh38
rs934709132 Chr.X: 47624278-47624278 on Build GRCh38
rs55972826 Chr.X: 47624192-47624192 on Build GRCh38
rs751266279 Chr.X: 47624381-47624381 on Build GRCh38
rs960236393 Chr.X: 47624198-47624198 on Build GRCh38
rs58944244 Chr.X: 47624214-47624214 on Build GRCh38
rs768994510 Chr.X: 47624325-47624325 on Build GRCh38
rs8177081 Chr.X: 47624215-47624215 on Build GRCh38
rs773405705 Chr.X: 47624361-47624361 on Build GRCh38
rs978755234 Chr.X: 47624265-47624265 on Build GRCh38
rs752325590 Chr.X: 47624394-47624394 on Build GRCh38
rs765506323 Chr.X: 47624357-47624357 on Build GRCh38
rs758358839 Chr.X: 47624456-47624456 on Build GRCh38
rs754834825 Chr.X: 47624261-47624261 on Build GRCh38
rs1013228436 Chr.X: 47624427-47624427 on Build GRCh38
rs141133000 Chr.X: 47624353-47624353 on Build GRCh38
rs747277925 Chr.X: 47624434-47624434 on Build GRCh38
rs375725149 Chr.X: 47624268-47624268 on Build GRCh38
rs968983928 Chr.X: 47624260-47624260 on Build GRCh38
rs762270639 Chr.X: 47624372-47624372 on Build GRCh38

View Full Product Details