Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47983159-47983564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF182
Entrez Gene ID
7569
Gene Name
zinc finger protein 182
Gene Aliases
HHZ150, KOX14, ZNF21, Zfp182
UniGene
Hs.189690
Gene Chromosome Location
Chr.X: 47974851-48003995 on Build GRCh38
Transcripts
NM_001007088.1 NM_001178099.1 NM_006962.1

SNP Detail

SNP ID Chromosome Location
rs911442695 Chr.X: 47983519-47983519 on Build GRCh38
rs369374114 Chr.X: 47983221-47983221 on Build GRCh38
rs782079254 Chr.X: 47983321-47983321 on Build GRCh38
rs75509461 Chr.X: 47983307-47983307 on Build GRCh38
rs782544016 Chr.X: 47983441-47983441 on Build GRCh38
rs781858932 Chr.X: 47983364-47983364 on Build GRCh38
rs782331652 Chr.X: 47983277-47983277 on Build GRCh38
rs782441070 Chr.X: 47983414-47983414 on Build GRCh38
rs376661048 Chr.X: 47983425-47983425 on Build GRCh38
rs368847467 Chr.X: 47983434-47983434 on Build GRCh38
rs782586926 Chr.X: 47983424-47983424 on Build GRCh38
rs1022474963 Chr.X: 47983228-47983228 on Build GRCh38
rs782536079 Chr.X: 47983384-47983384 on Build GRCh38
rs782014270 Chr.X: 47983292-47983292 on Build GRCh38
rs782255303 Chr.X: 47983462-47983462 on Build GRCh38
rs894805922 Chr.X: 47983199-47983199 on Build GRCh38
rs184830327 Chr.X: 47983514-47983514 on Build GRCh38
rs149501545 Chr.X: 47983324-47983324 on Build GRCh38
rs6608756 Chr.X: 47983387-47983387 on Build GRCh38
rs781843178 Chr.X: 47983410-47983410 on Build GRCh38
rs782725171 Chr.X: 47983407-47983407 on Build GRCh38
rs61249955 Chr.X: 47983359-47983359 on Build GRCh38
rs782639244 Chr.X: 47983457-47983457 on Build GRCh38
rs782366047 Chr.X: 47983300-47983300 on Build GRCh38
rs146965366 Chr.X: 47983299-47983299 on Build GRCh38
rs782743069 Chr.X: 47983322-47983322 on Build GRCh38
rs371978166 Chr.X: 47983345-47983345 on Build GRCh38

View Full Product Details