Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48071554-48072059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF630
Entrez Gene ID
57232
Gene Name
zinc finger protein 630
Gene Aliases
dJ54B20.2
UniGene
Hs.650883
Gene Chromosome Location
Chr.X: 48058178-48071658 on Build GRCh38
Transcripts
NM_001190255.2 NM_001282201.1 NM_001282202.1

SNP Detail

SNP ID Chromosome Location
rs191458010 Chr.X: 48071710-48071710 on Build GRCh38
rs989113317 Chr.X: 48071912-48071912 on Build GRCh38
rs1040466701 Chr.X: 48071681-48071681 on Build GRCh38
rs913272645 Chr.X: 48071643-48071643 on Build GRCh38
rs1049555174 Chr.X: 48071744-48071744 on Build GRCh38
rs888236736 Chr.X: 48071748-48071748 on Build GRCh38
rs1005505000 Chr.X: 48071751-48071751 on Build GRCh38
rs976065174 Chr.X: 48071969-48071969 on Build GRCh38
rs782703068 Chr.X: 48071861-48071861 on Build GRCh38
rs781953411 Chr.X: 48071785-48071785 on Build GRCh38
rs782383948 Chr.X: 48071590-48071590 on Build GRCh38
rs996185936 Chr.X: 48071692-48071692 on Build GRCh38
rs932068977 Chr.X: 48072005-48072005 on Build GRCh38
rs781904200 Chr.X: 48071878-48071878 on Build GRCh38
rs781799077 Chr.X: 48071690-48071690 on Build GRCh38
rs187020743 Chr.X: 48071632-48071632 on Build GRCh38
rs1023952925 Chr.X: 48071833-48071833 on Build GRCh38
rs782580879 Chr.X: 48072024-48072024 on Build GRCh38
rs782183541 Chr.X: 48072029-48072029 on Build GRCh38
rs143872363 Chr.X: 48071800-48071800 on Build GRCh38
rs139782495 Chr.X: 48071993-48071993 on Build GRCh38
rs782264524 Chr.X: 48071741-48071741 on Build GRCh38
rs17332463 Chr.X: 48071612-48071612 on Build GRCh38
rs782425250 Chr.X: 48071592-48071592 on Build GRCh38
rs957770513 Chr.X: 48071873-48071873 on Build GRCh38
rs782033676 Chr.X: 48072007-48072007 on Build GRCh38
rs913463364 Chr.X: 48071943-48071943 on Build GRCh38
rs782782325 Chr.X: 48071651-48071651 on Build GRCh38
rs183419027 Chr.X: 48071837-48071837 on Build GRCh38
rs150629936 Chr.X: 48071865-48071865 on Build GRCh38
rs782369061 Chr.X: 48071763-48071763 on Build GRCh38
rs782522779 Chr.X: 48071863-48071863 on Build GRCh38
rs944750077 Chr.X: 48071966-48071966 on Build GRCh38

View Full Product Details