Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48902843-48903343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQBP1
Entrez Gene ID
10084
Gene Name
polyglutamine binding protein 1
Gene Aliases
MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
UniGene
Hs.534384
Gene Chromosome Location
Chr.X: 48897862-48903145 on Build GRCh38
Transcripts
NM_001032381.1 NM_001032382.1 NM_001032383.1 NM_001032384.1 NM_001167989.1 NM_001167990.1 NM_001167992.1 NM_005710.2 NM_144495.2 XM_005272571.3 XM_005272572.4 XM_011543884.2 XM_017029207.1 NM_001282647.1 NM_005660.2

Gene Symbol
SLC35A2
Entrez Gene ID
7355
Gene Name
solute carrier family 35 member A2
Gene Aliases
CDG2M, CDGX, UDP-Gal-Tr, UGALT, UGAT, UGT, UGT1, UGT2, UGTL
UniGene
Hs.21899
Gene Chromosome Location
Chr.X: 48903180-48911958 on Build GRCh38
Transcripts
NM_001032381.1 NM_001032382.1 NM_001032383.1 NM_001032384.1 NM_001167989.1 NM_001167990.1 NM_001167992.1 NM_005710.2 NM_144495.2 XM_005272571.3 XM_005272572.4 XM_011543884.2 XM_017029207.1 NM_001282647.1 NM_005660.2

SNP Detail

SNP ID Chromosome Location
rs947555647 Chr.X: 48903138-48903138 on Build GRCh38
rs782395941 Chr.X: 48903062-48903062 on Build GRCh38
rs1030376983 Chr.X: 48903313-48903313 on Build GRCh38
rs782596874 Chr.X: 48903014-48903014 on Build GRCh38
rs998932518 Chr.X: 48903251-48903251 on Build GRCh38
rs781855851 Chr.X: 48902985-48902985 on Build GRCh38
rs185473267 Chr.X: 48903258-48903258 on Build GRCh38
rs782373232 Chr.X: 48902922-48902922 on Build GRCh38
rs782569786 Chr.X: 48902958-48902958 on Build GRCh38
rs375045493 Chr.X: 48902925-48902925 on Build GRCh38
rs55729004 Chr.X: 48903182-48903182 on Build GRCh38
rs367752180 Chr.X: 48902976-48902976 on Build GRCh38
rs782361685 Chr.X: 48903187-48903187 on Build GRCh38
rs199778115 Chr.X: 48903081-48903081 on Build GRCh38
rs781921383 Chr.X: 48903232-48903232 on Build GRCh38
rs782011373 Chr.X: 48903172-48903172 on Build GRCh38
rs782015028 Chr.X: 48902919-48902919 on Build GRCh38
rs919654718 Chr.X: 48902913-48902913 on Build GRCh38
rs782092434 Chr.X: 48902933-48902933 on Build GRCh38
rs781932497 Chr.X: 48902880-48902880 on Build GRCh38
rs181088115 Chr.X: 48903090-48903090 on Build GRCh38
rs781987043 Chr.X: 48903064-48903064 on Build GRCh38
rs376272913 Chr.X: 48903092-48903092 on Build GRCh38
rs1057518357 Chr.X: 48903032-48903032 on Build GRCh38
rs782711223 Chr.X: 48903265-48903265 on Build GRCh38
rs782335530 Chr.X: 48902878-48902878 on Build GRCh38
rs112160560 Chr.X: 48903168-48903168 on Build GRCh38
rs782736516 Chr.X: 48902934-48902934 on Build GRCh38
rs145527627 Chr.X: 48902885-48902885 on Build GRCh38
rs782271597 Chr.X: 48903051-48903051 on Build GRCh38
rs903234425 Chr.X: 48903248-48903248 on Build GRCh38
rs782816951 Chr.X: 48903197-48903197 on Build GRCh38
rs781908587 Chr.X: 48903312-48903312 on Build GRCh38
rs781944808 Chr.X: 48903087-48903087 on Build GRCh38
rs782136545 Chr.X: 48902921-48902921 on Build GRCh38
rs782023194 Chr.X: 48903094-48903094 on Build GRCh38
rs370524032 Chr.X: 48902903-48902903 on Build GRCh38
rs782104904 Chr.X: 48903101-48903101 on Build GRCh38
rs782287803 Chr.X: 48902900-48902900 on Build GRCh38
rs782072163 Chr.X: 48903250-48903250 on Build GRCh38
rs782196111 Chr.X: 48903018-48903018 on Build GRCh38
rs782479139 Chr.X: 48902994-48902994 on Build GRCh38
rs973807964 Chr.X: 48902888-48902888 on Build GRCh38
rs878853145 Chr.X: 48903017-48903017 on Build GRCh38
rs782687130 Chr.X: 48903039-48903039 on Build GRCh38
rs781795216 Chr.X: 48902943-48902943 on Build GRCh38

View Full Product Details