Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49062361-49062765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC120
Entrez Gene ID
90060
Gene Name
coiled-coil domain containing 120
Gene Aliases
JM11
UniGene
Hs.522643
Gene Chromosome Location
Chr.X: 49053432-49069858 on Build GRCh38
Transcripts
NM_001163321.2 NM_001163322.2 NM_001163323.2 NM_001271835.1 NM_001271836.1 NM_033626.3

SNP Detail

SNP ID Chromosome Location
rs782304295 Chr.X: 49062527-49062527 on Build GRCh38
rs782463146 Chr.X: 49062515-49062515 on Build GRCh38
rs782038798 Chr.X: 49062537-49062537 on Build GRCh38
rs782006096 Chr.X: 49062485-49062485 on Build GRCh38
rs782746698 Chr.X: 49062616-49062616 on Build GRCh38
rs782816907 Chr.X: 49062494-49062494 on Build GRCh38
rs782124595 Chr.X: 49062492-49062492 on Build GRCh38
rs782564721 Chr.X: 49062641-49062641 on Build GRCh38
rs369880011 Chr.X: 49062516-49062516 on Build GRCh38
rs186417996 Chr.X: 49062452-49062452 on Build GRCh38
rs781936159 Chr.X: 49062460-49062460 on Build GRCh38
rs782754700 Chr.X: 49062481-49062481 on Build GRCh38
rs200695722 Chr.X: 49062628-49062628 on Build GRCh38
rs782799136 Chr.X: 49062630-49062630 on Build GRCh38
rs781888962 Chr.X: 49062631-49062631 on Build GRCh38
rs781852609 Chr.X: 49062395-49062395 on Build GRCh38
rs377375139 Chr.X: 49062530-49062530 on Build GRCh38
rs368479848 Chr.X: 49062714-49062714 on Build GRCh38
rs781800216 Chr.X: 49062657-49062657 on Build GRCh38
rs781936640 Chr.X: 49062659-49062659 on Build GRCh38
rs781840376 Chr.X: 49062514-49062514 on Build GRCh38
rs147084360 Chr.X: 49062510-49062510 on Build GRCh38
rs944667488 Chr.X: 49062721-49062721 on Build GRCh38
rs782293227 Chr.X: 49062439-49062439 on Build GRCh38
rs370700735 Chr.X: 49062579-49062579 on Build GRCh38
rs781869149 Chr.X: 49062618-49062618 on Build GRCh38
rs782002309 Chr.X: 49062531-49062531 on Build GRCh38
rs782671018 Chr.X: 49062559-49062559 on Build GRCh38
rs782039361 Chr.X: 49062479-49062479 on Build GRCh38
rs199599217 Chr.X: 49062509-49062509 on Build GRCh38
rs201911974 Chr.X: 49062578-49062578 on Build GRCh38
rs782447032 Chr.X: 49062638-49062638 on Build GRCh38
rs782101079 Chr.X: 49062612-49062612 on Build GRCh38
rs917481602 Chr.X: 49062473-49062473 on Build GRCh38
rs782238126 Chr.X: 49062517-49062517 on Build GRCh38
rs782018905 Chr.X: 49062502-49062502 on Build GRCh38
rs373303313 Chr.X: 49062520-49062520 on Build GRCh38
rs782429479 Chr.X: 49062671-49062671 on Build GRCh38
rs782550931 Chr.X: 49062519-49062519 on Build GRCh38
rs151098006 Chr.X: 49062529-49062529 on Build GRCh38
rs781925365 Chr.X: 49062604-49062604 on Build GRCh38

View Full Product Details