Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101553476-101553970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX1
Entrez Gene ID
51309
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
ALEX1, GASP7
UniGene
Hs.9728
Gene Chromosome Location
Chr.X: 101550531-101554700 on Build GRCh38
Transcripts
NM_016608.1

SNP Detail

SNP ID Chromosome Location
rs782561018 Chr.X: 101553518-101553518 on Build GRCh38
rs781958602 Chr.X: 101553577-101553577 on Build GRCh38
rs192601560 Chr.X: 101553557-101553557 on Build GRCh38
rs781936103 Chr.X: 101553787-101553787 on Build GRCh38
rs782543108 Chr.X: 101553529-101553529 on Build GRCh38
rs782024205 Chr.X: 101553795-101553795 on Build GRCh38
rs201162255 Chr.X: 101553608-101553608 on Build GRCh38
rs921594464 Chr.X: 101553815-101553815 on Build GRCh38
rs781882847 Chr.X: 101553660-101553660 on Build GRCh38
rs782427731 Chr.X: 101553792-101553792 on Build GRCh38
rs782773484 Chr.X: 101553620-101553620 on Build GRCh38
rs782134425 Chr.X: 101553611-101553611 on Build GRCh38
rs371723828 Chr.X: 101553527-101553527 on Build GRCh38
rs782645448 Chr.X: 101553545-101553545 on Build GRCh38
rs150765680 Chr.X: 101553659-101553659 on Build GRCh38
rs781964539 Chr.X: 101553624-101553624 on Build GRCh38
rs781817677 Chr.X: 101553499-101553499 on Build GRCh38
rs782631305 Chr.X: 101553685-101553685 on Build GRCh38
rs781824199 Chr.X: 101553697-101553697 on Build GRCh38
rs141214930 Chr.X: 101553599-101553599 on Build GRCh38
rs782445638 Chr.X: 101553703-101553703 on Build GRCh38
rs782019553 Chr.X: 101553601-101553601 on Build GRCh38
rs782561639 Chr.X: 101553714-101553714 on Build GRCh38
rs782229645 Chr.X: 101553769-101553769 on Build GRCh38
rs368530130 Chr.X: 101553563-101553563 on Build GRCh38
rs782707804 Chr.X: 101553898-101553898 on Build GRCh38
rs782161930 Chr.X: 101553654-101553654 on Build GRCh38
rs782211130 Chr.X: 101553761-101553761 on Build GRCh38
rs781849892 Chr.X: 101553830-101553830 on Build GRCh38
rs782332356 Chr.X: 101553786-101553786 on Build GRCh38
rs782039307 Chr.X: 101553788-101553788 on Build GRCh38
rs782701859 Chr.X: 101553638-101553638 on Build GRCh38
rs782141841 Chr.X: 101553825-101553825 on Build GRCh38
rs183932643 Chr.X: 101553737-101553737 on Build GRCh38
rs782083089 Chr.X: 101553836-101553836 on Build GRCh38
rs368457888 Chr.X: 101553652-101553652 on Build GRCh38
rs782392131 Chr.X: 101553754-101553754 on Build GRCh38
rs782444968 Chr.X: 101553512-101553512 on Build GRCh38
rs376307003 Chr.X: 101553629-101553629 on Build GRCh38

View Full Product Details